Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1234102-1234595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBARP
Entrez Gene ID
255057
Gene Name
CACN beta subunit associated regulatory protein
Gene Aliases
BARP, C19orf26, DOS
UniGene
Hs.346575
Gene Chromosome Location
Chr.19: 1228287-1238512 on Build GRCh38
Transcripts
NM_152769.2 XM_017026555.1 XM_017026556.1 XM_017026557.1 XM_017026558.1 XM_017026559.1 XM_017026560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4833740 c.673C>G p.L225V Chr.19: 1234585-1234585 on Build GRCh38
4667138 c.686C>T p.A229V Chr.19: 1234255-1234255 on Build GRCh38
5891531 c.766G>A p.G256S Chr.19: 1234253-1234253 on Build GRCh38
4784977 c.723C>T p.G241G Chr.19: 1234296-1234296 on Build GRCh38
7315231 c.721G>T p.A241S Chr.19: 1234238-1234238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895168883 Chr.19: 1234458-1234458 on Build GRCh38
rs771389816 Chr.19: 1234542-1234542 on Build GRCh38
rs761599394 Chr.19: 1234296-1234296 on Build GRCh38
rs1009437048 Chr.19: 1234506-1234506 on Build GRCh38
rs775044212 Chr.19: 1234202-1234202 on Build GRCh38
rs773074379 Chr.19: 1234554-1234554 on Build GRCh38
rs974951257 Chr.19: 1234291-1234291 on Build GRCh38
rs1029114455 Chr.19: 1234327-1234327 on Build GRCh38
rs990335354 Chr.19: 1234356-1234356 on Build GRCh38
rs752929249 Chr.19: 1234536-1234536 on Build GRCh38
rs1047920248 Chr.19: 1234452-1234452 on Build GRCh38
rs114502625 Chr.19: 1234533-1234533 on Build GRCh38
rs912960055 Chr.19: 1234377-1234377 on Build GRCh38
rs58142912 Chr.19: 1234256-1234256 on Build GRCh38
rs763814316 Chr.19: 1234205-1234205 on Build GRCh38
rs928436466 Chr.19: 1234393-1234393 on Build GRCh38
rs149047560 Chr.19: 1234192-1234192 on Build GRCh38
rs534134247 Chr.19: 1234155-1234155 on Build GRCh38
rs769114052 Chr.19: 1234257-1234257 on Build GRCh38
rs779028431 Chr.19: 1234153-1234153 on Build GRCh38
rs375306397 Chr.19: 1234204-1234204 on Build GRCh38
rs929513023 Chr.19: 1234422-1234422 on Build GRCh38
rs144699048 Chr.19: 1234211-1234211 on Build GRCh38
rs866563653 Chr.19: 1234248-1234248 on Build GRCh38
rs777189721 Chr.19: 1234286-1234286 on Build GRCh38
rs1005073105 Chr.19: 1234267-1234267 on Build GRCh38
rs200987649 Chr.19: 1234158-1234158 on Build GRCh38
rs1051881459 Chr.19: 1234497-1234497 on Build GRCh38
rs565193202 Chr.19: 1234351-1234351 on Build GRCh38
rs900522193 Chr.19: 1234466-1234466 on Build GRCh38
rs747124011 Chr.19: 1234236-1234236 on Build GRCh38
rs1055632215 Chr.19: 1234430-1234430 on Build GRCh38
rs765120631 Chr.19: 1234306-1234306 on Build GRCh38
rs762870808 Chr.19: 1234566-1234566 on Build GRCh38
rs754691361 Chr.19: 1234547-1234547 on Build GRCh38
rs895148318 Chr.19: 1234226-1234226 on Build GRCh38
rs193226217 Chr.19: 1234162-1234162 on Build GRCh38
rs902008966 Chr.19: 1234516-1234516 on Build GRCh38
rs371405447 Chr.19: 1234249-1234249 on Build GRCh38
rs771481996 Chr.19: 1234271-1234271 on Build GRCh38
rs373046882 Chr.19: 1234540-1234540 on Build GRCh38
rs983165204 Chr.19: 1234568-1234568 on Build GRCh38
rs372344599 Chr.19: 1234177-1234177 on Build GRCh38
rs755828707 Chr.19: 1234147-1234147 on Build GRCh38
rs989912497 Chr.19: 1234164-1234164 on Build GRCh38
rs917642850 Chr.19: 1234403-1234403 on Build GRCh38
rs565281735 Chr.19: 1234468-1234468 on Build GRCh38
rs762474879 Chr.19: 1234510-1234510 on Build GRCh38
rs10414700 Chr.19: 1234189-1234189 on Build GRCh38
rs766178161 Chr.19: 1234573-1234573 on Build GRCh38
rs566704788 Chr.19: 1234499-1234499 on Build GRCh38
rs375352813 Chr.19: 1234152-1234152 on Build GRCh38
rs143104137 Chr.19: 1234389-1234389 on Build GRCh38
rs558899960 Chr.19: 1234519-1234519 on Build GRCh38
rs950510276 Chr.19: 1234562-1234562 on Build GRCh38
rs935751884 Chr.19: 1234163-1234163 on Build GRCh38
rs766981286 Chr.19: 1234194-1234194 on Build GRCh38
rs761546307 Chr.19: 1234550-1234550 on Build GRCh38
rs765854923 Chr.19: 1234532-1234532 on Build GRCh38
rs369637626 Chr.19: 1234538-1234538 on Build GRCh38
rs758531324 Chr.19: 1234157-1234157 on Build GRCh38
rs1009798419 Chr.19: 1234289-1234289 on Build GRCh38
rs540573275 Chr.19: 1234311-1234311 on Build GRCh38
rs766497164 Chr.19: 1234229-1234231 on Build GRCh38
rs758478510 Chr.19: 1234221-1234221 on Build GRCh38
rs763789967 Chr.19: 1234149-1234149 on Build GRCh38
rs368354333 Chr.19: 1234176-1234176 on Build GRCh38
rs547964674 Chr.19: 1234496-1234496 on Build GRCh38
rs750303420 Chr.19: 1234314-1234314 on Build GRCh38
rs376983695 Chr.19: 1234151-1234151 on Build GRCh38
rs778145931 Chr.19: 1234254-1234254 on Build GRCh38
rs975685235 Chr.19: 1234170-1234170 on Build GRCh38
rs114112602 Chr.19: 1234382-1234382 on Build GRCh38
rs532628028 Chr.19: 1234501-1234502 on Build GRCh38
rs776328551 Chr.19: 1234287-1234287 on Build GRCh38
rs542734546 Chr.19: 1234238-1234238 on Build GRCh38
rs753541878 Chr.19: 1234206-1234206 on Build GRCh38
rs776800497 Chr.19: 1234172-1234172 on Build GRCh38
rs750354718 Chr.19: 1234212-1234212 on Build GRCh38
rs529829090 Chr.19: 1234470-1234470 on Build GRCh38
rs112181417 Chr.19: 1234479-1234479 on Build GRCh38
rs776071664 Chr.19: 1234548-1234548 on Build GRCh38
rs751552463 Chr.19: 1234527-1234527 on Build GRCh38
rs765770219 Chr.19: 1234465-1234465 on Build GRCh38
rs376111449 Chr.19: 1234159-1234159 on Build GRCh38
rs781508045 Chr.19: 1234239-1234239 on Build GRCh38
rs533926715 Chr.19: 1234509-1234509 on Build GRCh38
rs942267021 Chr.19: 1234462-1234462 on Build GRCh38
rs1045644718 Chr.19: 1234184-1234184 on Build GRCh38
rs184411443 Chr.19: 1234250-1234250 on Build GRCh38
rs774799579 Chr.19: 1234272-1234272 on Build GRCh38
rs1020867368 Chr.19: 1234310-1234310 on Build GRCh38
rs991937369 Chr.19: 1234399-1234399 on Build GRCh38
rs746415022 Chr.19: 1234545-1234545 on Build GRCh38
rs769584644 Chr.19: 1234552-1234552 on Build GRCh38
rs1000758309 Chr.19: 1234522-1234522 on Build GRCh38
rs779500133 Chr.19: 1234543-1234543 on Build GRCh38
rs368975309 Chr.19: 1234326-1234326 on Build GRCh38
rs1029233377 Chr.19: 1234520-1234520 on Build GRCh38
rs778094622 Chr.19: 1234539-1234539 on Build GRCh38
rs367906910 Chr.19: 1234222-1234222 on Build GRCh38
rs957052614 Chr.19: 1234345-1234345 on Build GRCh38
rs75522877 Chr.19: 1234398-1234398 on Build GRCh38
rs757102508 Chr.19: 1234208-1234208 on Build GRCh38
rs773726822 Chr.19: 1234188-1234188 on Build GRCh38
rs1039262274 Chr.19: 1234463-1234463 on Build GRCh38

View Full Product Details