Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2189642-2190146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOT1L
Entrez Gene ID
84444
Gene Name
DOT1 like histone lysine methyltransferase
Gene Aliases
DOT1, KMT4
UniGene
Hs.713641
Gene Chromosome Location
Chr.19: 2163963-2232578 on Build GRCh38
Transcripts
NM_032482.2 XM_005259659.3 XM_005259660.3 XM_011528359.2 XM_011528360.1 XM_017027366.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1589919 c.202T>C p.F68L Chr.19: 2189733-2189733 on Build GRCh38
7244135 c.219G>A p.R73R Chr.19: 2189750-2189750 on Build GRCh38
6648709 c.225C>T p.C75C Chr.19: 2189756-2189756 on Build GRCh38
993706 c.202T>C p.F68L Chr.19: 2189733-2189733 on Build GRCh38
3103069 c.204C>T p.F68F Chr.19: 2189735-2189735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775691456 Chr.19: 2189685-2189685 on Build GRCh38
rs747205311 Chr.19: 2189833-2189838 on Build GRCh38
rs199909162 Chr.19: 2189756-2189756 on Build GRCh38
rs182174397 Chr.19: 2190043-2190043 on Build GRCh38
rs537079884 Chr.19: 2189678-2189678 on Build GRCh38
rs369044072 Chr.19: 2189843-2189843 on Build GRCh38
rs771739927 Chr.19: 2189915-2189916 on Build GRCh38
rs764139743 Chr.19: 2189716-2189716 on Build GRCh38
rs913160679 Chr.19: 2189665-2189665 on Build GRCh38
rs757442864 Chr.19: 2189723-2189723 on Build GRCh38
rs777157799 Chr.19: 2189777-2189777 on Build GRCh38
rs1020534490 Chr.19: 2190076-2190076 on Build GRCh38
rs74384750 Chr.19: 2190098-2190098 on Build GRCh38
rs994705028 Chr.19: 2190032-2190032 on Build GRCh38
rs568151989 Chr.19: 2189987-2189987 on Build GRCh38
rs751426391 Chr.19: 2189803-2189803 on Build GRCh38
rs958675088 Chr.19: 2190123-2190123 on Build GRCh38
rs1054421602 Chr.19: 2189933-2189933 on Build GRCh38
rs888708392 Chr.19: 2189724-2189724 on Build GRCh38
rs1047683473 Chr.19: 2189956-2189956 on Build GRCh38
rs150316325 Chr.19: 2189750-2189750 on Build GRCh38
rs779102020 Chr.19: 2189729-2189729 on Build GRCh38
rs992800502 Chr.19: 2190124-2190124 on Build GRCh38
rs755870956 Chr.19: 2189711-2189711 on Build GRCh38
rs989066425 Chr.19: 2189809-2189809 on Build GRCh38
rs968614827 Chr.19: 2190086-2190086 on Build GRCh38
rs1057124392 Chr.19: 2189922-2189922 on Build GRCh38
rs771719316 Chr.19: 2189965-2189965 on Build GRCh38
rs769020592 Chr.19: 2189765-2189765 on Build GRCh38
rs1050112814 Chr.19: 2189677-2189677 on Build GRCh38
rs773332079 Chr.19: 2189820-2189820 on Build GRCh38
rs900154168 Chr.19: 2189990-2189990 on Build GRCh38
rs935478625 Chr.19: 2190122-2190122 on Build GRCh38
rs758450615 Chr.19: 2189835-2189835 on Build GRCh38
rs529330428 Chr.19: 2189877-2189877 on Build GRCh38
rs746397721 Chr.19: 2190008-2190008 on Build GRCh38
rs985759511 Chr.19: 2190060-2190060 on Build GRCh38
rs769051965 Chr.19: 2189691-2189691 on Build GRCh38
rs186572260 Chr.19: 2189699-2189699 on Build GRCh38
rs1041233088 Chr.19: 2189695-2189695 on Build GRCh38
rs79010603 Chr.19: 2189858-2189858 on Build GRCh38
rs559585222 Chr.19: 2189923-2189923 on Build GRCh38
rs368740625 Chr.19: 2189707-2189707 on Build GRCh38
rs759089252 Chr.19: 2189702-2189702 on Build GRCh38
rs917348715 Chr.19: 2189924-2189924 on Build GRCh38
rs376001115 Chr.19: 2189808-2189808 on Build GRCh38
rs962157637 Chr.19: 2190027-2190027 on Build GRCh38
rs935528624 Chr.19: 2189932-2189932 on Build GRCh38
rs760407860 Chr.19: 2189824-2189824 on Build GRCh38
rs144956568 Chr.19: 2189979-2189979 on Build GRCh38
rs370257976 Chr.19: 2189697-2189697 on Build GRCh38
rs779217633 Chr.19: 2189989-2189989 on Build GRCh38
rs954624534 Chr.19: 2190038-2190038 on Build GRCh38
rs1043460223 Chr.19: 2189689-2189689 on Build GRCh38
rs763939653 Chr.19: 2189825-2189825 on Build GRCh38
rs888663787 Chr.19: 2189974-2189974 on Build GRCh38
rs537375770 Chr.19: 2190007-2190007 on Build GRCh38
rs548658346 Chr.19: 2190039-2190039 on Build GRCh38
rs201346710 Chr.19: 2189875-2189875 on Build GRCh38
rs191141666 Chr.19: 2189802-2189802 on Build GRCh38
rs924401011 Chr.19: 2190113-2190113 on Build GRCh38
rs1015016205 Chr.19: 2190016-2190016 on Build GRCh38
rs978741462 Chr.19: 2189862-2189862 on Build GRCh38
rs924072599 Chr.19: 2189925-2189925 on Build GRCh38
rs371636480 Chr.19: 2189783-2189783 on Build GRCh38
rs372161368 Chr.19: 2189735-2189735 on Build GRCh38
rs757236929 Chr.19: 2189827-2189827 on Build GRCh38
rs569995120 Chr.19: 2190005-2190005 on Build GRCh38
rs555421971 Chr.19: 2189681-2189681 on Build GRCh38
rs534414260 Chr.19: 2190075-2190075 on Build GRCh38
rs370237255 Chr.19: 2189914-2189914 on Build GRCh38
rs977547771 Chr.19: 2190112-2190112 on Build GRCh38
rs753779025 Chr.19: 2189717-2189717 on Build GRCh38
rs200627822 Chr.19: 2189804-2189804 on Build GRCh38
rs765461750 Chr.19: 2189828-2189828 on Build GRCh38
rs758532148 Chr.19: 2189736-2189736 on Build GRCh38
rs367786294 Chr.19: 2189700-2189700 on Build GRCh38
rs771205855 Chr.19: 2189841-2189843 on Build GRCh38
rs970829295 Chr.19: 2189789-2189789 on Build GRCh38
rs945262129 Chr.19: 2190109-2190109 on Build GRCh38
rs552688015 Chr.19: 2190087-2190087 on Build GRCh38
rs912396374 Chr.19: 2190108-2190108 on Build GRCh38
rs930131693 Chr.19: 2189952-2189952 on Build GRCh38
rs543423366 Chr.19: 2189864-2189864 on Build GRCh38
rs750461832 Chr.19: 2189831-2189831 on Build GRCh38
rs761298039 Chr.19: 2189687-2189687 on Build GRCh38
rs762475482 Chr.19: 2189698-2189698 on Build GRCh38
rs376425351 Chr.19: 2189844-2189844 on Build GRCh38
rs775148921 Chr.19: 2189823-2189823 on Build GRCh38
rs535643516 Chr.19: 2190011-2190011 on Build GRCh38

View Full Product Details