Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2206624-2207120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOT1L
Entrez Gene ID
84444
Gene Name
DOT1 like histone lysine methyltransferase
Gene Aliases
DOT1, KMT4
UniGene
Hs.713641
Gene Chromosome Location
Chr.19: 2163963-2232578 on Build GRCh38
Transcripts
NM_032482.2 XM_005259659.3 XM_005259660.3 XM_011528359.2 XM_011528360.1 XM_011528361.2 XM_017027366.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6950199 c.813_814insT p.A273fs*10 Chr.19: 2206754-2206755 on Build GRCh38
6969444 c.829_837delTTCAGAATA p.F277_I279delFRI Chr.19: 2206770-2206778 on Build GRCh38
5608302 c.804C>T p.S268S Chr.19: 2206745-2206745 on Build GRCh38
6577809 c.851T>G p.L284W Chr.19: 2206792-2206792 on Build GRCh38
6912133 c.812C>G p.P271R Chr.19: 2206753-2206753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902188119 Chr.19: 2207055-2207055 on Build GRCh38
rs58189866 Chr.19: 2207054-2207054 on Build GRCh38
rs942790805 Chr.19: 2207089-2207089 on Build GRCh38
rs778170732 Chr.19: 2206822-2206822 on Build GRCh38
rs990653269 Chr.19: 2206953-2206953 on Build GRCh38
rs763128974 Chr.19: 2206718-2206718 on Build GRCh38
rs917816382 Chr.19: 2206974-2206974 on Build GRCh38
rs760116051 Chr.19: 2206661-2206661 on Build GRCh38
rs201441854 Chr.19: 2206739-2206739 on Build GRCh38
rs201346501 Chr.19: 2206699-2206699 on Build GRCh38
rs368904514 Chr.19: 2207018-2207018 on Build GRCh38
rs966463955 Chr.19: 2206916-2206916 on Build GRCh38
rs1031372847 Chr.19: 2206997-2206997 on Build GRCh38
rs3746161 Chr.19: 2207006-2207006 on Build GRCh38
rs556517461 Chr.19: 2207022-2207022 on Build GRCh38
rs766071484 Chr.19: 2206690-2206691 on Build GRCh38
rs915023528 Chr.19: 2206833-2206833 on Build GRCh38
rs866305655 Chr.19: 2206653-2206653 on Build GRCh38
rs1001028322 Chr.19: 2206924-2206924 on Build GRCh38
rs1021943115 Chr.19: 2207014-2207014 on Build GRCh38
rs942122014 Chr.19: 2206796-2206796 on Build GRCh38
rs746714161 Chr.19: 2206828-2206828 on Build GRCh38
rs778598047 Chr.19: 2206812-2206814 on Build GRCh38
rs532542990 Chr.19: 2207011-2207011 on Build GRCh38
rs201645007 Chr.19: 2206762-2206762 on Build GRCh38
rs1008340404 Chr.19: 2206662-2206662 on Build GRCh38
rs1057494372 Chr.19: 2206872-2206872 on Build GRCh38
rs1051955712 Chr.19: 2207021-2207021 on Build GRCh38
rs189294966 Chr.19: 2207065-2207065 on Build GRCh38
rs372927694 Chr.19: 2206764-2206764 on Build GRCh38
rs770463736 Chr.19: 2206913-2206913 on Build GRCh38
rs959192437 Chr.19: 2206951-2206951 on Build GRCh38
rs71337090 Chr.19: 2206842-2206842 on Build GRCh38
rs907884288 Chr.19: 2206983-2206983 on Build GRCh38
rs200310379 Chr.19: 2206811-2206811 on Build GRCh38
rs745513425 Chr.19: 2207017-2207017 on Build GRCh38
rs527849544 Chr.19: 2206876-2206876 on Build GRCh38
rs764682973 Chr.19: 2206806-2206806 on Build GRCh38
rs752659817 Chr.19: 2206818-2206820 on Build GRCh38
rs763429734 Chr.19: 2207002-2207002 on Build GRCh38
rs7507579 Chr.19: 2206860-2206860 on Build GRCh38
rs778072754 Chr.19: 2206679-2206679 on Build GRCh38
rs748224363 Chr.19: 2206694-2206694 on Build GRCh38
rs1020698386 Chr.19: 2206668-2206668 on Build GRCh38
rs369397123 Chr.19: 2206697-2206697 on Build GRCh38
rs199768042 Chr.19: 2206748-2206748 on Build GRCh38
rs749596107 Chr.19: 2206871-2206871 on Build GRCh38
rs971082223 Chr.19: 2206976-2206976 on Build GRCh38
rs545098955 Chr.19: 2207043-2207043 on Build GRCh38
rs927415348 Chr.19: 2206853-2206853 on Build GRCh38
rs143646595 Chr.19: 2207071-2207071 on Build GRCh38
rs1006902270 Chr.19: 2206919-2206919 on Build GRCh38
rs371116678 Chr.19: 2206707-2206707 on Build GRCh38
rs552353831 Chr.19: 2206940-2206940 on Build GRCh38
rs147017323 Chr.19: 2206995-2206995 on Build GRCh38
rs1021054743 Chr.19: 2206897-2206897 on Build GRCh38
rs879097034 Chr.19: 2206921-2206921 on Build GRCh38
rs1007873824 Chr.19: 2206895-2206895 on Build GRCh38
rs1032104419 Chr.19: 2206931-2206931 on Build GRCh38
rs141421870 Chr.19: 2206854-2206854 on Build GRCh38
rs3746160 Chr.19: 2207042-2207042 on Build GRCh38
rs545452591 Chr.19: 2206831-2206831 on Build GRCh38
rs938888033 Chr.19: 2207048-2207048 on Build GRCh38
rs993003667 Chr.19: 2207050-2207050 on Build GRCh38
rs775927698 Chr.19: 2206952-2206952 on Build GRCh38
rs888549385 Chr.19: 2206909-2206909 on Build GRCh38
rs367900456 Chr.19: 2206696-2206696 on Build GRCh38
rs1032049566 Chr.19: 2207066-2207066 on Build GRCh38
rs980282701 Chr.19: 2206845-2206845 on Build GRCh38
rs184925366 Chr.19: 2206996-2206996 on Build GRCh38
rs756594301 Chr.19: 2206733-2206733 on Build GRCh38
rs1018071450 Chr.19: 2206968-2206968 on Build GRCh38
rs754764341 Chr.19: 2206809-2206810 on Build GRCh38
rs745394926 Chr.19: 2206683-2206683 on Build GRCh38
rs779685711 Chr.19: 2206823-2206823 on Build GRCh38
rs1012023414 Chr.19: 2207075-2207075 on Build GRCh38
rs753439756 Chr.19: 2206806-2206807 on Build GRCh38
rs758017919 Chr.19: 2206812-2206812 on Build GRCh38
rs574936318 Chr.19: 2206807-2206807 on Build GRCh38
rs983441841 Chr.19: 2206979-2206979 on Build GRCh38
rs191575628 Chr.19: 2207081-2207081 on Build GRCh38
rs780653744 Chr.19: 2206835-2206835 on Build GRCh38
rs753107452 Chr.19: 2206760-2206760 on Build GRCh38
rs757932091 Chr.19: 2206685-2206685 on Build GRCh38
rs890741050 Chr.19: 2206894-2206894 on Build GRCh38
rs375736617 Chr.19: 2206808-2206808 on Build GRCh38

View Full Product Details