Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3939658-3940065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NMRK2
Entrez Gene ID
27231
Gene Name
nicotinamide riboside kinase 2
Gene Aliases
ITGB1BP3, MIBP, NRK2
UniGene
Hs.135458
Gene Chromosome Location
Chr.19: 3933103-3942416 on Build GRCh38
Transcripts
NM_001289117.1 NM_170678.2 NR_110316.1 XM_006722725.3 XM_006722726.3 XM_017026597.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6273313 c.395+9G>T p.? Chr.19: 3939980-3939980 on Build GRCh38
1680521 c.346C>T p.R116C Chr.19: 3939922-3939922 on Build GRCh38
996190 c.369G>T p.P123P Chr.19: 3939945-3939945 on Build GRCh38
6053823 c.351G>A p.R117R Chr.19: 3939927-3939927 on Build GRCh38
4404019 c.366C>T p.V122V Chr.19: 3939942-3939942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544946486 Chr.19: 3939863-3939864 on Build GRCh38
rs763706235 Chr.19: 3939849-3939849 on Build GRCh38
rs1037292039 Chr.19: 3940040-3940040 on Build GRCh38
rs527662906 Chr.19: 3939752-3939752 on Build GRCh38
rs879659954 Chr.19: 3940032-3940032 on Build GRCh38
rs140113877 Chr.19: 3939926-3939926 on Build GRCh38
rs575881344 Chr.19: 3939730-3939730 on Build GRCh38
rs367875659 Chr.19: 3939939-3939939 on Build GRCh38
rs749363799 Chr.19: 3940003-3940003 on Build GRCh38
rs762210846 Chr.19: 3939925-3939925 on Build GRCh38
rs756309223 Chr.19: 3940010-3940010 on Build GRCh38
rs746483461 Chr.19: 3939980-3939980 on Build GRCh38
rs539348131 Chr.19: 3939959-3939959 on Build GRCh38
rs144400706 Chr.19: 3939923-3939923 on Build GRCh38
rs892808269 Chr.19: 3940026-3940026 on Build GRCh38
rs757731623 Chr.19: 3939857-3939857 on Build GRCh38
rs902963358 Chr.19: 3939861-3939861 on Build GRCh38
rs34544544 Chr.19: 3939688-3939689 on Build GRCh38
rs775325451 Chr.19: 3939962-3939962 on Build GRCh38
rs759589757 Chr.19: 3939995-3939995 on Build GRCh38
rs752829863 Chr.19: 3940009-3940009 on Build GRCh38
rs372177012 Chr.19: 3939870-3939870 on Build GRCh38
rs756025963 Chr.19: 3939949-3939950 on Build GRCh38
rs1030133391 Chr.19: 3939832-3939832 on Build GRCh38
rs1035054556 Chr.19: 3939778-3939778 on Build GRCh38
rs972502783 Chr.19: 3939747-3939747 on Build GRCh38
rs556081033 Chr.19: 3939991-3939991 on Build GRCh38
rs1054100987 Chr.19: 3940020-3940020 on Build GRCh38
rs4806966 Chr.19: 3939705-3939705 on Build GRCh38
rs200154352 Chr.19: 3939888-3939888 on Build GRCh38
rs982222745 Chr.19: 3939716-3939716 on Build GRCh38
rs954702373 Chr.19: 3939841-3939841 on Build GRCh38
rs775044733 Chr.19: 3939900-3939900 on Build GRCh38
rs564732253 Chr.19: 3939846-3939846 on Build GRCh38
rs556342655 Chr.19: 3939956-3939956 on Build GRCh38
rs747560231 Chr.19: 3939685-3939685 on Build GRCh38
rs531686734 Chr.19: 3939821-3939821 on Build GRCh38
rs570369559 Chr.19: 3939949-3939949 on Build GRCh38
rs776281746 Chr.19: 3939985-3939985 on Build GRCh38
rs763766121 Chr.19: 3939922-3939922 on Build GRCh38
rs373037496 Chr.19: 3939983-3939983 on Build GRCh38
rs919892927 Chr.19: 3939964-3939964 on Build GRCh38
rs541950755 Chr.19: 3939731-3939731 on Build GRCh38
rs766859762 Chr.19: 3939993-3939993 on Build GRCh38
rs1012765883 Chr.19: 3939706-3939706 on Build GRCh38
rs763435803 Chr.19: 3939856-3939856 on Build GRCh38
rs781300477 Chr.19: 3939885-3939885 on Build GRCh38
rs144659918 Chr.19: 3939940-3939940 on Build GRCh38
rs759141138 Chr.19: 3939942-3939942 on Build GRCh38
rs113469967 Chr.19: 3940027-3940027 on Build GRCh38
rs766236376 Chr.19: 3939954-3939954 on Build GRCh38
rs910845544 Chr.19: 3939965-3939965 on Build GRCh38
rs760146517 Chr.19: 3939915-3939915 on Build GRCh38
rs4807536 Chr.19: 3939930-3939930 on Build GRCh38
rs369553268 Chr.19: 3939945-3939945 on Build GRCh38
rs1003088140 Chr.19: 3939768-3939769 on Build GRCh38
rs962145208 Chr.19: 3939745-3939745 on Build GRCh38
rs920576931 Chr.19: 3939782-3939782 on Build GRCh38
rs1056038411 Chr.19: 3939886-3939886 on Build GRCh38
rs977185095 Chr.19: 3939828-3939828 on Build GRCh38
rs747105897 Chr.19: 3939878-3939878 on Build GRCh38
rs149777713 Chr.19: 3939880-3939880 on Build GRCh38
rs754203086 Chr.19: 3939851-3939851 on Build GRCh38
rs8107013 Chr.19: 3939714-3939714 on Build GRCh38
rs533521180 Chr.19: 3939944-3939944 on Build GRCh38
rs562246434 Chr.19: 3939750-3939750 on Build GRCh38
rs930900936 Chr.19: 3939802-3939802 on Build GRCh38
rs139753247 Chr.19: 3939825-3939825 on Build GRCh38
rs761554532 Chr.19: 3939987-3939987 on Build GRCh38
rs745797871 Chr.19: 3939893-3939893 on Build GRCh38
rs769617345 Chr.19: 3939989-3939989 on Build GRCh38
rs998964231 Chr.19: 3939684-3939684 on Build GRCh38
rs377372767 Chr.19: 3940031-3940031 on Build GRCh38
rs748370713 Chr.19: 3939947-3939947 on Build GRCh38
rs115477365 Chr.19: 3939994-3939994 on Build GRCh38
rs377251978 Chr.19: 3940028-3940028 on Build GRCh38
rs113983965 Chr.19: 3939877-3939877 on Build GRCh38
rs111903284 Chr.19: 3939948-3939948 on Build GRCh38
rs765790768 Chr.19: 3939865-3939865 on Build GRCh38
rs944967433 Chr.19: 3939977-3939977 on Build GRCh38
rs150650227 Chr.19: 3939711-3939711 on Build GRCh38
rs772058188 Chr.19: 3939894-3939894 on Build GRCh38
rs780139139 Chr.19: 3939962-3939964 on Build GRCh38
rs368814465 Chr.19: 3939848-3939848 on Build GRCh38
rs910730893 Chr.19: 3939826-3939826 on Build GRCh38
rs767487870 Chr.19: 3940007-3940007 on Build GRCh38

View Full Product Details