Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5727246-5727782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPERD
Entrez Gene ID
257062
Gene Name
cation channel sperm associated auxiliary subunit delta
Gene Aliases
TMEM146
UniGene
Hs.631842
Gene Chromosome Location
Chr.19: 5720385-5778734 on Build GRCh38
Transcripts
NM_152784.3 XM_011527885.2 XM_011527886.2 XM_011527887.2 XM_011527888.2 XM_011527889.2 XM_011527890.2 XM_017026564.1 XM_017026565.1 XM_017026566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1243688 c.197A>G p.Y66C Chr.19: 5727338-5727338 on Build GRCh38
1165362 c.131G>A p.R44H Chr.19: 5727272-5727272 on Build GRCh38
6869220 c.195A>G p.L65L Chr.19: 5727336-5727336 on Build GRCh38
6620810 c.177C>T p.C59C Chr.19: 5727318-5727318 on Build GRCh38
3371522 c.146C>A p.P49H Chr.19: 5727287-5727287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775178619 Chr.19: 5727299-5727299 on Build GRCh38
rs773678262 Chr.19: 5727372-5727372 on Build GRCh38
rs749282599 Chr.19: 5727284-5727284 on Build GRCh38
rs5826899 Chr.19: 5727403-5727404 on Build GRCh38
rs1040475849 Chr.19: 5727663-5727663 on Build GRCh38
rs535783541 Chr.19: 5727746-5727746 on Build GRCh38
rs746455349 Chr.19: 5727301-5727301 on Build GRCh38
rs778749794 Chr.19: 5727296-5727296 on Build GRCh38
rs938485724 Chr.19: 5727616-5727616 on Build GRCh38
rs145553037 Chr.19: 5727521-5727521 on Build GRCh38
rs901165517 Chr.19: 5727704-5727704 on Build GRCh38
rs888007594 Chr.19: 5727438-5727438 on Build GRCh38
rs942914478 Chr.19: 5727665-5727665 on Build GRCh38
rs1015306299 Chr.19: 5727491-5727491 on Build GRCh38
rs968579875 Chr.19: 5727535-5727535 on Build GRCh38
rs998164923 Chr.19: 5727720-5727720 on Build GRCh38
rs1050526592 Chr.19: 5727673-5727673 on Build GRCh38
rs945749135 Chr.19: 5727274-5727274 on Build GRCh38
rs187902259 Chr.19: 5727370-5727370 on Build GRCh38
rs918308087 Chr.19: 5727620-5727620 on Build GRCh38
rs892530437 Chr.19: 5727756-5727756 on Build GRCh38
rs78781569 Chr.19: 5727298-5727298 on Build GRCh38
rs913180671 Chr.19: 5727614-5727614 on Build GRCh38
rs73544733 Chr.19: 5727757-5727757 on Build GRCh38
rs1039729825 Chr.19: 5727678-5727678 on Build GRCh38
rs1031508508 Chr.19: 5727728-5727728 on Build GRCh38
rs1006844288 Chr.19: 5727694-5727694 on Build GRCh38
rs182825334 Chr.19: 5727272-5727272 on Build GRCh38
rs756765868 Chr.19: 5727484-5727485 on Build GRCh38
rs888713699 Chr.19: 5727644-5727644 on Build GRCh38
rs1006443041 Chr.19: 5727444-5727444 on Build GRCh38
rs969489965 Chr.19: 5727516-5727516 on Build GRCh38
rs998658069 Chr.19: 5727486-5727486 on Build GRCh38
rs201332713 Chr.19: 5727400-5727401 on Build GRCh38
rs762452547 Chr.19: 5727690-5727690 on Build GRCh38
rs962071448 Chr.19: 5727503-5727503 on Build GRCh38
rs1048570500 Chr.19: 5727641-5727641 on Build GRCh38
rs567552095 Chr.19: 5727618-5727618 on Build GRCh38
rs926934538 Chr.19: 5727600-5727600 on Build GRCh38
rs754287311 Chr.19: 5727631-5727631 on Build GRCh38
rs931751498 Chr.19: 5727667-5727667 on Build GRCh38
rs1030202628 Chr.19: 5727505-5727505 on Build GRCh38
rs776962512 Chr.19: 5727319-5727319 on Build GRCh38
rs1021400814 Chr.19: 5727758-5727758 on Build GRCh38
rs979767635 Chr.19: 5727552-5727552 on Build GRCh38
rs772014392 Chr.19: 5727708-5727708 on Build GRCh38
rs887955475 Chr.19: 5727675-5727675 on Build GRCh38
rs762403499 Chr.19: 5727321-5727321 on Build GRCh38
rs1018257855 Chr.19: 5727385-5727385 on Build GRCh38
rs901284602 Chr.19: 5727433-5727433 on Build GRCh38
rs200448036 Chr.19: 5727303-5727303 on Build GRCh38
rs763471291 Chr.19: 5727384-5727384 on Build GRCh38
rs768213227 Chr.19: 5727318-5727318 on Build GRCh38
rs756956309 Chr.19: 5727289-5727289 on Build GRCh38
rs1048683556 Chr.19: 5727276-5727276 on Build GRCh38
rs1021518263 Chr.19: 5727523-5727523 on Build GRCh38
rs537135469 Chr.19: 5727483-5727483 on Build GRCh38
rs956077499 Chr.19: 5727515-5727515 on Build GRCh38
rs765711880 Chr.19: 5727336-5727336 on Build GRCh38
rs990089437 Chr.19: 5727589-5727589 on Build GRCh38

View Full Product Details