Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5748137-5748683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPERD
Entrez Gene ID
257062
Gene Name
cation channel sperm associated auxiliary subunit delta
Gene Aliases
TMEM146
UniGene
Hs.631842
Gene Chromosome Location
Chr.19: 5720385-5778734 on Build GRCh38
Transcripts
NM_152784.3 XM_011527885.2 XM_011527886.2 XM_011527887.2 XM_011527888.2 XM_011527889.2 XM_011527890.2 XM_011527891.1 XM_017026564.1 XM_017026565.1 XM_017026566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5665051 c.845A>T p.D282V Chr.19: 5748196-5748196 on Build GRCh38
5390917 c.834G>A p.V278V Chr.19: 5748185-5748185 on Build GRCh38
3540277 c.863C>T p.S288F Chr.19: 5748214-5748214 on Build GRCh38
4417371 c.816C>G p.L272L Chr.19: 5748167-5748167 on Build GRCh38
357853 c.829C>T p.R277W Chr.19: 5748180-5748180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766108792 Chr.19: 5748259-5748259 on Build GRCh38
rs999523906 Chr.19: 5748468-5748468 on Build GRCh38
rs368431820 Chr.19: 5748245-5748245 on Build GRCh38
rs73920054 Chr.19: 5748177-5748177 on Build GRCh38
rs866701427 Chr.19: 5748220-5748220 on Build GRCh38
rs892510623 Chr.19: 5748656-5748656 on Build GRCh38
rs201831364 Chr.19: 5748197-5748197 on Build GRCh38
rs921804089 Chr.19: 5748378-5748378 on Build GRCh38
rs948166762 Chr.19: 5748452-5748452 on Build GRCh38
rs771234851 Chr.19: 5748214-5748214 on Build GRCh38
rs995216840 Chr.19: 5748355-5748355 on Build GRCh38
rs192744307 Chr.19: 5748170-5748170 on Build GRCh38
rs778386653 Chr.19: 5748272-5748272 on Build GRCh38
rs187632509 Chr.19: 5748397-5748397 on Build GRCh38
rs958115665 Chr.19: 5748509-5748509 on Build GRCh38
rs904111720 Chr.19: 5748229-5748229 on Build GRCh38
rs529155205 Chr.19: 5748181-5748181 on Build GRCh38
rs1052717258 Chr.19: 5748544-5748544 on Build GRCh38
rs372663902 Chr.19: 5748182-5748182 on Build GRCh38
rs369400230 Chr.19: 5748270-5748270 on Build GRCh38
rs1049929435 Chr.19: 5748359-5748359 on Build GRCh38
rs373844202 Chr.19: 5748179-5748179 on Build GRCh38
rs758138933 Chr.19: 5748176-5748176 on Build GRCh38
rs149492736 Chr.19: 5748265-5748265 on Build GRCh38
rs746059692 Chr.19: 5748225-5748225 on Build GRCh38
rs376369654 Chr.19: 5748298-5748298 on Build GRCh38
rs371809557 Chr.19: 5748191-5748191 on Build GRCh38
rs772382021 Chr.19: 5748230-5748230 on Build GRCh38
rs1008233687 Chr.19: 5748560-5748560 on Build GRCh38
rs990838048 Chr.19: 5748513-5748513 on Build GRCh38
rs904059414 Chr.19: 5748497-5748497 on Build GRCh38
rs899906469 Chr.19: 5748616-5748617 on Build GRCh38
rs376650447 Chr.19: 5748266-5748266 on Build GRCh38
rs936896341 Chr.19: 5748508-5748508 on Build GRCh38
rs201202287 Chr.19: 5748168-5748168 on Build GRCh38
rs1019715229 Chr.19: 5748304-5748304 on Build GRCh38
rs757835155 Chr.19: 5748299-5748299 on Build GRCh38
rs1020083552 Chr.19: 5748613-5748614 on Build GRCh38
rs917676375 Chr.19: 5748441-5748441 on Build GRCh38
rs1032877324 Chr.19: 5748504-5748504 on Build GRCh38
rs1019845884 Chr.19: 5748423-5748423 on Build GRCh38
rs77442187 Chr.19: 5748167-5748167 on Build GRCh38
rs978603822 Chr.19: 5748204-5748204 on Build GRCh38
rs1056592475 Chr.19: 5748430-5748430 on Build GRCh38
rs760798679 Chr.19: 5748239-5748239 on Build GRCh38
rs762767140 Chr.19: 5748258-5748258 on Build GRCh38
rs907519484 Chr.19: 5748428-5748428 on Build GRCh38
rs372911128 Chr.19: 5748295-5748295 on Build GRCh38
rs779121013 Chr.19: 5748303-5748303 on Build GRCh38
rs974556815 Chr.19: 5748357-5748357 on Build GRCh38
rs563097211 Chr.19: 5748342-5748342 on Build GRCh38
rs377253618 Chr.19: 5748260-5748260 on Build GRCh38
rs371642511 Chr.19: 5748243-5748243 on Build GRCh38
rs1031004254 Chr.19: 5748264-5748264 on Build GRCh38
rs889929546 Chr.19: 5748396-5748396 on Build GRCh38
rs1028619214 Chr.19: 5748159-5748159 on Build GRCh38
rs780918618 Chr.19: 5748559-5748559 on Build GRCh38
rs367967767 Chr.19: 5748180-5748180 on Build GRCh38
rs750202905 Chr.19: 5748171-5748171 on Build GRCh38
rs369486913 Chr.19: 5748474-5748474 on Build GRCh38
rs866376155 Chr.19: 5748343-5748343 on Build GRCh38
rs774586641 Chr.19: 5748222-5748222 on Build GRCh38
rs773137259 Chr.19: 5748253-5748253 on Build GRCh38
rs12982570 Chr.19: 5748374-5748374 on Build GRCh38
rs139627891 Chr.19: 5748246-5748246 on Build GRCh38
rs536534122 Chr.19: 5748612-5748612 on Build GRCh38
rs752198648 Chr.19: 5748268-5748268 on Build GRCh38
rs939690694 Chr.19: 5748277-5748277 on Build GRCh38
rs984697257 Chr.19: 5748419-5748419 on Build GRCh38
rs72983135 Chr.19: 5748429-5748429 on Build GRCh38
rs981795886 Chr.19: 5748585-5748585 on Build GRCh38
rs375060881 Chr.19: 5748244-5748244 on Build GRCh38
rs567993540 Chr.19: 5748556-5748556 on Build GRCh38
rs775539339 Chr.19: 5748234-5748234 on Build GRCh38

View Full Product Details