Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5838525-5839030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT6
Entrez Gene ID
2528
Gene Name
fucosyltransferase 6
Gene Aliases
FCT3A, FT1A, Fuc-TVI, FucT-VI
UniGene
Hs.631846
Gene Chromosome Location
Chr.19: 5830626-5839753 on Build GRCh38
Transcripts
NM_000150.2 NM_001040701.1 XM_005259526.4 XM_005259527.3 XM_011527868.2 XM_011527869.2 XM_011527870.2 XM_011527872.2 XM_011527873.2 XM_011527874.2 XM_011527875.2 XM_011527876.2 XM_011527878.2 XM_011527879.2

SNP Detail

SNP ID Chromosome Location
rs558261281 Chr.19: 5838652-5838652 on Build GRCh38
rs185856449 Chr.19: 5838607-5838607 on Build GRCh38
rs562997995 Chr.19: 5838750-5838750 on Build GRCh38
rs8101385 Chr.19: 5838584-5838584 on Build GRCh38
rs942904708 Chr.19: 5838860-5838860 on Build GRCh38
rs989139953 Chr.19: 5838836-5838836 on Build GRCh38
rs147625395 Chr.19: 5838649-5838649 on Build GRCh38
rs960642801 Chr.19: 5838768-5838768 on Build GRCh38
rs147005931 Chr.19: 5838642-5838642 on Build GRCh38
rs1002558028 Chr.19: 5838683-5838683 on Build GRCh38
rs1030986998 Chr.19: 5838801-5838801 on Build GRCh38
rs1044547744 Chr.19: 5838671-5838671 on Build GRCh38
rs965355099 Chr.19: 5838625-5838625 on Build GRCh38
rs764386049 Chr.19: 5838559-5838559 on Build GRCh38
rs531703784 Chr.19: 5838989-5838989 on Build GRCh38
rs1027595297 Chr.19: 5838595-5838595 on Build GRCh38
rs372298888 Chr.19: 5838903-5838903 on Build GRCh38
rs916210621 Chr.19: 5838791-5838791 on Build GRCh38
rs544596903 Chr.19: 5838719-5838719 on Build GRCh38
rs552575987 Chr.19: 5838911-5838911 on Build GRCh38
rs1039957654 Chr.19: 5838862-5838862 on Build GRCh38
rs931362828 Chr.19: 5838907-5838908 on Build GRCh38
rs531178578 Chr.19: 5838606-5838606 on Build GRCh38
rs562717920 Chr.19: 5838923-5838923 on Build GRCh38
rs965323868 Chr.19: 5838796-5838796 on Build GRCh38
rs752674801 Chr.19: 5838735-5838735 on Build GRCh38
rs912748397 Chr.19: 5838848-5838848 on Build GRCh38
rs758520742 Chr.19: 5838775-5838775 on Build GRCh38
rs746952727 Chr.19: 5838873-5838873 on Build GRCh38
rs923752476 Chr.19: 5838808-5838808 on Build GRCh38
rs1034788787 Chr.19: 5838988-5838988 on Build GRCh38
rs914380313 Chr.19: 5838870-5838870 on Build GRCh38
rs930203964 Chr.19: 5838946-5838946 on Build GRCh38
rs1012539302 Chr.19: 5839006-5839006 on Build GRCh38
rs888590737 Chr.19: 5838985-5838985 on Build GRCh38
rs116792659 Chr.19: 5838825-5838825 on Build GRCh38
rs896342506 Chr.19: 5838994-5838994 on Build GRCh38
rs1049129097 Chr.19: 5838959-5838959 on Build GRCh38
rs950553399 Chr.19: 5838552-5838552 on Build GRCh38
rs1027876017 Chr.19: 5838551-5838551 on Build GRCh38
rs561211197 Chr.19: 5838721-5838721 on Build GRCh38
rs975940998 Chr.19: 5838632-5838632 on Build GRCh38
rs986994719 Chr.19: 5838835-5838835 on Build GRCh38
rs927922288 Chr.19: 5838604-5838604 on Build GRCh38
rs1004661296 Chr.19: 5838914-5838914 on Build GRCh38
rs536418528 Chr.19: 5838638-5838638 on Build GRCh38
rs75998859 Chr.19: 5838643-5838643 on Build GRCh38
rs923208302 Chr.19: 5838637-5838637 on Build GRCh38
rs911181776 Chr.19: 5838613-5838613 on Build GRCh38
rs896862973 Chr.19: 5838929-5838929 on Build GRCh38
rs73920069 Chr.19: 5838718-5838718 on Build GRCh38
rs374029480 Chr.19: 5838928-5838928 on Build GRCh38
rs960361154 Chr.19: 5838605-5838605 on Build GRCh38
rs574699260 Chr.19: 5838725-5838725 on Build GRCh38
rs368365032 Chr.19: 5838891-5838891 on Build GRCh38
rs182398606 Chr.19: 5838912-5838912 on Build GRCh38
rs190182678 Chr.19: 5838769-5838769 on Build GRCh38
rs372435962 Chr.19: 5838563-5838563 on Build GRCh38
rs887218034 Chr.19: 5838913-5838913 on Build GRCh38
rs1025733540 Chr.19: 5838720-5838720 on Build GRCh38
rs994860293 Chr.19: 5838724-5838724 on Build GRCh38
rs905498430 Chr.19: 5838679-5838679 on Build GRCh38
rs1003093439 Chr.19: 5838553-5838553 on Build GRCh38
rs898454402 Chr.19: 5838892-5838892 on Build GRCh38
rs533576544 Chr.19: 5838797-5838797 on Build GRCh38
rs540380256 Chr.19: 5838748-5838748 on Build GRCh38

View Full Product Details