Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5838770-5839283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT6
Entrez Gene ID
2528
Gene Name
fucosyltransferase 6
Gene Aliases
FCT3A, FT1A, Fuc-TVI, FucT-VI
UniGene
Hs.631846
Gene Chromosome Location
Chr.19: 5830626-5839753 on Build GRCh38
Transcripts
NM_000150.2 NM_001040701.1 XM_005259526.4 XM_005259527.3 XM_011527868.2 XM_011527869.2 XM_011527870.2 XM_011527872.2 XM_011527873.2 XM_011527874.2 XM_011527875.2 XM_011527876.2 XM_011527878.2 XM_011527879.2

SNP Detail

SNP ID Chromosome Location
rs918224757 Chr.19: 5839156-5839156 on Build GRCh38
rs938408462 Chr.19: 5839116-5839116 on Build GRCh38
rs938745159 Chr.19: 5839206-5839212 on Build GRCh38
rs998266015 Chr.19: 5839045-5839045 on Build GRCh38
rs992578989 Chr.19: 5839146-5839146 on Build GRCh38
rs942904708 Chr.19: 5838860-5838860 on Build GRCh38
rs989139953 Chr.19: 5838836-5838836 on Build GRCh38
rs1030986998 Chr.19: 5838801-5838801 on Build GRCh38
rs1047088307 Chr.19: 5839157-5839157 on Build GRCh38
rs1021980327 Chr.19: 5839072-5839072 on Build GRCh38
rs531703784 Chr.19: 5838989-5838989 on Build GRCh38
rs372298888 Chr.19: 5838903-5838903 on Build GRCh38
rs1012117005 Chr.19: 5839234-5839234 on Build GRCh38
rs552575987 Chr.19: 5838911-5838911 on Build GRCh38
rs546817300 Chr.19: 5839093-5839093 on Build GRCh38
rs1039957654 Chr.19: 5838862-5838862 on Build GRCh38
rs567060960 Chr.19: 5839106-5839106 on Build GRCh38
rs965323868 Chr.19: 5838796-5838796 on Build GRCh38
rs931362828 Chr.19: 5838907-5838908 on Build GRCh38
rs562717920 Chr.19: 5838923-5838923 on Build GRCh38
rs912748397 Chr.19: 5838848-5838848 on Build GRCh38
rs746952727 Chr.19: 5838873-5838873 on Build GRCh38
rs923752476 Chr.19: 5838808-5838808 on Build GRCh38
rs914380313 Chr.19: 5838870-5838870 on Build GRCh38
rs1034788787 Chr.19: 5838988-5838988 on Build GRCh38
rs930203964 Chr.19: 5838946-5838946 on Build GRCh38
rs1012539302 Chr.19: 5839006-5839006 on Build GRCh38
rs888590737 Chr.19: 5838985-5838985 on Build GRCh38
rs896342506 Chr.19: 5838994-5838994 on Build GRCh38
rs116792659 Chr.19: 5838825-5838825 on Build GRCh38
rs912694544 Chr.19: 5839107-5839108 on Build GRCh38
rs1049129097 Chr.19: 5838959-5838959 on Build GRCh38
rs956413152 Chr.19: 5839100-5839100 on Build GRCh38
rs769711323 Chr.19: 5839215-5839215 on Build GRCh38
rs887125001 Chr.19: 5839181-5839181 on Build GRCh38
rs1030741964 Chr.19: 5839081-5839081 on Build GRCh38
rs986994719 Chr.19: 5838835-5838835 on Build GRCh38
rs527821634 Chr.19: 5839115-5839115 on Build GRCh38
rs1004661296 Chr.19: 5838914-5838914 on Build GRCh38
rs896862973 Chr.19: 5838929-5838929 on Build GRCh38
rs1030923231 Chr.19: 5839054-5839054 on Build GRCh38
rs374029480 Chr.19: 5838928-5838928 on Build GRCh38
rs149084677 Chr.19: 5839032-5839032 on Build GRCh38
rs956931989 Chr.19: 5839061-5839061 on Build GRCh38
rs558590761 Chr.19: 5839218-5839218 on Build GRCh38
rs920231014 Chr.19: 5839133-5839133 on Build GRCh38
rs368365032 Chr.19: 5838891-5838891 on Build GRCh38
rs182398606 Chr.19: 5838912-5838912 on Build GRCh38
rs887218034 Chr.19: 5838913-5838913 on Build GRCh38
rs929644322 Chr.19: 5839208-5839208 on Build GRCh38
rs116953194 Chr.19: 5839214-5839214 on Build GRCh38
rs988718651 Chr.19: 5839063-5839063 on Build GRCh38
rs567529239 Chr.19: 5839059-5839059 on Build GRCh38
rs968687949 Chr.19: 5839075-5839075 on Build GRCh38
rs1007485769 Chr.19: 5839017-5839017 on Build GRCh38
rs942888456 Chr.19: 5839111-5839111 on Build GRCh38
rs533576544 Chr.19: 5838797-5838797 on Build GRCh38
rs898454402 Chr.19: 5838892-5838892 on Build GRCh38
rs901251098 Chr.19: 5839033-5839033 on Build GRCh38
rs754897390 Chr.19: 5839010-5839010 on Build GRCh38
rs536455808 Chr.19: 5839065-5839065 on Build GRCh38
rs764054379 Chr.19: 5839143-5839146 on Build GRCh38
rs747911807 Chr.19: 5839167-5839167 on Build GRCh38

View Full Product Details