Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12878291-12878798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE2
Entrez Gene ID
1777
Gene Name
deoxyribonuclease II, lysosomal
Gene Aliases
DNASE2A, DNL, DNL2
UniGene
Hs.118243
Gene Chromosome Location
Chr.19: 12875211-12881521 on Build GRCh38
Transcripts
NM_001375.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1179780 c.435C>T p.S145S Chr.19: 12878746-12878746 on Build GRCh38
438651 c.415delG p.A139fs*36 Chr.19: 12878766-12878766 on Build GRCh38
6647463 c.476G>C p.C159S Chr.19: 12878705-12878705 on Build GRCh38
4428200 c.603C>G p.V201V Chr.19: 12878488-12878488 on Build GRCh38
4437206 c.447C>T p.S149S Chr.19: 12878734-12878734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753419724 Chr.19: 12878433-12878433 on Build GRCh38
rs764898961 Chr.19: 12878526-12878526 on Build GRCh38
rs1019467719 Chr.19: 12878512-12878512 on Build GRCh38
rs16978744 Chr.19: 12878480-12878480 on Build GRCh38
rs780051331 Chr.19: 12878644-12878644 on Build GRCh38
rs559395836 Chr.19: 12878694-12878694 on Build GRCh38
rs867875402 Chr.19: 12878766-12878766 on Build GRCh38
rs747955427 Chr.19: 12878662-12878662 on Build GRCh38
rs774019136 Chr.19: 12878733-12878733 on Build GRCh38
rs758549402 Chr.19: 12878688-12878688 on Build GRCh38
rs1046653514 Chr.19: 12878356-12878356 on Build GRCh38
rs2293682 Chr.19: 12878746-12878746 on Build GRCh38
rs763188696 Chr.19: 12878768-12878768 on Build GRCh38
rs745911181 Chr.19: 12878491-12878491 on Build GRCh38
rs1016647190 Chr.19: 12878612-12878612 on Build GRCh38
rs771117083 Chr.19: 12878486-12878486 on Build GRCh38
rs761471696 Chr.19: 12878440-12878440 on Build GRCh38
rs375732425 Chr.19: 12878421-12878421 on Build GRCh38
rs776317136 Chr.19: 12878656-12878656 on Build GRCh38
rs774427647 Chr.19: 12878392-12878392 on Build GRCh38
rs574232701 Chr.19: 12878731-12878731 on Build GRCh38
rs1056459011 Chr.19: 12878655-12878655 on Build GRCh38
rs778611100 Chr.19: 12878455-12878455 on Build GRCh38
rs367700946 Chr.19: 12878505-12878505 on Build GRCh38
rs757441500 Chr.19: 12878624-12878624 on Build GRCh38
rs368624630 Chr.19: 12878345-12878345 on Build GRCh38
rs760522934 Chr.19: 12878506-12878506 on Build GRCh38
rs558856830 Chr.19: 12878683-12878683 on Build GRCh38
rs748948347 Chr.19: 12878727-12878727 on Build GRCh38
rs147237033 Chr.19: 12878516-12878516 on Build GRCh38
rs750747420 Chr.19: 12878767-12878767 on Build GRCh38
rs763803681 Chr.19: 12878430-12878430 on Build GRCh38
rs772650354 Chr.19: 12878546-12878546 on Build GRCh38
rs373190378 Chr.19: 12878722-12878722 on Build GRCh38
rs916716760 Chr.19: 12878680-12878680 on Build GRCh38
rs940646510 Chr.19: 12878594-12878594 on Build GRCh38
rs779171808 Chr.19: 12878626-12878626 on Build GRCh38
rs747045824 Chr.19: 12878704-12878704 on Build GRCh38
rs1024208306 Chr.19: 12878461-12878461 on Build GRCh38
rs754190671 Chr.19: 12878613-12878613 on Build GRCh38
rs148522375 Chr.19: 12878428-12878428 on Build GRCh38
rs748478088 Chr.19: 12878338-12878338 on Build GRCh38
rs772044130 Chr.19: 12878499-12878499 on Build GRCh38
rs756628525 Chr.19: 12878586-12878586 on Build GRCh38
rs540721218 Chr.19: 12878689-12878689 on Build GRCh38
rs577376992 Chr.19: 12878684-12878684 on Build GRCh38
rs764367413 Chr.19: 12878591-12878591 on Build GRCh38
rs768453783 Chr.19: 12878511-12878511 on Build GRCh38
rs762523522 Chr.19: 12878550-12878550 on Build GRCh38
rs762606971 Chr.19: 12878669-12878669 on Build GRCh38
rs768202850 Chr.19: 12878551-12878551 on Build GRCh38
rs903636310 Chr.19: 12878465-12878465 on Build GRCh38
rs763046911 Chr.19: 12878695-12878695 on Build GRCh38
rs746953632 Chr.19: 12878651-12878651 on Build GRCh38
rs147892950 Chr.19: 12878734-12878734 on Build GRCh38
rs930786202 Chr.19: 12878570-12878570 on Build GRCh38
rs202170563 Chr.19: 12878475-12878475 on Build GRCh38
rs1030626745 Chr.19: 12878476-12878476 on Build GRCh38
rs747962653 Chr.19: 12878721-12878721 on Build GRCh38
rs746015779 Chr.19: 12878639-12878639 on Build GRCh38
rs544157644 Chr.19: 12878643-12878643 on Build GRCh38
rs1056699939 Chr.19: 12878351-12878351 on Build GRCh38
rs754923904 Chr.19: 12878709-12878709 on Build GRCh38
rs376167745 Chr.19: 12878665-12878665 on Build GRCh38
rs935346827 Chr.19: 12878438-12878438 on Build GRCh38
rs781716298 Chr.19: 12878332-12878332 on Build GRCh38
rs749911697 Chr.19: 12878444-12878444 on Build GRCh38
rs765996726 Chr.19: 12878671-12878671 on Build GRCh38
rs140738663 Chr.19: 12878677-12878677 on Build GRCh38
rs757609925 Chr.19: 12878681-12878681 on Build GRCh38
rs771955952 Chr.19: 12878419-12878419 on Build GRCh38
rs919748766 Chr.19: 12878561-12878561 on Build GRCh38
rs547496555 Chr.19: 12878707-12878707 on Build GRCh38
rs768544258 Chr.19: 12878652-12878652 on Build GRCh38
rs781155237 Chr.19: 12878710-12878710 on Build GRCh38
rs775803168 Chr.19: 12878585-12878585 on Build GRCh38
rs374218185 Chr.19: 12878672-12878672 on Build GRCh38
rs998494644 Chr.19: 12878493-12878493 on Build GRCh38
rs770193338 Chr.19: 12878363-12878363 on Build GRCh38
rs765639857 Chr.19: 12878762-12878762 on Build GRCh38
rs753149246 Chr.19: 12878464-12878464 on Build GRCh38
rs369135205 Chr.19: 12878585-12878585 on Build GRCh38
rs565891293 Chr.19: 12878427-12878427 on Build GRCh38
rs778890192 Chr.19: 12878490-12878490 on Build GRCh38
rs761407102 Chr.19: 12878524-12878524 on Build GRCh38
rs975301847 Chr.19: 12878554-12878554 on Build GRCh38
rs530686133 Chr.19: 12878747-12878747 on Build GRCh38
rs554251996 Chr.19: 12878390-12878390 on Build GRCh38
rs371265511 Chr.19: 12878663-12878663 on Build GRCh38
rs764662113 Chr.19: 12878443-12878443 on Build GRCh38
rs772156292 Chr.19: 12878483-12878483 on Build GRCh38
rs754201510 Chr.19: 12878678-12878678 on Build GRCh38
rs1018517407 Chr.19: 12878482-12878482 on Build GRCh38
rs759396010 Chr.19: 12878406-12878406 on Build GRCh38
rs749448953 Chr.19: 12878373-12878373 on Build GRCh38
rs781759575 Chr.19: 12878463-12878463 on Build GRCh38
rs778108310 Chr.19: 12878364-12878364 on Build GRCh38

View Full Product Details