Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43976507-43977004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0B
Entrez Gene ID
533
Gene Name
ATPase H+ transporting V0 subunit b
Gene Aliases
ATP6F, HATPL, VMA16
UniGene
Hs.596514
Gene Chromosome Location
Chr.1: 43974648-43978300 on Build GRCh38
Transcripts
NM_001039457.2 NM_001294333.1 NM_004047.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5381066 c.301G>A p.A101T Chr.1: 43976612-43976612 on Build GRCh38
5025264 c.286T>A p.F96I Chr.1: 43976597-43976597 on Build GRCh38
7296236 c.362C>T p.T121I Chr.1: 43976786-43976786 on Build GRCh38
2171351 c.378C>T p.I126I Chr.1: 43976802-43976802 on Build GRCh38
2171350 c.318C>T p.I106I Chr.1: 43976629-43976629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004517634 Chr.1: 43976672-43976672 on Build GRCh38
rs570568258 Chr.1: 43976867-43976867 on Build GRCh38
rs555279277 Chr.1: 43976951-43976951 on Build GRCh38
rs767136073 Chr.1: 43976730-43976730 on Build GRCh38
rs746213678 Chr.1: 43976647-43976647 on Build GRCh38
rs753401849 Chr.1: 43976851-43976851 on Build GRCh38
rs777944365 Chr.1: 43976809-43976809 on Build GRCh38
rs781639921 Chr.1: 43976980-43976980 on Build GRCh38
rs1041216516 Chr.1: 43976914-43976914 on Build GRCh38
rs115663196 Chr.1: 43976960-43976960 on Build GRCh38
rs774120589 Chr.1: 43976829-43976829 on Build GRCh38
rs774140781 Chr.1: 43976725-43976725 on Build GRCh38
rs766950507 Chr.1: 43976596-43976596 on Build GRCh38
rs901325443 Chr.1: 43976927-43976927 on Build GRCh38
rs191439288 Chr.1: 43976885-43976885 on Build GRCh38
rs41270393 Chr.1: 43976695-43976695 on Build GRCh38
rs949017181 Chr.1: 43976926-43976926 on Build GRCh38
rs759394656 Chr.1: 43976832-43976832 on Build GRCh38
rs755962790 Chr.1: 43976858-43976858 on Build GRCh38
rs773042993 Chr.1: 43976544-43976544 on Build GRCh38
rs759003608 Chr.1: 43976753-43976753 on Build GRCh38
rs760396755 Chr.1: 43976838-43976838 on Build GRCh38
rs765752256 Chr.1: 43976749-43976749 on Build GRCh38
rs1046140142 Chr.1: 43976944-43976944 on Build GRCh38
rs199866002 Chr.1: 43976629-43976629 on Build GRCh38
rs369492376 Chr.1: 43976682-43976682 on Build GRCh38
rs201804028 Chr.1: 43976671-43976671 on Build GRCh38
rs760144170 Chr.1: 43976741-43976741 on Build GRCh38
rs372745081 Chr.1: 43976683-43976683 on Build GRCh38
rs200988092 Chr.1: 43976839-43976839 on Build GRCh38
rs1028370427 Chr.1: 43976797-43976797 on Build GRCh38
rs887316878 Chr.1: 43976546-43976546 on Build GRCh38
rs1026683156 Chr.1: 43976531-43976531 on Build GRCh38
rs937865463 Chr.1: 43976970-43976970 on Build GRCh38
rs767110678 Chr.1: 43976800-43976800 on Build GRCh38
rs765097306 Chr.1: 43976539-43976539 on Build GRCh38
rs763710464 Chr.1: 43976537-43976537 on Build GRCh38
rs374119264 Chr.1: 43976804-43976804 on Build GRCh38
rs924690911 Chr.1: 43976722-43976722 on Build GRCh38
rs755235142 Chr.1: 43976767-43976767 on Build GRCh38
rs780497975 Chr.1: 43976755-43976755 on Build GRCh38
rs755334693 Chr.1: 43976873-43976873 on Build GRCh38
rs769717078 Chr.1: 43976806-43976806 on Build GRCh38
rs761448712 Chr.1: 43976570-43976570 on Build GRCh38
rs749662178 Chr.1: 43976850-43976854 on Build GRCh38
rs762904303 Chr.1: 43976707-43976707 on Build GRCh38
rs888345613 Chr.1: 43976648-43976648 on Build GRCh38
rs770870823 Chr.1: 43976816-43976816 on Build GRCh38
rs548133772 Chr.1: 43976763-43976763 on Build GRCh38
rs1004341714 Chr.1: 43976560-43976560 on Build GRCh38
rs759132891 Chr.1: 43976728-43976728 on Build GRCh38
rs779356770 Chr.1: 43976642-43976642 on Build GRCh38
rs750846538 Chr.1: 43976752-43976752 on Build GRCh38
rs377647424 Chr.1: 43976704-43976704 on Build GRCh38
rs374872009 Chr.1: 43976592-43976592 on Build GRCh38
rs751252390 Chr.1: 43976569-43976569 on Build GRCh38
rs748224999 Chr.1: 43976802-43976802 on Build GRCh38
rs377754670 Chr.1: 43976810-43976810 on Build GRCh38
rs778167105 Chr.1: 43976830-43976831 on Build GRCh38
rs763494081 Chr.1: 43976855-43976855 on Build GRCh38
rs371099886 Chr.1: 43976735-43976735 on Build GRCh38
rs1001960030 Chr.1: 43976641-43976641 on Build GRCh38
rs773076922 Chr.1: 43976706-43976706 on Build GRCh38
rs905037694 Chr.1: 43976601-43976601 on Build GRCh38
rs752826674 Chr.1: 43976982-43976982 on Build GRCh38
rs770676237 Chr.1: 43976710-43976710 on Build GRCh38
rs12410334 Chr.1: 43976849-43976849 on Build GRCh38
rs539194978 Chr.1: 43976869-43976869 on Build GRCh38
rs771724220 Chr.1: 43976833-43976833 on Build GRCh38
rs892114317 Chr.1: 43976552-43976552 on Build GRCh38
rs1046128856 Chr.1: 43976571-43976571 on Build GRCh38
rs762679268 Chr.1: 43976530-43976530 on Build GRCh38
rs781331446 Chr.1: 43976778-43976778 on Build GRCh38
rs752164497 Chr.1: 43976620-43976620 on Build GRCh38
rs978877537 Chr.1: 43976717-43976717 on Build GRCh38
rs917733288 Chr.1: 43976793-43976793 on Build GRCh38
rs751167166 Chr.1: 43976584-43976584 on Build GRCh38
rs769003302 Chr.1: 43976689-43976689 on Build GRCh38
rs142793826 Chr.1: 43976910-43976910 on Build GRCh38
rs535357237 Chr.1: 43976949-43976949 on Build GRCh38
rs775055228 Chr.1: 43976835-43976835 on Build GRCh38
rs748230317 Chr.1: 43976703-43976703 on Build GRCh38

View Full Product Details