Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44784463-44784983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEST4
Entrez Gene ID
266675
Gene Name
bestrophin 4
Gene Aliases
VMD2L2
UniGene
Hs.302513
Gene Chromosome Location
Chr.1: 44781836-44792828 on Build GRCh38
Transcripts
NM_153274.2 XM_017001020.1 XM_017001021.1 XM_017001022.1 XM_017001023.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4459278 c.1119C>T p.P373P Chr.1: 44784658-44784658 on Build GRCh38
7425897 c.1104C>A p.A368A Chr.1: 44784673-44784673 on Build GRCh38
4439776 c.1032T>C p.L344L Chr.1: 44784745-44784745 on Build GRCh38
4008436 c.1037C>T p.P346L Chr.1: 44784740-44784740 on Build GRCh38
6611369 c.1009G>A p.V337M Chr.1: 44784768-44784768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966114563 Chr.1: 44784647-44784647 on Build GRCh38
rs762198297 Chr.1: 44784883-44784883 on Build GRCh38
rs778549996 Chr.1: 44784916-44784916 on Build GRCh38
rs372498770 Chr.1: 44784912-44784912 on Build GRCh38
rs202109934 Chr.1: 44784604-44784604 on Build GRCh38
rs781569913 Chr.1: 44784586-44784586 on Build GRCh38
rs773705288 Chr.1: 44784858-44784858 on Build GRCh38
rs776266325 Chr.1: 44784702-44784702 on Build GRCh38
rs753116814 Chr.1: 44784710-44784715 on Build GRCh38
rs913241832 Chr.1: 44784501-44784501 on Build GRCh38
rs779163006 Chr.1: 44784682-44784682 on Build GRCh38
rs542213146 Chr.1: 44784595-44784595 on Build GRCh38
rs762584742 Chr.1: 44784832-44784832 on Build GRCh38
rs767954927 Chr.1: 44784669-44784669 on Build GRCh38
rs779216302 Chr.1: 44784772-44784772 on Build GRCh38
rs770637220 Chr.1: 44784816-44784816 on Build GRCh38
rs753592848 Chr.1: 44784621-44784621 on Build GRCh38
rs1012656124 Chr.1: 44784877-44784877 on Build GRCh38
rs377207881 Chr.1: 44784921-44784921 on Build GRCh38
rs372933274 Chr.1: 44784823-44784823 on Build GRCh38
rs761357873 Chr.1: 44784634-44784634 on Build GRCh38
rs774553146 Chr.1: 44784601-44784601 on Build GRCh38
rs758614240 Chr.1: 44784781-44784781 on Build GRCh38
rs772723521 Chr.1: 44784710-44784710 on Build GRCh38
rs749575880 Chr.1: 44784679-44784679 on Build GRCh38
rs559437180 Chr.1: 44784560-44784560 on Build GRCh38
rs749471926 Chr.1: 44784597-44784597 on Build GRCh38
rs772800009 Chr.1: 44784522-44784522 on Build GRCh38
rs746548544 Chr.1: 44784568-44784568 on Build GRCh38
rs959136954 Chr.1: 44784518-44784518 on Build GRCh38
rs772145581 Chr.1: 44784695-44784695 on Build GRCh38
rs572875585 Chr.1: 44784575-44784575 on Build GRCh38
rs997168788 Chr.1: 44784671-44784671 on Build GRCh38
rs752116565 Chr.1: 44784497-44784497 on Build GRCh38
rs765870318 Chr.1: 44784720-44784720 on Build GRCh38
rs781647396 Chr.1: 44784518-44784518 on Build GRCh38
rs748074273 Chr.1: 44784931-44784931 on Build GRCh38
rs779983381 Chr.1: 44784799-44784799 on Build GRCh38
rs780927449 Chr.1: 44784916-44784916 on Build GRCh38
rs764563258 Chr.1: 44784742-44784742 on Build GRCh38
rs917816812 Chr.1: 44784786-44784786 on Build GRCh38
rs750730789 Chr.1: 44784887-44784887 on Build GRCh38
rs1041631425 Chr.1: 44784783-44784783 on Build GRCh38
rs1002135708 Chr.1: 44784527-44784527 on Build GRCh38
rs749901864 Chr.1: 44784656-44784656 on Build GRCh38
rs201461479 Chr.1: 44784862-44784862 on Build GRCh38
rs16832241 Chr.1: 44784907-44784907 on Build GRCh38
rs564255785 Chr.1: 44784680-44784680 on Build GRCh38
rs768552808 Chr.1: 44784698-44784698 on Build GRCh38
rs201994250 Chr.1: 44784906-44784906 on Build GRCh38
rs950049962 Chr.1: 44784505-44784505 on Build GRCh38
rs1002510939 Chr.1: 44784812-44784812 on Build GRCh38
rs925120042 Chr.1: 44784706-44784706 on Build GRCh38
rs765967549 Chr.1: 44784835-44784835 on Build GRCh38
rs769443238 Chr.1: 44784826-44784826 on Build GRCh38
rs753008617 Chr.1: 44784672-44784672 on Build GRCh38
rs376989433 Chr.1: 44784652-44784652 on Build GRCh38
rs1003074418 Chr.1: 44784498-44784498 on Build GRCh38
rs754241463 Chr.1: 44784881-44784881 on Build GRCh38
rs748497240 Chr.1: 44784587-44784587 on Build GRCh38
rs1046249159 Chr.1: 44784510-44784510 on Build GRCh38
rs777990349 Chr.1: 44784674-44784674 on Build GRCh38
rs768444892 Chr.1: 44784819-44784819 on Build GRCh38
rs756572035 Chr.1: 44784673-44784673 on Build GRCh38
rs750247002 Chr.1: 44784787-44784787 on Build GRCh38
rs755503437 Chr.1: 44784664-44784664 on Build GRCh38
rs760457800 Chr.1: 44784617-44784617 on Build GRCh38
rs972297246 Chr.1: 44784548-44784548 on Build GRCh38
rs749013433 Chr.1: 44784555-44784555 on Build GRCh38
rs954732268 Chr.1: 44784605-44784605 on Build GRCh38
rs546689077 Chr.1: 44784724-44784724 on Build GRCh38
rs112195869 Chr.1: 44784519-44784519 on Build GRCh38
rs772092532 Chr.1: 44784609-44784609 on Build GRCh38
rs780220883 Chr.1: 44784896-44784896 on Build GRCh38
rs994881861 Chr.1: 44784591-44784591 on Build GRCh38
rs369252209 Chr.1: 44784886-44784886 on Build GRCh38
rs937440816 Chr.1: 44784904-44784904 on Build GRCh38
rs761565620 Chr.1: 44784703-44784703 on Build GRCh38
rs745942679 Chr.1: 44784780-44784780 on Build GRCh38
rs771197040 Chr.1: 44784598-44784598 on Build GRCh38
rs745891320 Chr.1: 44784685-44784685 on Build GRCh38
rs1049253156 Chr.1: 44784567-44784567 on Build GRCh38
rs375787125 Chr.1: 44784865-44784865 on Build GRCh38
rs1039535996 Chr.1: 44784721-44784721 on Build GRCh38
rs988031054 Chr.1: 44784743-44784743 on Build GRCh38
rs776518836 Chr.1: 44784821-44784821 on Build GRCh38
rs942555009 Chr.1: 44784716-44784716 on Build GRCh38
rs138819993 Chr.1: 44784739-44784739 on Build GRCh38
rs533175337 Chr.1: 44784696-44784696 on Build GRCh38
rs754115252 Chr.1: 44784746-44784746 on Build GRCh38
rs150797829 Chr.1: 44784923-44784923 on Build GRCh38
rs772916692 Chr.1: 44784831-44784831 on Build GRCh38
rs763653505 Chr.1: 44784620-44784620 on Build GRCh38
rs566574541 Chr.1: 44784807-44784807 on Build GRCh38
rs1029183229 Chr.1: 44784678-44784678 on Build GRCh38
rs894204398 Chr.1: 44784846-44784846 on Build GRCh38
rs756736992 Chr.1: 44784810-44784811 on Build GRCh38
rs758671494 Chr.1: 44784889-44784889 on Build GRCh38
rs762383507 Chr.1: 44784715-44784715 on Build GRCh38
rs755452954 Chr.1: 44784583-44784583 on Build GRCh38
rs769636382 Chr.1: 44784948-44784948 on Build GRCh38
rs141308464 Chr.1: 44784704-44784704 on Build GRCh38
rs540228706 Chr.1: 44784549-44784549 on Build GRCh38
rs769923561 Chr.1: 44784594-44784594 on Build GRCh38
rs757574175 Chr.1: 44784747-44784747 on Build GRCh38
rs530826885 Chr.1: 44784611-44784611 on Build GRCh38
rs369088764 Chr.1: 44784944-44784944 on Build GRCh38

View Full Product Details