Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13890440-13890959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf57
Entrez Gene ID
79173
Gene Name
chromosome 19 open reading frame 57
Gene Aliases
-
UniGene
Hs.143288
Gene Chromosome Location
Chr.19: 13882348-13906125 on Build GRCh38
Transcripts
NM_024323.3 XM_006722885.3 XM_011528286.2 XM_011528287.2 XM_017027300.1 XM_017027301.1

SNP Detail

SNP ID Chromosome Location
rs988245522 Chr.19: 13890636-13890636 on Build GRCh38
rs776590882 Chr.19: 13890656-13890656 on Build GRCh38
rs907612843 Chr.19: 13890613-13890613 on Build GRCh38
rs999475971 Chr.19: 13890665-13890665 on Build GRCh38
rs750394425 Chr.19: 13890491-13890491 on Build GRCh38
rs905119001 Chr.19: 13890908-13890908 on Build GRCh38
rs527677218 Chr.19: 13890502-13890502 on Build GRCh38
rs539137085 Chr.19: 13890562-13890565 on Build GRCh38
rs534176910 Chr.19: 13890526-13890526 on Build GRCh38
rs911619624 Chr.19: 13890720-13890720 on Build GRCh38
rs940433600 Chr.19: 13890759-13890759 on Build GRCh38
rs147832236 Chr.19: 13890626-13890626 on Build GRCh38
rs975835934 Chr.19: 13890867-13890867 on Build GRCh38
rs866667281 Chr.19: 13890785-13890785 on Build GRCh38
rs921622831 Chr.19: 13890465-13890465 on Build GRCh38
rs1027232052 Chr.19: 13890789-13890789 on Build GRCh38
rs564975656 Chr.19: 13890802-13890802 on Build GRCh38
rs567634280 Chr.19: 13890514-13890514 on Build GRCh38
rs571070283 Chr.19: 13890551-13890551 on Build GRCh38
rs779118658 Chr.19: 13890511-13890511 on Build GRCh38
rs192371042 Chr.19: 13890634-13890634 on Build GRCh38
rs189227404 Chr.19: 13890554-13890554 on Build GRCh38
rs1023615322 Chr.19: 13890589-13890589 on Build GRCh38
rs960939871 Chr.19: 13890509-13890509 on Build GRCh38
rs1003577277 Chr.19: 13890600-13890600 on Build GRCh38
rs199703310 Chr.19: 13890492-13890492 on Build GRCh38
rs987136188 Chr.19: 13890719-13890719 on Build GRCh38
rs374647698 Chr.19: 13890816-13890816 on Build GRCh38
rs944578141 Chr.19: 13890622-13890622 on Build GRCh38
rs995188828 Chr.19: 13890588-13890588 on Build GRCh38
rs1024236435 Chr.19: 13890738-13890738 on Build GRCh38
rs956791458 Chr.19: 13890631-13890631 on Build GRCh38
rs755156121 Chr.19: 13890510-13890510 on Build GRCh38
rs920444098 Chr.19: 13890799-13890799 on Build GRCh38
rs111233692 Chr.19: 13890639-13890639 on Build GRCh38
rs572336664 Chr.19: 13890630-13890630 on Build GRCh38
rs921767280 Chr.19: 13890925-13890925 on Build GRCh38
rs905234227 Chr.19: 13890591-13890591 on Build GRCh38
rs549880309 Chr.19: 13890531-13890531 on Build GRCh38
rs1043586225 Chr.19: 13890653-13890653 on Build GRCh38
rs578216134 Chr.19: 13890815-13890815 on Build GRCh38
rs970031865 Chr.19: 13890521-13890521 on Build GRCh38
rs567545985 Chr.19: 13890892-13890892 on Build GRCh38
rs184917010 Chr.19: 13890933-13890933 on Build GRCh38
rs921605876 Chr.19: 13890686-13890686 on Build GRCh38
rs1052509995 Chr.19: 13890668-13890668 on Build GRCh38
rs780050658 Chr.19: 13890500-13890500 on Build GRCh38
rs931784133 Chr.19: 13890931-13890931 on Build GRCh38
rs892523081 Chr.19: 13890926-13890926 on Build GRCh38
rs576122016 Chr.19: 13890652-13890652 on Build GRCh38
rs767496887 Chr.19: 13890489-13890489 on Build GRCh38
rs549392956 Chr.19: 13890513-13890513 on Build GRCh38
rs1013783088 Chr.19: 13890708-13890708 on Build GRCh38
rs1056410259 Chr.19: 13890918-13890918 on Build GRCh38
rs34679323 Chr.19: 13890703-13890703 on Build GRCh38
rs1045423325 Chr.19: 13890856-13890856 on Build GRCh38
rs976108981 Chr.19: 13890657-13890657 on Build GRCh38
rs761715891 Chr.19: 13890488-13890488 on Build GRCh38
rs528016862 Chr.19: 13890912-13890912 on Build GRCh38
rs200387098 Chr.19: 13890468-13890468 on Build GRCh38
rs560607474 Chr.19: 13890819-13890819 on Build GRCh38
rs969545104 Chr.19: 13890775-13890775 on Build GRCh38

View Full Product Details