Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14381409-14381905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE5
Entrez Gene ID
976
Gene Name
adhesion G protein-coupled receptor E5
Gene Aliases
CD97, TM7LN1
UniGene
Hs.466039
Gene Chromosome Location
Chr.19: 14381144-14408725 on Build GRCh38
Transcripts
NM_001025160.2 NM_001784.4 NM_078481.3 XM_011528451.2 XM_017027547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623967 c.5G>A p.G2E Chr.19: 14381528-14381528 on Build GRCh38
6623975 c.7G>T p.G3C Chr.19: 14381530-14381530 on Build GRCh38
1611670 c.4G>C p.G2R Chr.19: 14381527-14381527 on Build GRCh38
50463 c.22G>C p.A8P Chr.19: 14381545-14381545 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756503408 Chr.19: 14381516-14381516 on Build GRCh38
rs574118088 Chr.19: 14381603-14381603 on Build GRCh38
rs578000310 Chr.19: 14381683-14381683 on Build GRCh38
rs561034171 Chr.19: 14381451-14381451 on Build GRCh38
rs901414645 Chr.19: 14381652-14381652 on Build GRCh38
rs531385483 Chr.19: 14381807-14381807 on Build GRCh38
rs904560060 Chr.19: 14381712-14381712 on Build GRCh38
rs2302997 Chr.19: 14381453-14381453 on Build GRCh38
rs538564533 Chr.19: 14381617-14381617 on Build GRCh38
rs755499613 Chr.19: 14381531-14381531 on Build GRCh38
rs777093304 Chr.19: 14381481-14381481 on Build GRCh38
rs745703535 Chr.19: 14381491-14381491 on Build GRCh38
rs201356058 Chr.19: 14381446-14381446 on Build GRCh38
rs73515682 Chr.19: 14381768-14381768 on Build GRCh38
rs878914780 Chr.19: 14381695-14381695 on Build GRCh38
rs535897631 Chr.19: 14381496-14381496 on Build GRCh38
rs1028305347 Chr.19: 14381476-14381476 on Build GRCh38
rs550853961 Chr.19: 14381880-14381880 on Build GRCh38
rs746554560 Chr.19: 14381533-14381533 on Build GRCh38
rs879535384 Chr.19: 14381646-14381646 on Build GRCh38
rs1049707922 Chr.19: 14381711-14381711 on Build GRCh38
rs371030779 Chr.19: 14381467-14381467 on Build GRCh38
rs980416490 Chr.19: 14381513-14381513 on Build GRCh38
rs369055764 Chr.19: 14381480-14381480 on Build GRCh38
rs750596785 Chr.19: 14381512-14381512 on Build GRCh38
rs2302996 Chr.19: 14381460-14381460 on Build GRCh38
rs550098988 Chr.19: 14381873-14381873 on Build GRCh38
rs549788944 Chr.19: 14381454-14381454 on Build GRCh38
rs367758297 Chr.19: 14381545-14381545 on Build GRCh38
rs908304737 Chr.19: 14381879-14381879 on Build GRCh38
rs767091071 Chr.19: 14381796-14381796 on Build GRCh38
rs767776449 Chr.19: 14381511-14381511 on Build GRCh38
rs374718424 Chr.19: 14381474-14381474 on Build GRCh38
rs892267868 Chr.19: 14381770-14381770 on Build GRCh38
rs766589387 Chr.19: 14381523-14381523 on Build GRCh38
rs769634013 Chr.19: 14381569-14381569 on Build GRCh38
rs779473458 Chr.19: 14381532-14381532 on Build GRCh38
rs561168456 Chr.19: 14381792-14381792 on Build GRCh38
rs376149429 Chr.19: 14381825-14381825 on Build GRCh38
rs752458327 Chr.19: 14381534-14381535 on Build GRCh38
rs755930239 Chr.19: 14381843-14381843 on Build GRCh38
rs988431351 Chr.19: 14381498-14381498 on Build GRCh38
rs775089878 Chr.19: 14381573-14381573 on Build GRCh38
rs530817007 Chr.19: 14381505-14381505 on Build GRCh38
rs141605989 Chr.19: 14381544-14381544 on Build GRCh38
rs371403378 Chr.19: 14381501-14381501 on Build GRCh38
rs751091427 Chr.19: 14381767-14381767 on Build GRCh38
rs1041289860 Chr.19: 14381466-14381466 on Build GRCh38
rs549199439 Chr.19: 14381441-14381441 on Build GRCh38
rs932932079 Chr.19: 14381656-14381656 on Build GRCh38
rs931048600 Chr.19: 14381564-14381564 on Build GRCh38
rs34025302 Chr.19: 14381754-14381754 on Build GRCh38
rs761851309 Chr.19: 14381510-14381510 on Build GRCh38
rs185833329 Chr.19: 14381750-14381750 on Build GRCh38
rs565025561 Chr.19: 14381874-14381874 on Build GRCh38
rs976747573 Chr.19: 14381493-14381493 on Build GRCh38
rs140117277 Chr.19: 14381651-14381651 on Build GRCh38
rs115061086 Chr.19: 14381485-14381485 on Build GRCh38
rs778564424 Chr.19: 14381494-14381494 on Build GRCh38
rs149873493 Chr.19: 14381773-14381773 on Build GRCh38
rs754384842 Chr.19: 14381525-14381525 on Build GRCh38
rs940216170 Chr.19: 14381649-14381649 on Build GRCh38
rs967108634 Chr.19: 14381487-14381487 on Build GRCh38
rs376980277 Chr.19: 14381495-14381495 on Build GRCh38
rs758142503 Chr.19: 14381560-14381561 on Build GRCh38
rs749094992 Chr.19: 14381590-14381590 on Build GRCh38
rs781010964 Chr.19: 14381542-14381542 on Build GRCh38
rs375635407 Chr.19: 14381534-14381534 on Build GRCh38
rs952415125 Chr.19: 14381846-14381846 on Build GRCh38
rs1002157122 Chr.19: 14381842-14381842 on Build GRCh38

View Full Product Details