Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14401412-14401847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE5
Entrez Gene ID
976
Gene Name
adhesion G protein-coupled receptor E5
Gene Aliases
CD97, TM7LN1
UniGene
Hs.466039
Gene Chromosome Location
Chr.19: 14381144-14408725 on Build GRCh38
Transcripts
NM_001025160.2 NM_001784.4 NM_078481.3 XM_011528451.2 XM_017027547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623969 c.1052C>T p.T351I Chr.19: 14401540-14401540 on Build GRCh38
3362696 c.1141A>G p.M381V Chr.19: 14401718-14401718 on Build GRCh38
3692249 c.1021C>A p.L341M Chr.19: 14401509-14401509 on Build GRCh38
1390936 c.1173C>T p.A391A Chr.19: 14401750-14401750 on Build GRCh38
6242098 c.1183+1G>T p.? Chr.19: 14401761-14401761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568316578 Chr.19: 14401790-14401790 on Build GRCh38
rs746207246 Chr.19: 14401479-14401479 on Build GRCh38
rs569128990 Chr.19: 14401686-14401686 on Build GRCh38
rs765247258 Chr.19: 14401535-14401535 on Build GRCh38
rs1003158202 Chr.19: 14401489-14401489 on Build GRCh38
rs770014400 Chr.19: 14401598-14401598 on Build GRCh38
rs770383003 Chr.19: 14401490-14401490 on Build GRCh38
rs767028684 Chr.19: 14401585-14401585 on Build GRCh38
rs1026900432 Chr.19: 14401611-14401611 on Build GRCh38
rs145167749 Chr.19: 14401718-14401718 on Build GRCh38
rs956075882 Chr.19: 14401517-14401517 on Build GRCh38
rs767883884 Chr.19: 14401698-14401698 on Build GRCh38
rs770357759 Chr.19: 14401733-14401733 on Build GRCh38
rs1012979522 Chr.19: 14401569-14401569 on Build GRCh38
rs372215749 Chr.19: 14401734-14401734 on Build GRCh38
rs747173884 Chr.19: 14401464-14401464 on Build GRCh38
rs145137370 Chr.19: 14401751-14401751 on Build GRCh38
rs1031506181 Chr.19: 14401499-14401499 on Build GRCh38
rs139618356 Chr.19: 14401539-14401539 on Build GRCh38
rs746258131 Chr.19: 14401596-14401596 on Build GRCh38
rs951409320 Chr.19: 14401616-14401616 on Build GRCh38
rs28548336 Chr.19: 14401636-14401636 on Build GRCh38
rs201125993 Chr.19: 14401582-14401582 on Build GRCh38
rs1043275920 Chr.19: 14401510-14401510 on Build GRCh38
rs758516153 Chr.19: 14401541-14401541 on Build GRCh38
rs772694731 Chr.19: 14401511-14401511 on Build GRCh38
rs61733714 Chr.19: 14401444-14401444 on Build GRCh38
rs760469641 Chr.19: 14401435-14401435 on Build GRCh38
rs751688182 Chr.19: 14401556-14401556 on Build GRCh38
rs750607968 Chr.19: 14401705-14401705 on Build GRCh38
rs775345308 Chr.19: 14401682-14401682 on Build GRCh38
rs370979328 Chr.19: 14401555-14401555 on Build GRCh38
rs537262542 Chr.19: 14401498-14401498 on Build GRCh38
rs781734469 Chr.19: 14401478-14401478 on Build GRCh38
rs16979636 Chr.19: 14401448-14401448 on Build GRCh38
rs61733711 Chr.19: 14401690-14401690 on Build GRCh38
rs374807034 Chr.19: 14401712-14401712 on Build GRCh38
rs2230748 Chr.19: 14401677-14401677 on Build GRCh38
rs796816912 Chr.19: 14401771-14401771 on Build GRCh38
rs775192070 Chr.19: 14401750-14401750 on Build GRCh38
rs149991001 Chr.19: 14401716-14401716 on Build GRCh38
rs775811992 Chr.19: 14401494-14401494 on Build GRCh38
rs749702711 Chr.19: 14401627-14401627 on Build GRCh38
rs200898011 Chr.19: 14401780-14401780 on Build GRCh38
rs376677729 Chr.19: 14401531-14401531 on Build GRCh38
rs141443086 Chr.19: 14401722-14401722 on Build GRCh38
rs545710498 Chr.19: 14401691-14401691 on Build GRCh38
rs78841685 Chr.19: 14401676-14401676 on Build GRCh38
rs369475079 Chr.19: 14401768-14401768 on Build GRCh38
rs367986615 Chr.19: 14401603-14401603 on Build GRCh38
rs912190793 Chr.19: 14401659-14401659 on Build GRCh38
rs754176571 Chr.19: 14401788-14401788 on Build GRCh38
rs73924180 Chr.19: 14401770-14401770 on Build GRCh38
rs1023067985 Chr.19: 14401573-14401573 on Build GRCh38
rs759273933 Chr.19: 14401534-14401534 on Build GRCh38
rs770592573 Chr.19: 14401647-14401647 on Build GRCh38
rs766600227 Chr.19: 14401786-14401786 on Build GRCh38
rs756516696 Chr.19: 14401711-14401711 on Build GRCh38
rs771266840 Chr.19: 14401467-14401467 on Build GRCh38
rs758365972 Chr.19: 14401455-14401455 on Build GRCh38
rs761418785 Chr.19: 14401496-14401496 on Build GRCh38
rs762418893 Chr.19: 14401699-14401699 on Build GRCh38
rs777230363 Chr.19: 14401588-14401589 on Build GRCh38
rs761989966 Chr.19: 14401769-14401769 on Build GRCh38
rs199620684 Chr.19: 14401470-14401470 on Build GRCh38
rs778693810 Chr.19: 14401452-14401452 on Build GRCh38
rs778057042 Chr.19: 14401456-14401456 on Build GRCh38
rs769484947 Chr.19: 14401749-14401749 on Build GRCh38
rs559079747 Chr.19: 14401519-14401519 on Build GRCh38
rs746549606 Chr.19: 14401730-14401730 on Build GRCh38
rs757596890 Chr.19: 14401577-14401577 on Build GRCh38
rs753623982 Chr.19: 14401445-14401445 on Build GRCh38
rs367751456 Chr.19: 14401715-14401715 on Build GRCh38
rs923554059 Chr.19: 14401580-14401580 on Build GRCh38
rs376738555 Chr.19: 14401739-14401739 on Build GRCh38
rs375567054 Chr.19: 14401520-14401520 on Build GRCh38
rs891652853 Chr.19: 14401757-14401757 on Build GRCh38
rs547426676 Chr.19: 14401756-14401756 on Build GRCh38
rs370104930 Chr.19: 14401683-14401683 on Build GRCh38
rs528906484 Chr.19: 14401821-14401821 on Build GRCh38
rs760154980 Chr.19: 14401637-14401637 on Build GRCh38
rs372231585 Chr.19: 14401789-14401789 on Build GRCh38
rs752841394 Chr.19: 14401454-14401454 on Build GRCh38

View Full Product Details