Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15237355-15237864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD4
Entrez Gene ID
23476
Gene Name
bromodomain containing 4
Gene Aliases
CAP, HUNK1, HUNKI, MCAP
UniGene
Hs.187763
Gene Chromosome Location
Chr.19: 15236836-15332543 on Build GRCh38
Transcripts
NM_058243.2 XM_011527854.1

SNP Detail

SNP ID Chromosome Location
rs868413450 Chr.19: 15237838-15237838 on Build GRCh38
rs201490111 Chr.19: 15237485-15237485 on Build GRCh38
rs200374596 Chr.19: 15237681-15237681 on Build GRCh38
rs200950657 Chr.19: 15237495-15237495 on Build GRCh38
rs1029339859 Chr.19: 15237608-15237608 on Build GRCh38
rs1028057802 Chr.19: 15237715-15237715 on Build GRCh38
rs908793111 Chr.19: 15237763-15237763 on Build GRCh38
rs200086721 Chr.19: 15237773-15237773 on Build GRCh38
rs200880810 Chr.19: 15237624-15237624 on Build GRCh38
rs988586510 Chr.19: 15237387-15237387 on Build GRCh38
rs1054021952 Chr.19: 15237795-15237796 on Build GRCh38
rs562862448 Chr.19: 15237631-15237632 on Build GRCh38
rs1037219371 Chr.19: 15237732-15237732 on Build GRCh38
rs201620037 Chr.19: 15237631-15237631 on Build GRCh38
rs200335584 Chr.19: 15237480-15237480 on Build GRCh38
rs199726418 Chr.19: 15237499-15237499 on Build GRCh38
rs200736169 Chr.19: 15237525-15237525 on Build GRCh38
rs940215071 Chr.19: 15237424-15237424 on Build GRCh38
rs967802478 Chr.19: 15237698-15237698 on Build GRCh38
rs1022090482 Chr.19: 15237686-15237686 on Build GRCh38
rs199827334 Chr.19: 15237561-15237561 on Build GRCh38
rs920006948 Chr.19: 15237482-15237482 on Build GRCh38
rs564816332 Chr.19: 15237645-15237645 on Build GRCh38
rs202192143 Chr.19: 15237663-15237663 on Build GRCh38
rs888529250 Chr.19: 15237522-15237524 on Build GRCh38
rs201387076 Chr.19: 15237830-15237830 on Build GRCh38
rs199512390 Chr.19: 15237494-15237494 on Build GRCh38
rs201471124 Chr.19: 15237650-15237650 on Build GRCh38
rs1010363587 Chr.19: 15237835-15237836 on Build GRCh38
rs1009824554 Chr.19: 15237644-15237644 on Build GRCh38
rs920066846 Chr.19: 15237840-15237840 on Build GRCh38
rs200550709 Chr.19: 15237643-15237643 on Build GRCh38
rs941395050 Chr.19: 15237730-15237730 on Build GRCh38
rs776107705 Chr.19: 15237512-15237512 on Build GRCh38
rs1002814281 Chr.19: 15237500-15237500 on Build GRCh38
rs933629777 Chr.19: 15237402-15237402 on Build GRCh38
rs147277784 Chr.19: 15237406-15237406 on Build GRCh38
rs749354061 Chr.19: 15237767-15237767 on Build GRCh38
rs200697397 Chr.19: 15237820-15237820 on Build GRCh38
rs1032749180 Chr.19: 15237623-15237634 on Build GRCh38
rs201309790 Chr.19: 15237750-15237750 on Build GRCh38
rs892375099 Chr.19: 15237540-15237540 on Build GRCh38
rs201785680 Chr.19: 15237762-15237762 on Build GRCh38
rs982153875 Chr.19: 15237436-15237439 on Build GRCh38
rs201350886 Chr.19: 15237571-15237571 on Build GRCh38
rs998818737 Chr.19: 15237602-15237602 on Build GRCh38
rs201662899 Chr.19: 15237503-15237503 on Build GRCh38
rs201480234 Chr.19: 15237497-15237497 on Build GRCh38
rs988674712 Chr.19: 15237632-15237632 on Build GRCh38
rs1020514584 Chr.19: 15237632-15237636 on Build GRCh38
rs949980609 Chr.19: 15237411-15237411 on Build GRCh38
rs199828251 Chr.19: 15237724-15237724 on Build GRCh38
rs1038500798 Chr.19: 15237444-15237444 on Build GRCh38
rs200955284 Chr.19: 15237757-15237757 on Build GRCh38
rs892907417 Chr.19: 15237804-15237804 on Build GRCh38
rs116952107 Chr.19: 15237511-15237511 on Build GRCh38
rs974663828 Chr.19: 15237803-15237803 on Build GRCh38
rs545785918 Chr.19: 15237826-15237826 on Build GRCh38
rs957105175 Chr.19: 15237631-15237631 on Build GRCh38
rs199524751 Chr.19: 15237754-15237754 on Build GRCh38
rs201929816 Chr.19: 15237555-15237555 on Build GRCh38
rs544892393 Chr.19: 15237490-15237490 on Build GRCh38
rs934169564 Chr.19: 15237739-15237739 on Build GRCh38
rs768019295 Chr.19: 15237791-15237794 on Build GRCh38
rs971343012 Chr.19: 15237635-15237635 on Build GRCh38
rs1037591375 Chr.19: 15237544-15237544 on Build GRCh38
rs889484799 Chr.19: 15237621-15237629 on Build GRCh38
rs941531804 Chr.19: 15237543-15237543 on Build GRCh38
rs1006511863 Chr.19: 15237622-15237623 on Build GRCh38
rs200619207 Chr.19: 15237501-15237501 on Build GRCh38
rs201899245 Chr.19: 15237582-15237582 on Build GRCh38
rs908790387 Chr.19: 15237415-15237415 on Build GRCh38
rs1006541179 Chr.19: 15237780-15237780 on Build GRCh38
rs200093683 Chr.19: 15237836-15237836 on Build GRCh38
rs190600735 Chr.19: 15237714-15237714 on Build GRCh38
rs200068139 Chr.19: 15237592-15237592 on Build GRCh38
rs909994497 Chr.19: 15237726-15237726 on Build GRCh38
rs200753158 Chr.19: 15237736-15237736 on Build GRCh38
rs897417842 Chr.19: 15237586-15237591 on Build GRCh38

View Full Product Details