Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15237758-15238263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD4
Entrez Gene ID
23476
Gene Name
bromodomain containing 4
Gene Aliases
CAP, HUNK1, HUNKI, MCAP
UniGene
Hs.187763
Gene Chromosome Location
Chr.19: 15236836-15332543 on Build GRCh38
Transcripts
NM_058243.2 XM_011527854.1

SNP Detail

SNP ID Chromosome Location
rs199792504 Chr.19: 15237935-15237935 on Build GRCh38
rs903711317 Chr.19: 15237900-15237900 on Build GRCh38
rs566903761 Chr.19: 15237986-15237986 on Build GRCh38
rs995579787 Chr.19: 15238083-15238083 on Build GRCh38
rs971670731 Chr.19: 15237867-15237867 on Build GRCh38
rs550569511 Chr.19: 15237966-15237966 on Build GRCh38
rs201412762 Chr.19: 15237939-15237939 on Build GRCh38
rs1054021952 Chr.19: 15237795-15237796 on Build GRCh38
rs200086926 Chr.19: 15237960-15237960 on Build GRCh38
rs535929150 Chr.19: 15237987-15237987 on Build GRCh38
rs201386454 Chr.19: 15238196-15238196 on Build GRCh38
rs201913608 Chr.19: 15238179-15238179 on Build GRCh38
rs938742063 Chr.19: 15237953-15237953 on Build GRCh38
rs200116413 Chr.19: 15237976-15237976 on Build GRCh38
rs1034290856 Chr.19: 15237870-15237870 on Build GRCh38
rs1006057479 Chr.19: 15238202-15238205 on Build GRCh38
rs201387076 Chr.19: 15237830-15237830 on Build GRCh38
rs1016215015 Chr.19: 15238233-15238233 on Build GRCh38
rs200367617 Chr.19: 15238030-15238030 on Build GRCh38
rs1010363587 Chr.19: 15237835-15237836 on Build GRCh38
rs920066846 Chr.19: 15237840-15237840 on Build GRCh38
rs2286019 Chr.19: 15238091-15238091 on Build GRCh38
rs955071577 Chr.19: 15237906-15237906 on Build GRCh38
rs200378959 Chr.19: 15237931-15237931 on Build GRCh38
rs1026180941 Chr.19: 15238031-15238031 on Build GRCh38
rs200697397 Chr.19: 15237820-15237820 on Build GRCh38
rs200524246 Chr.19: 15237885-15237885 on Build GRCh38
rs201172236 Chr.19: 15238034-15238034 on Build GRCh38
rs200892899 Chr.19: 15238001-15238001 on Build GRCh38
rs1036823728 Chr.19: 15238197-15238197 on Build GRCh38
rs201442549 Chr.19: 15237863-15237863 on Build GRCh38
rs201030374 Chr.19: 15237888-15237888 on Build GRCh38
rs201938372 Chr.19: 15237892-15237892 on Build GRCh38
rs892907417 Chr.19: 15237804-15237804 on Build GRCh38
rs545785918 Chr.19: 15237826-15237826 on Build GRCh38
rs903765096 Chr.19: 15237988-15237988 on Build GRCh38
rs200681653 Chr.19: 15237945-15237945 on Build GRCh38
rs201599512 Chr.19: 15237950-15237950 on Build GRCh38
rs896843872 Chr.19: 15238080-15238080 on Build GRCh38
rs202015466 Chr.19: 15238169-15238169 on Build GRCh38
rs1006541179 Chr.19: 15237780-15237780 on Build GRCh38
rs200067207 Chr.19: 15238145-15238145 on Build GRCh38
rs943851915 Chr.19: 15238195-15238195 on Build GRCh38
rs199505733 Chr.19: 15238023-15238023 on Build GRCh38
rs199509209 Chr.19: 15238223-15238223 on Build GRCh38
rs200093683 Chr.19: 15237836-15237836 on Build GRCh38
rs201236634 Chr.19: 15238163-15238163 on Build GRCh38
rs200420468 Chr.19: 15237905-15237905 on Build GRCh38
rs995194192 Chr.19: 15238231-15238231 on Build GRCh38
rs202033132 Chr.19: 15237984-15237984 on Build GRCh38
rs567892805 Chr.19: 15237925-15237925 on Build GRCh38
rs199752874 Chr.19: 15238036-15238036 on Build GRCh38
rs890989168 Chr.19: 15238193-15238193 on Build GRCh38
rs896892337 Chr.19: 15238206-15238206 on Build GRCh38
rs201907959 Chr.19: 15237969-15237969 on Build GRCh38
rs948878358 Chr.19: 15237849-15237849 on Build GRCh38
rs200187809 Chr.19: 15237998-15237998 on Build GRCh38
rs996679489 Chr.19: 15237904-15237904 on Build GRCh38
rs1009464707 Chr.19: 15237890-15237890 on Build GRCh38
rs200272821 Chr.19: 15238174-15238174 on Build GRCh38
rs868413450 Chr.19: 15237838-15237838 on Build GRCh38
rs974380887 Chr.19: 15237938-15237938 on Build GRCh38
rs962828463 Chr.19: 15237887-15237887 on Build GRCh38
rs1007947523 Chr.19: 15238048-15238048 on Build GRCh38
rs199798128 Chr.19: 15238084-15238084 on Build GRCh38
rs201379224 Chr.19: 15238021-15238021 on Build GRCh38
rs183154747 Chr.19: 15238125-15238125 on Build GRCh38
rs980234094 Chr.19: 15237947-15237947 on Build GRCh38
rs200850854 Chr.19: 15238097-15238097 on Build GRCh38
rs201096850 Chr.19: 15237881-15237881 on Build GRCh38
rs202162163 Chr.19: 15238018-15238018 on Build GRCh38
rs200846538 Chr.19: 15237936-15237936 on Build GRCh38
rs199886125 Chr.19: 15238114-15238114 on Build GRCh38
rs986948585 Chr.19: 15238051-15238052 on Build GRCh38
rs928731751 Chr.19: 15237951-15237951 on Build GRCh38
rs113991798 Chr.19: 15238115-15238115 on Build GRCh38
rs201053103 Chr.19: 15237983-15237983 on Build GRCh38
rs201418824 Chr.19: 15237914-15237914 on Build GRCh38
rs980242317 Chr.19: 15238122-15238122 on Build GRCh38
rs199837307 Chr.19: 15237869-15237869 on Build GRCh38
rs199535844 Chr.19: 15237943-15237943 on Build GRCh38
rs201010946 Chr.19: 15238147-15238147 on Build GRCh38
rs200574105 Chr.19: 15238062-15238062 on Build GRCh38
rs199612127 Chr.19: 15237918-15237918 on Build GRCh38
rs201054777 Chr.19: 15238175-15238175 on Build GRCh38
rs772466769 Chr.19: 15237978-15237978 on Build GRCh38
rs1034153380 Chr.19: 15237979-15237979 on Build GRCh38
rs527735632 Chr.19: 15238126-15238126 on Build GRCh38
rs989237826 Chr.19: 15237954-15237954 on Build GRCh38
rs111874520 Chr.19: 15237893-15237893 on Build GRCh38
rs547162799 Chr.19: 15238205-15238205 on Build GRCh38
rs202161903 Chr.19: 15237882-15237882 on Build GRCh38
rs200844588 Chr.19: 15238244-15238244 on Build GRCh38
rs938718759 Chr.19: 15237875-15237875 on Build GRCh38
rs974663828 Chr.19: 15237803-15237803 on Build GRCh38
rs200341856 Chr.19: 15238180-15238180 on Build GRCh38
rs768019295 Chr.19: 15237791-15237794 on Build GRCh38
rs370301575 Chr.19: 15238188-15238188 on Build GRCh38
rs200119535 Chr.19: 15238151-15238151 on Build GRCh38
rs544828079 Chr.19: 15238046-15238046 on Build GRCh38
rs200895322 Chr.19: 15237858-15237858 on Build GRCh38
rs906957866 Chr.19: 15238152-15238152 on Build GRCh38
rs200527661 Chr.19: 15238019-15238019 on Build GRCh38
rs201460595 Chr.19: 15237932-15237932 on Build GRCh38
rs201072641 Chr.19: 15237968-15237968 on Build GRCh38
rs201897143 Chr.19: 15238063-15238063 on Build GRCh38
rs988933550 Chr.19: 15238146-15238146 on Build GRCh38
rs202092653 Chr.19: 15238150-15238150 on Build GRCh38
rs201667557 Chr.19: 15238099-15238099 on Build GRCh38

View Full Product Details