Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15238952-15239448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD4
Entrez Gene ID
23476
Gene Name
bromodomain containing 4
Gene Aliases
CAP, HUNK1, HUNKI, MCAP
UniGene
Hs.187763
Gene Chromosome Location
Chr.19: 15236836-15332543 on Build GRCh38
Transcripts
NM_058243.2 XM_011527854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4835958 c.3673G>A p.E1225K Chr.19: 15239168-15239168 on Build GRCh38
1391072 c.3629C>T p.P1210L Chr.19: 15239212-15239212 on Build GRCh38
6003528 c.3741C>T p.H1247H Chr.19: 15239100-15239100 on Build GRCh38
1325444 c.3772G>C p.E1258Q Chr.19: 15239069-15239069 on Build GRCh38
6979135 c.3688G>A p.A1230T Chr.19: 15239153-15239153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191065610 Chr.19: 15238985-15238985 on Build GRCh38
rs766976215 Chr.19: 15239238-15239238 on Build GRCh38
rs775209814 Chr.19: 15239154-15239154 on Build GRCh38
rs776249512 Chr.19: 15239209-15239209 on Build GRCh38
rs573999854 Chr.19: 15239318-15239318 on Build GRCh38
rs200683030 Chr.19: 15239021-15239021 on Build GRCh38
rs746206112 Chr.19: 15239026-15239026 on Build GRCh38
rs759445995 Chr.19: 15239000-15239000 on Build GRCh38
rs745807590 Chr.19: 15239168-15239168 on Build GRCh38
rs764870849 Chr.19: 15239283-15239283 on Build GRCh38
rs755419721 Chr.19: 15239268-15239268 on Build GRCh38
rs200730677 Chr.19: 15238984-15238984 on Build GRCh38
rs770961898 Chr.19: 15239063-15239063 on Build GRCh38
rs926195469 Chr.19: 15239273-15239273 on Build GRCh38
rs747757800 Chr.19: 15239046-15239046 on Build GRCh38
rs763100026 Chr.19: 15239094-15239094 on Build GRCh38
rs775717426 Chr.19: 15239079-15239079 on Build GRCh38
rs201209267 Chr.19: 15239106-15239106 on Build GRCh38
rs752887812 Chr.19: 15239006-15239006 on Build GRCh38
rs760849221 Chr.19: 15239357-15239357 on Build GRCh38
rs192715927 Chr.19: 15239366-15239366 on Build GRCh38
rs202177646 Chr.19: 15239261-15239261 on Build GRCh38
rs1034211332 Chr.19: 15239396-15239396 on Build GRCh38
rs771511153 Chr.19: 15238976-15238976 on Build GRCh38
rs766766367 Chr.19: 15238989-15238990 on Build GRCh38
rs148010154 Chr.19: 15239160-15239160 on Build GRCh38
rs201489245 Chr.19: 15239277-15239277 on Build GRCh38
rs781690964 Chr.19: 15239025-15239025 on Build GRCh38
rs202033213 Chr.19: 15239195-15239195 on Build GRCh38
rs756432777 Chr.19: 15239109-15239109 on Build GRCh38
rs201061856 Chr.19: 15239367-15239367 on Build GRCh38
rs199756709 Chr.19: 15239354-15239354 on Build GRCh38
rs751201371 Chr.19: 15239371-15239371 on Build GRCh38
rs200270814 Chr.19: 15239211-15239211 on Build GRCh38
rs769618538 Chr.19: 15239307-15239307 on Build GRCh38
rs200566874 Chr.19: 15239345-15239345 on Build GRCh38
rs772582937 Chr.19: 15239052-15239052 on Build GRCh38
rs757981891 Chr.19: 15239139-15239139 on Build GRCh38
rs199514099 Chr.19: 15239344-15239344 on Build GRCh38
rs201430449 Chr.19: 15238990-15238990 on Build GRCh38
rs974664417 Chr.19: 15239322-15239323 on Build GRCh38
rs760702028 Chr.19: 15238992-15238992 on Build GRCh38
rs761335076 Chr.19: 15238971-15238971 on Build GRCh38
rs745444017 Chr.19: 15239285-15239285 on Build GRCh38
rs867367622 Chr.19: 15239178-15239178 on Build GRCh38
rs141704077 Chr.19: 15239253-15239253 on Build GRCh38
rs536597957 Chr.19: 15239348-15239348 on Build GRCh38
rs200733287 Chr.19: 15239389-15239389 on Build GRCh38
rs769576955 Chr.19: 15239169-15239169 on Build GRCh38
rs1003096587 Chr.19: 15239049-15239049 on Build GRCh38
rs367705172 Chr.19: 15239310-15239310 on Build GRCh38
rs201932061 Chr.19: 15239401-15239401 on Build GRCh38
rs759358078 Chr.19: 15239075-15239075 on Build GRCh38
rs201682548 Chr.19: 15238993-15238993 on Build GRCh38
rs367944319 Chr.19: 15239100-15239100 on Build GRCh38
rs201653061 Chr.19: 15239136-15239136 on Build GRCh38
rs200078058 Chr.19: 15239099-15239099 on Build GRCh38
rs752009541 Chr.19: 15239017-15239017 on Build GRCh38
rs200491118 Chr.19: 15239270-15239270 on Build GRCh38
rs199536004 Chr.19: 15238994-15238994 on Build GRCh38
rs199836962 Chr.19: 15239374-15239374 on Build GRCh38
rs141774763 Chr.19: 15239098-15239098 on Build GRCh38
rs773134670 Chr.19: 15238991-15238991 on Build GRCh38
rs199833998 Chr.19: 15239417-15239417 on Build GRCh38
rs118000160 Chr.19: 15239151-15239151 on Build GRCh38
rs775025837 Chr.19: 15239327-15239327 on Build GRCh38
rs368540009 Chr.19: 15239402-15239402 on Build GRCh38
rs774575755 Chr.19: 15239203-15239203 on Build GRCh38
rs777608829 Chr.19: 15239147-15239147 on Build GRCh38
rs764974504 Chr.19: 15239076-15239076 on Build GRCh38
rs776909894 Chr.19: 15239274-15239275 on Build GRCh38
rs750870100 Chr.19: 15239228-15239228 on Build GRCh38
rs764438642 Chr.19: 15239097-15239097 on Build GRCh38
rs369998665 Chr.19: 15239028-15239028 on Build GRCh38
rs762877879 Chr.19: 15239179-15239179 on Build GRCh38
rs748982689 Chr.19: 15239315-15239315 on Build GRCh38
rs777722255 Chr.19: 15239014-15239014 on Build GRCh38
rs775138875 Chr.19: 15239177-15239177 on Build GRCh38
rs150535175 Chr.19: 15239409-15239409 on Build GRCh38
rs945737822 Chr.19: 15239247-15239247 on Build GRCh38
rs1023216999 Chr.19: 15239146-15239146 on Build GRCh38
rs200780861 Chr.19: 15239031-15239031 on Build GRCh38
rs374034348 Chr.19: 15239053-15239053 on Build GRCh38
rs187455857 Chr.19: 15239353-15239353 on Build GRCh38
rs756564257 Chr.19: 15239230-15239230 on Build GRCh38
rs759612692 Chr.19: 15239288-15239295 on Build GRCh38
rs370801637 Chr.19: 15239280-15239280 on Build GRCh38
rs201079722 Chr.19: 15239207-15239207 on Build GRCh38
rs376929445 Chr.19: 15239365-15239365 on Build GRCh38
rs758467136 Chr.19: 15239010-15239010 on Build GRCh38
rs780294079 Chr.19: 15239113-15239113 on Build GRCh38
rs765476238 Chr.19: 15239003-15239003 on Build GRCh38
rs947440360 Chr.19: 15239337-15239337 on Build GRCh38
rs183321935 Chr.19: 15239050-15239050 on Build GRCh38
rs564040267 Chr.19: 15239376-15239376 on Build GRCh38
rs748002758 Chr.19: 15239416-15239416 on Build GRCh38
rs1040899844 Chr.19: 15239334-15239334 on Build GRCh38
rs776759685 Chr.19: 15239067-15239067 on Build GRCh38
rs1034617603 Chr.19: 15239057-15239057 on Build GRCh38
rs373150587 Chr.19: 15239181-15239181 on Build GRCh38

View Full Product Details