Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15239581-15240086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD4
Entrez Gene ID
23476
Gene Name
bromodomain containing 4
Gene Aliases
CAP, HUNK1, HUNKI, MCAP
UniGene
Hs.187763
Gene Chromosome Location
Chr.19: 15236836-15332543 on Build GRCh38
Transcripts
NM_058243.2 XM_011527854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3529580 c.3445C>T p.R1149W Chr.19: 15239659-15239659 on Build GRCh38
4444652 c.3217A>G p.M1073V Chr.19: 15239975-15239975 on Build GRCh38
6932980 c.3211C>T p.P1071S Chr.19: 15239981-15239981 on Build GRCh38
7303838 c.3266T>C p.V1089A Chr.19: 15239926-15239926 on Build GRCh38
3422491 c.3197T>C p.L1066P Chr.19: 15239995-15239995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759440567 Chr.19: 15239987-15239987 on Build GRCh38
rs755212384 Chr.19: 15239610-15239610 on Build GRCh38
rs201671277 Chr.19: 15239680-15239680 on Build GRCh38
rs372021833 Chr.19: 15239786-15239786 on Build GRCh38
rs35676845 Chr.19: 15239814-15239814 on Build GRCh38
rs779611413 Chr.19: 15239842-15239842 on Build GRCh38
rs776518007 Chr.19: 15239906-15239906 on Build GRCh38
rs376743001 Chr.19: 15239706-15239706 on Build GRCh38
rs200799413 Chr.19: 15239890-15239890 on Build GRCh38
rs371555101 Chr.19: 15239862-15239862 on Build GRCh38
rs200957651 Chr.19: 15239674-15239674 on Build GRCh38
rs200400609 Chr.19: 15239846-15239846 on Build GRCh38
rs200327440 Chr.19: 15239868-15239868 on Build GRCh38
rs976384639 Chr.19: 15240015-15240015 on Build GRCh38
rs748633137 Chr.19: 15239721-15239721 on Build GRCh38
rs761598463 Chr.19: 15239693-15239693 on Build GRCh38
rs200501260 Chr.19: 15239850-15239850 on Build GRCh38
rs754011460 Chr.19: 15239792-15239792 on Build GRCh38
rs201646539 Chr.19: 15239803-15239803 on Build GRCh38
rs984889293 Chr.19: 15240063-15240063 on Build GRCh38
rs757573992 Chr.19: 15239888-15239888 on Build GRCh38
rs201908293 Chr.19: 15239997-15239997 on Build GRCh38
rs778881856 Chr.19: 15239804-15239804 on Build GRCh38
rs376162076 Chr.19: 15239716-15239716 on Build GRCh38
rs1032797771 Chr.19: 15239959-15239959 on Build GRCh38
rs572578556 Chr.19: 15239975-15239975 on Build GRCh38
rs375354544 Chr.19: 15239828-15239828 on Build GRCh38
rs772125141 Chr.19: 15239858-15239858 on Build GRCh38
rs567489459 Chr.19: 15239927-15239927 on Build GRCh38
rs771140069 Chr.19: 15239774-15239776 on Build GRCh38
rs556458475 Chr.19: 15239627-15239627 on Build GRCh38
rs199517872 Chr.19: 15239626-15239626 on Build GRCh38
rs757054890 Chr.19: 15239640-15239640 on Build GRCh38
rs762435043 Chr.19: 15239684-15239684 on Build GRCh38
rs199544374 Chr.19: 15239678-15239678 on Build GRCh38
rs750742416 Chr.19: 15240027-15240027 on Build GRCh38
rs200095218 Chr.19: 15239711-15239711 on Build GRCh38
rs200705995 Chr.19: 15239992-15239992 on Build GRCh38
rs377217681 Chr.19: 15239644-15239644 on Build GRCh38
rs773297674 Chr.19: 15239860-15239860 on Build GRCh38
rs547510807 Chr.19: 15239869-15239869 on Build GRCh38
rs369685357 Chr.19: 15239902-15239902 on Build GRCh38
rs995848451 Chr.19: 15239863-15239864 on Build GRCh38
rs200850373 Chr.19: 15239638-15239638 on Build GRCh38
rs908131984 Chr.19: 15240057-15240057 on Build GRCh38
rs777398745 Chr.19: 15240043-15240043 on Build GRCh38
rs762152914 Chr.19: 15239615-15239615 on Build GRCh38
rs765535087 Chr.19: 15239866-15239866 on Build GRCh38
rs1026979615 Chr.19: 15240019-15240019 on Build GRCh38
rs776258180 Chr.19: 15239978-15239978 on Build GRCh38
rs757629079 Chr.19: 15239833-15239833 on Build GRCh38
rs746128795 Chr.19: 15239647-15239647 on Build GRCh38
rs143993362 Chr.19: 15239990-15239990 on Build GRCh38
rs201100073 Chr.19: 15239762-15239762 on Build GRCh38
rs540369634 Chr.19: 15239694-15239694 on Build GRCh38
rs200902225 Chr.19: 15240011-15240011 on Build GRCh38
rs573909313 Chr.19: 15239738-15239738 on Build GRCh38
rs761637834 Chr.19: 15239775-15239775 on Build GRCh38
rs200026456 Chr.19: 15239864-15239864 on Build GRCh38
rs764075971 Chr.19: 15239685-15239685 on Build GRCh38
rs775177082 Chr.19: 15239757-15239757 on Build GRCh38
rs754585456 Chr.19: 15239905-15239905 on Build GRCh38
rs752061417 Chr.19: 15239881-15239881 on Build GRCh38
rs774749936 Chr.19: 15239611-15239611 on Build GRCh38
rs768141679 Chr.19: 15240014-15240014 on Build GRCh38
rs148664288 Chr.19: 15240013-15240013 on Build GRCh38
rs187907043 Chr.19: 15239768-15239768 on Build GRCh38
rs976668161 Chr.19: 15239723-15239723 on Build GRCh38
rs1055671896 Chr.19: 15239623-15239623 on Build GRCh38
rs199530457 Chr.19: 15239884-15239884 on Build GRCh38
rs747605396 Chr.19: 15239921-15239921 on Build GRCh38
rs749421543 Chr.19: 15239650-15239650 on Build GRCh38
rs780262620 Chr.19: 15239649-15239649 on Build GRCh38
rs772188667 Chr.19: 15239642-15239643 on Build GRCh38
rs1007248843 Chr.19: 15239979-15239979 on Build GRCh38
rs376613701 Chr.19: 15240012-15240012 on Build GRCh38
rs1044584643 Chr.19: 15239772-15239772 on Build GRCh38
rs930830089 Chr.19: 15240017-15240017 on Build GRCh38
rs146449278 Chr.19: 15240040-15240040 on Build GRCh38
rs201082202 Chr.19: 15239840-15239840 on Build GRCh38
rs200345800 Chr.19: 15239617-15239617 on Build GRCh38
rs200882606 Chr.19: 15239785-15239785 on Build GRCh38
rs1047396764 Chr.19: 15239671-15239671 on Build GRCh38
rs778364833 Chr.19: 15240030-15240030 on Build GRCh38
rs201935715 Chr.19: 15239733-15239733 on Build GRCh38
rs553993519 Chr.19: 15239700-15239700 on Build GRCh38
rs767605554 Chr.19: 15239780-15239780 on Build GRCh38
rs11669901 Chr.19: 15239783-15239783 on Build GRCh38
rs201578775 Chr.19: 15239675-15239675 on Build GRCh38
rs201317873 Chr.19: 15239872-15239872 on Build GRCh38
rs778688396 Chr.19: 15239920-15239920 on Build GRCh38
rs1046048062 Chr.19: 15239878-15239878 on Build GRCh38
rs967545322 Chr.19: 15239682-15239682 on Build GRCh38
rs781392567 Chr.19: 15239898-15239898 on Build GRCh38
rs570161414 Chr.19: 15239631-15239631 on Build GRCh38
rs201477031 Chr.19: 15239819-15239819 on Build GRCh38
rs762462881 Chr.19: 15239758-15239758 on Build GRCh38
rs772431443 Chr.19: 15239737-15239737 on Build GRCh38
rs201460342 Chr.19: 15239847-15239847 on Build GRCh38
rs202076913 Chr.19: 15239942-15239942 on Build GRCh38
rs200703650 Chr.19: 15239715-15239715 on Build GRCh38
rs370451418 Chr.19: 15239652-15239652 on Build GRCh38
rs199727585 Chr.19: 15239974-15239974 on Build GRCh38
rs747411770 Chr.19: 15239747-15239747 on Build GRCh38
rs373309106 Chr.19: 15239928-15239928 on Build GRCh38
rs773269235 Chr.19: 15239761-15239763 on Build GRCh38
rs920724739 Chr.19: 15239776-15239776 on Build GRCh38
rs747153554 Chr.19: 15239765-15239767 on Build GRCh38
rs1032745641 Chr.19: 15239690-15239690 on Build GRCh38
rs367680758 Chr.19: 15239883-15239883 on Build GRCh38
rs201844945 Chr.19: 15239843-15239843 on Build GRCh38
rs201451134 Chr.19: 15240039-15240039 on Build GRCh38
rs759208540 Chr.19: 15239676-15239676 on Build GRCh38
rs200117669 Chr.19: 15239998-15239998 on Build GRCh38
rs778317398 Chr.19: 15239816-15239816 on Build GRCh38
rs779348570 Chr.19: 15239659-15239659 on Build GRCh38
rs144097229 Chr.19: 15239970-15239970 on Build GRCh38
rs759824399 Chr.19: 15240057-15240057 on Build GRCh38
rs199839337 Chr.19: 15239743-15239743 on Build GRCh38
rs200628536 Chr.19: 15239679-15239679 on Build GRCh38
rs747867996 Chr.19: 15239617-15239618 on Build GRCh38
rs199735840 Chr.19: 15239624-15239624 on Build GRCh38
rs368982771 Chr.19: 15239618-15239618 on Build GRCh38
rs779410862 Chr.19: 15239720-15239720 on Build GRCh38
rs751462158 Chr.19: 15239686-15239686 on Build GRCh38
rs756217610 Chr.19: 15239648-15239648 on Build GRCh38

View Full Product Details