Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15680270-15680757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F12
Entrez Gene ID
66002
Gene Name
cytochrome P450 family 4 subfamily F member 12
Gene Aliases
CYPIVF12, F22329_1
UniGene
Hs.131459
Gene Chromosome Location
Chr.19: 15673018-15698819 on Build GRCh38
Transcripts
NM_023944.3 NR_117085.1 XM_006722850.3 XM_011528202.2 XM_011528203.2 XM_011528204.2 XM_011528205.2 XM_011528207.2 XM_011528208.2 XM_017027172.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3283998 c.456G>A p.T152T Chr.19: 15680450-15680450 on Build GRCh38
3822190 c.491T>C p.I164T Chr.19: 15680485-15680485 on Build GRCh38
5598204 c.381C>T p.F127F Chr.19: 15680281-15680281 on Build GRCh38
7211864 c.498C>G p.I166M Chr.19: 15680492-15680492 on Build GRCh38
709647 c.455C>T p.T152M Chr.19: 15680449-15680449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954142297 Chr.19: 15680578-15680578 on Build GRCh38
rs372150293 Chr.19: 15680337-15680337 on Build GRCh38
rs536209285 Chr.19: 15680705-15680705 on Build GRCh38
rs377107512 Chr.19: 15680437-15680437 on Build GRCh38
rs386807228 Chr.19: 15680400-15680402 on Build GRCh38
rs201073886 Chr.19: 15680414-15680414 on Build GRCh38
rs192633367 Chr.19: 15680481-15680481 on Build GRCh38
rs147972597 Chr.19: 15680389-15680389 on Build GRCh38
rs754352800 Chr.19: 15680600-15680600 on Build GRCh38
rs770504560 Chr.19: 15680618-15680618 on Build GRCh38
rs927960898 Chr.19: 15680612-15680612 on Build GRCh38
rs566080636 Chr.19: 15680548-15680548 on Build GRCh38
rs774998075 Chr.19: 15680571-15680571 on Build GRCh38
rs762783755 Chr.19: 15680570-15680570 on Build GRCh38
rs573565787 Chr.19: 15680469-15680469 on Build GRCh38
rs767869616 Chr.19: 15680588-15680588 on Build GRCh38
rs879526751 Chr.19: 15680675-15680675 on Build GRCh38
rs756590184 Chr.19: 15680503-15680503 on Build GRCh38
rs745331588 Chr.19: 15680632-15680632 on Build GRCh38
rs780946073 Chr.19: 15680312-15680312 on Build GRCh38
rs969444395 Chr.19: 15680606-15680606 on Build GRCh38
rs567598197 Chr.19: 15680398-15680398 on Build GRCh38
rs761590121 Chr.19: 15680403-15680403 on Build GRCh38
rs768805870 Chr.19: 15680324-15680324 on Build GRCh38
rs770246280 Chr.19: 15680401-15680401 on Build GRCh38
rs116266410 Chr.19: 15680341-15680341 on Build GRCh38
rs372624515 Chr.19: 15680484-15680484 on Build GRCh38
rs553958185 Chr.19: 15680547-15680547 on Build GRCh38
rs371537480 Chr.19: 15680350-15680350 on Build GRCh38
rs750351950 Chr.19: 15680367-15680367 on Build GRCh38
rs201807535 Chr.19: 15680441-15680441 on Build GRCh38
rs367934794 Chr.19: 15680333-15680333 on Build GRCh38
rs148891614 Chr.19: 15680368-15680368 on Build GRCh38
rs998249113 Chr.19: 15680470-15680470 on Build GRCh38
rs868644208 Chr.19: 15680351-15680351 on Build GRCh38
rs755305719 Chr.19: 15680604-15680604 on Build GRCh38
rs8100667 Chr.19: 15680370-15680370 on Build GRCh38
rs530094833 Chr.19: 15680399-15680399 on Build GRCh38
rs758590070 Chr.19: 15680423-15680423 on Build GRCh38
rs375594895 Chr.19: 15680388-15680388 on Build GRCh38
rs780433013 Chr.19: 15680517-15680517 on Build GRCh38
rs1017896805 Chr.19: 15680363-15680363 on Build GRCh38
rs766919632 Chr.19: 15680408-15680408 on Build GRCh38
rs756826295 Chr.19: 15680305-15680305 on Build GRCh38
rs187651397 Chr.19: 15680378-15680378 on Build GRCh38
rs143608773 Chr.19: 15680387-15680387 on Build GRCh38
rs982533782 Chr.19: 15680609-15680609 on Build GRCh38
rs376384018 Chr.19: 15680496-15680496 on Build GRCh38
rs769032711 Chr.19: 15680529-15680529 on Build GRCh38
rs752469422 Chr.19: 15680472-15680472 on Build GRCh38
rs751823223 Chr.19: 15680573-15680573 on Build GRCh38
rs763726603 Chr.19: 15680324-15680324 on Build GRCh38
rs771685280 Chr.19: 15680440-15680440 on Build GRCh38
rs772647986 Chr.19: 15680410-15680410 on Build GRCh38
rs556006739 Chr.19: 15680488-15680488 on Build GRCh38
rs769234053 Chr.19: 15680634-15680634 on Build GRCh38
rs386807227 Chr.19: 15680341-15680343 on Build GRCh38
rs749623963 Chr.19: 15680439-15680439 on Build GRCh38
rs750413369 Chr.19: 15680320-15680320 on Build GRCh38
rs767742680 Chr.19: 15680703-15680703 on Build GRCh38
rs373355202 Chr.19: 15680450-15680450 on Build GRCh38
rs754831729 Chr.19: 15680380-15680380 on Build GRCh38
rs751251110 Chr.19: 15680352-15680352 on Build GRCh38
rs775464004 Chr.19: 15680636-15680636 on Build GRCh38
rs535388110 Chr.19: 15680430-15680430 on Build GRCh38
rs138277223 Chr.19: 15680670-15680670 on Build GRCh38
rs778147286 Chr.19: 15680426-15680426 on Build GRCh38
rs2074568 Chr.19: 15680322-15680322 on Build GRCh38
rs375976293 Chr.19: 15680415-15680415 on Build GRCh38
rs564765640 Chr.19: 15680679-15680679 on Build GRCh38
rs866922722 Chr.19: 15680369-15680369 on Build GRCh38
rs761687729 Chr.19: 15680392-15680396 on Build GRCh38
rs766411170 Chr.19: 15680727-15680727 on Build GRCh38
rs750453342 Chr.19: 15680589-15680589 on Build GRCh38
rs369172315 Chr.19: 15680449-15680449 on Build GRCh38
rs138713818 Chr.19: 15680400-15680400 on Build GRCh38
rs768553860 Chr.19: 15680664-15680664 on Build GRCh38
rs763048040 Chr.19: 15680653-15680653 on Build GRCh38
rs1029303921 Chr.19: 15680474-15680474 on Build GRCh38
rs115910547 Chr.19: 15680327-15680327 on Build GRCh38
rs764017281 Chr.19: 15680572-15680572 on Build GRCh38
rs761210668 Chr.19: 15680359-15680359 on Build GRCh38
rs776517845 Chr.19: 15680454-15680454 on Build GRCh38
rs765101244 Chr.19: 15680471-15680471 on Build GRCh38
rs199991130 Chr.19: 15680453-15680453 on Build GRCh38
rs201727887 Chr.19: 15680431-15680431 on Build GRCh38
rs377219094 Chr.19: 15680366-15680366 on Build GRCh38
rs149631245 Chr.19: 15680722-15680722 on Build GRCh38
rs762180054 Chr.19: 15680583-15680583 on Build GRCh38
rs763822619 Chr.19: 15680293-15680293 on Build GRCh38
rs374385810 Chr.19: 15680677-15680677 on Build GRCh38
rs762799959 Chr.19: 15680318-15680337 on Build GRCh38
rs749627356 Chr.19: 15680518-15680518 on Build GRCh38
rs752849351 Chr.19: 15680421-15680421 on Build GRCh38
rs115514058 Chr.19: 15680343-15680343 on Build GRCh38
rs754706185 Chr.19: 15680417-15680417 on Build GRCh38
rs145798366 Chr.19: 15680396-15680396 on Build GRCh38
rs780737722 Chr.19: 15680626-15680626 on Build GRCh38
rs201190186 Chr.19: 15680534-15680534 on Build GRCh38
rs775310967 Chr.19: 15680558-15680558 on Build GRCh38
rs746705007 Chr.19: 15680535-15680535 on Build GRCh38
rs146729462 Chr.19: 15680414-15680415 on Build GRCh38
rs750969888 Chr.19: 15680300-15680300 on Build GRCh38
rs771409674 Chr.19: 15680338-15680338 on Build GRCh38
rs201942691 Chr.19: 15680543-15680543 on Build GRCh38
rs751525227 Chr.19: 15680390-15680392 on Build GRCh38
rs769729293 Chr.19: 15680556-15680556 on Build GRCh38
rs781759781 Chr.19: 15680434-15680434 on Build GRCh38
rs140731978 Chr.19: 15680393-15680393 on Build GRCh38
rs374559031 Chr.19: 15680420-15680420 on Build GRCh38
rs371223441 Chr.19: 15680560-15680560 on Build GRCh38
rs772214781 Chr.19: 15680335-15680335 on Build GRCh38
rs147497103 Chr.19: 15680598-15680598 on Build GRCh38
rs770423243 Chr.19: 15680540-15680540 on Build GRCh38
rs756182056 Chr.19: 15680596-15680596 on Build GRCh38
rs61731188 Chr.19: 15680402-15680402 on Build GRCh38
rs746651729 Chr.19: 15680616-15680616 on Build GRCh38
rs141478890 Chr.19: 15680489-15680489 on Build GRCh38
rs539254205 Chr.19: 15680412-15680412 on Build GRCh38
rs555713012 Chr.19: 15680629-15680629 on Build GRCh38
rs965838651 Chr.19: 15680303-15680303 on Build GRCh38
rs746430660 Chr.19: 15680436-15680436 on Build GRCh38
rs748889613 Chr.19: 15680689-15680689 on Build GRCh38

View Full Product Details