Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16502673-16503217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf44
Entrez Gene ID
84167
Gene Name
chromosome 19 open reading frame 44
Gene Aliases
-
UniGene
Hs.631627
Gene Chromosome Location
Chr.19: 16496311-16521369 on Build GRCh38
Transcripts
NM_001288834.1 NM_032207.3 XM_006722920.3 XM_011528354.2 XM_017027350.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3284658 c.795C>T p.S265S Chr.19: 16503100-16503100 on Build GRCh38
148528 c.877C>T p.R293C Chr.19: 16503182-16503182 on Build GRCh38
1290891 c.771A>G p.Q257Q Chr.19: 16503076-16503076 on Build GRCh38
3530029 c.911C>T p.S304F Chr.19: 16503216-16503216 on Build GRCh38
4075164 c.791C>A p.P264H Chr.19: 16503096-16503096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201337843 Chr.19: 16503133-16503133 on Build GRCh38
rs760184451 Chr.19: 16503107-16503108 on Build GRCh38
rs913046744 Chr.19: 16502851-16502851 on Build GRCh38
rs148082583 Chr.19: 16503085-16503085 on Build GRCh38
rs891345509 Chr.19: 16502997-16502997 on Build GRCh38
rs200951543 Chr.19: 16503100-16503100 on Build GRCh38
rs150685247 Chr.19: 16503186-16503186 on Build GRCh38
rs8111282 Chr.19: 16502922-16502922 on Build GRCh38
rs200824240 Chr.19: 16502825-16502825 on Build GRCh38
rs1026323383 Chr.19: 16503164-16503165 on Build GRCh38
rs776040312 Chr.19: 16503176-16503176 on Build GRCh38
rs777127184 Chr.19: 16503068-16503068 on Build GRCh38
rs375144704 Chr.19: 16502824-16502824 on Build GRCh38
rs944280393 Chr.19: 16502999-16502999 on Build GRCh38
rs199764040 Chr.19: 16503182-16503182 on Build GRCh38
rs1045227155 Chr.19: 16503109-16503109 on Build GRCh38
rs754834760 Chr.19: 16503146-16503146 on Build GRCh38
rs778880692 Chr.19: 16503148-16503148 on Build GRCh38
rs750082780 Chr.19: 16503117-16503117 on Build GRCh38
rs947277649 Chr.19: 16502854-16502854 on Build GRCh38
rs111546920 Chr.19: 16503092-16503092 on Build GRCh38
rs766896291 Chr.19: 16503014-16503015 on Build GRCh38
rs770993395 Chr.19: 16503056-16503056 on Build GRCh38
rs781082813 Chr.19: 16503169-16503169 on Build GRCh38
rs758176968 Chr.19: 16503155-16503155 on Build GRCh38
rs8111141 Chr.19: 16502804-16502804 on Build GRCh38
rs142017069 Chr.19: 16503158-16503158 on Build GRCh38
rs777950896 Chr.19: 16503032-16503032 on Build GRCh38
rs573273210 Chr.19: 16502711-16502711 on Build GRCh38
rs570640890 Chr.19: 16502944-16502944 on Build GRCh38
rs181968124 Chr.19: 16502973-16502973 on Build GRCh38
rs61130725 Chr.19: 16502987-16502987 on Build GRCh38
rs12232822 Chr.19: 16503130-16503130 on Build GRCh38
rs970032813 Chr.19: 16502725-16502725 on Build GRCh38
rs866255904 Chr.19: 16502992-16502992 on Build GRCh38
rs752485439 Chr.19: 16503150-16503150 on Build GRCh38
rs897297606 Chr.19: 16503052-16503052 on Build GRCh38
rs988625571 Chr.19: 16502790-16502790 on Build GRCh38
rs761563346 Chr.19: 16503141-16503141 on Build GRCh38
rs1027093644 Chr.19: 16502705-16502705 on Build GRCh38
rs771011628 Chr.19: 16503168-16503168 on Build GRCh38
rs1037325944 Chr.19: 16503013-16503013 on Build GRCh38
rs531540207 Chr.19: 16502788-16502788 on Build GRCh38
rs1035700068 Chr.19: 16503132-16503132 on Build GRCh38
rs763343071 Chr.19: 16503105-16503105 on Build GRCh38
rs980110901 Chr.19: 16502728-16502728 on Build GRCh38
rs542228989 Chr.19: 16502737-16502737 on Build GRCh38
rs959946149 Chr.19: 16502761-16502761 on Build GRCh38
rs775675044 Chr.19: 16503101-16503101 on Build GRCh38
rs921700664 Chr.19: 16502930-16502930 on Build GRCh38
rs575239426 Chr.19: 16502946-16502946 on Build GRCh38
rs772321987 Chr.19: 16503014-16503014 on Build GRCh38
rs372928987 Chr.19: 16503048-16503048 on Build GRCh38
rs996172063 Chr.19: 16502730-16502730 on Build GRCh38
rs760358231 Chr.19: 16503189-16503189 on Build GRCh38
rs953141484 Chr.19: 16503194-16503194 on Build GRCh38
rs760270571 Chr.19: 16503121-16503121 on Build GRCh38
rs76297817 Chr.19: 16502745-16502745 on Build GRCh38
rs567728269 Chr.19: 16502994-16502994 on Build GRCh38
rs767323492 Chr.19: 16503115-16503115 on Build GRCh38
rs796725569 Chr.19: 16502998-16502998 on Build GRCh38
rs527436104 Chr.19: 16502923-16502923 on Build GRCh38
rs769134219 Chr.19: 16503183-16503183 on Build GRCh38
rs753839919 Chr.19: 16503144-16503144 on Build GRCh38
rs754238674 Chr.19: 16503054-16503055 on Build GRCh38
rs549936231 Chr.19: 16503163-16503163 on Build GRCh38
rs1018640910 Chr.19: 16502816-16502816 on Build GRCh38
rs965807229 Chr.19: 16502875-16502875 on Build GRCh38
rs746416232 Chr.19: 16503170-16503170 on Build GRCh38
rs757253497 Chr.19: 16502716-16502716 on Build GRCh38
rs1010496025 Chr.19: 16502824-16502824 on Build GRCh38
rs979777302 Chr.19: 16502998-16502998 on Build GRCh38
rs747150057 Chr.19: 16503050-16503050 on Build GRCh38

View Full Product Details