Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16509637-16510127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf44
Entrez Gene ID
84167
Gene Name
chromosome 19 open reading frame 44
Gene Aliases
-
UniGene
Hs.631627
Gene Chromosome Location
Chr.19: 16496311-16521369 on Build GRCh38
Transcripts
NM_001288834.1 NM_032207.3 XM_006722920.3 XM_011528354.2 XM_017027350.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5499356 c.1299G>A p.S433S Chr.19: 16509648-16509648 on Build GRCh38
992475 c.1566C>T p.H522H Chr.19: 16509915-16509915 on Build GRCh38
1471090 c.1385A>G p.N462S Chr.19: 16509734-16509734 on Build GRCh38
4642270 c.1316G>C p.S439T Chr.19: 16509665-16509665 on Build GRCh38
5942809 c.1376C>T p.P459L Chr.19: 16509725-16509725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372750890 Chr.19: 16510014-16510014 on Build GRCh38
rs540886492 Chr.19: 16509754-16509754 on Build GRCh38
rs746626897 Chr.19: 16510022-16510022 on Build GRCh38
rs758660986 Chr.19: 16509880-16509880 on Build GRCh38
rs763818017 Chr.19: 16510049-16510049 on Build GRCh38
rs377201342 Chr.19: 16509670-16509670 on Build GRCh38
rs1048380810 Chr.19: 16509902-16509902 on Build GRCh38
rs759992419 Chr.19: 16509828-16509828 on Build GRCh38
rs747081260 Chr.19: 16509759-16509759 on Build GRCh38
rs531527946 Chr.19: 16509686-16509686 on Build GRCh38
rs562591336 Chr.19: 16509668-16509668 on Build GRCh38
rs771360150 Chr.19: 16509685-16509685 on Build GRCh38
rs750562390 Chr.19: 16510066-16510066 on Build GRCh38
rs754801069 Chr.19: 16509740-16509740 on Build GRCh38
rs777951084 Chr.19: 16509888-16509888 on Build GRCh38
rs140516141 Chr.19: 16509704-16509704 on Build GRCh38
rs772162433 Chr.19: 16509755-16509755 on Build GRCh38
rs908826916 Chr.19: 16509827-16509827 on Build GRCh38
rs777180074 Chr.19: 16509781-16509781 on Build GRCh38
rs375608776 Chr.19: 16509923-16509923 on Build GRCh38
rs142240474 Chr.19: 16509975-16509975 on Build GRCh38
rs780714279 Chr.19: 16510092-16510092 on Build GRCh38
rs373701349 Chr.19: 16510039-16510039 on Build GRCh38
rs200961956 Chr.19: 16509953-16509953 on Build GRCh38
rs761411932 Chr.19: 16509713-16509713 on Build GRCh38
rs769078809 Chr.19: 16510013-16510013 on Build GRCh38
rs767600030 Chr.19: 16510061-16510061 on Build GRCh38
rs769661033 Chr.19: 16509893-16509893 on Build GRCh38
rs200982019 Chr.19: 16509803-16509803 on Build GRCh38
rs368527845 Chr.19: 16509801-16509801 on Build GRCh38
rs992592293 Chr.19: 16509727-16509727 on Build GRCh38
rs775891707 Chr.19: 16509841-16509842 on Build GRCh38
rs755962723 Chr.19: 16509731-16509731 on Build GRCh38
rs376553711 Chr.19: 16510037-16510037 on Build GRCh38
rs529997239 Chr.19: 16509703-16509703 on Build GRCh38
rs766122582 Chr.19: 16509994-16510002 on Build GRCh38
rs770380454 Chr.19: 16510023-16510023 on Build GRCh38
rs755419197 Chr.19: 16510096-16510096 on Build GRCh38
rs751736465 Chr.19: 16509983-16509983 on Build GRCh38
rs978521433 Chr.19: 16509989-16509989 on Build GRCh38
rs766296594 Chr.19: 16509836-16509836 on Build GRCh38
rs538831832 Chr.19: 16509985-16509985 on Build GRCh38
rs765129357 Chr.19: 16509972-16509972 on Build GRCh38
rs765588228 Chr.19: 16509838-16509839 on Build GRCh38
rs763484094 Chr.19: 16509886-16509887 on Build GRCh38
rs772886038 Chr.19: 16509829-16509829 on Build GRCh38
rs761121917 Chr.19: 16509682-16509682 on Build GRCh38
rs757352107 Chr.19: 16509890-16509890 on Build GRCh38
rs759920607 Chr.19: 16509699-16509699 on Build GRCh38
rs201070169 Chr.19: 16509833-16509833 on Build GRCh38
rs766987060 Chr.19: 16509986-16509987 on Build GRCh38
rs200035622 Chr.19: 16509869-16509869 on Build GRCh38
rs139036992 Chr.19: 16509954-16509954 on Build GRCh38
rs1036045941 Chr.19: 16509916-16509916 on Build GRCh38
rs761503718 Chr.19: 16509811-16509811 on Build GRCh38
rs150419921 Chr.19: 16509786-16509786 on Build GRCh38
rs776923708 Chr.19: 16509734-16509735 on Build GRCh38
rs762739951 Chr.19: 16510044-16510044 on Build GRCh38
rs931543726 Chr.19: 16509809-16509809 on Build GRCh38
rs771896382 Chr.19: 16509927-16509927 on Build GRCh38
rs529571469 Chr.19: 16510094-16510094 on Build GRCh38
rs772916260 Chr.19: 16509929-16509929 on Build GRCh38
rs746328167 Chr.19: 16509906-16509906 on Build GRCh38
rs766025646 Chr.19: 16510095-16510096 on Build GRCh38
rs759479233 Chr.19: 16509965-16509965 on Build GRCh38
rs78068439 Chr.19: 16509883-16509883 on Build GRCh38
rs557457298 Chr.19: 16510007-16510007 on Build GRCh38
rs750403686 Chr.19: 16509728-16509728 on Build GRCh38
rs756246452 Chr.19: 16510067-16510067 on Build GRCh38
rs202160274 Chr.19: 16509889-16509889 on Build GRCh38
rs58659693 Chr.19: 16509905-16509905 on Build GRCh38
rs138160349 Chr.19: 16509866-16509866 on Build GRCh38
rs767164051 Chr.19: 16509812-16509812 on Build GRCh38
rs749739462 Chr.19: 16510012-16510012 on Build GRCh38
rs370262229 Chr.19: 16510038-16510038 on Build GRCh38
rs762052186 Chr.19: 16510050-16510050 on Build GRCh38
rs770129493 Chr.19: 16509787-16509787 on Build GRCh38
rs749637051 Chr.19: 16509924-16509924 on Build GRCh38
rs201946733 Chr.19: 16509661-16509661 on Build GRCh38
rs369614715 Chr.19: 16509981-16509981 on Build GRCh38
rs746388656 Chr.19: 16510005-16510005 on Build GRCh38
rs374870382 Chr.19: 16509756-16509756 on Build GRCh38
rs760222715 Chr.19: 16509835-16509835 on Build GRCh38
rs752948221 Chr.19: 16509872-16509872 on Build GRCh38
rs755832391 Chr.19: 16509997-16509998 on Build GRCh38
rs145166150 Chr.19: 16509915-16509915 on Build GRCh38
rs774156051 Chr.19: 16509802-16509802 on Build GRCh38
rs753636800 Chr.19: 16509737-16509737 on Build GRCh38
rs370079916 Chr.19: 16509735-16509735 on Build GRCh38
rs771390136 Chr.19: 16509776-16509776 on Build GRCh38
rs773722664 Chr.19: 16509674-16509674 on Build GRCh38
rs767074023 Chr.19: 16509724-16509724 on Build GRCh38
rs569954503 Chr.19: 16509976-16509976 on Build GRCh38
rs753746894 Chr.19: 16509850-16509850 on Build GRCh38
rs760109719 Chr.19: 16509946-16509946 on Build GRCh38
rs781280277 Chr.19: 16510004-16510004 on Build GRCh38

View Full Product Details