Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17276399-17276900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BABAM1
Entrez Gene ID
29086
Gene Name
BRISC and BRCA1 A complex member 1
Gene Aliases
C19orf62, HSPC142, MERIT40, NBA1
UniGene
Hs.190722
Gene Chromosome Location
Chr.19: 17267376-17279353 on Build GRCh38
Transcripts
NM_001033549.2 NM_001288756.1 NM_001288757.1 NM_014173.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5174851 c.621G>A p.P207P Chr.19: 17276546-17276546 on Build GRCh38
5167074 c.655C>T p.R219C Chr.19: 17276580-17276580 on Build GRCh38
5451510 c.656G>A p.R219H Chr.19: 17276581-17276581 on Build GRCh38
3530305 c.642C>T p.I214I Chr.19: 17276567-17276567 on Build GRCh38
3285777 c.635G>A p.R212H Chr.19: 17276560-17276560 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768958596 Chr.19: 17276505-17276505 on Build GRCh38
rs184899067 Chr.19: 17276737-17276737 on Build GRCh38
rs750588177 Chr.19: 17276699-17276699 on Build GRCh38
rs534927439 Chr.19: 17276484-17276484 on Build GRCh38
rs769543927 Chr.19: 17276769-17276769 on Build GRCh38
rs773632474 Chr.19: 17276731-17276731 on Build GRCh38
rs10416551 Chr.19: 17276776-17276776 on Build GRCh38
rs762756608 Chr.19: 17276848-17276848 on Build GRCh38
rs374131612 Chr.19: 17276515-17276515 on Build GRCh38
rs775834500 Chr.19: 17276534-17276534 on Build GRCh38
rs201104012 Chr.19: 17276856-17276856 on Build GRCh38
rs749960676 Chr.19: 17276861-17276861 on Build GRCh38
rs546323439 Chr.19: 17276516-17276516 on Build GRCh38
rs751463325 Chr.19: 17276855-17276855 on Build GRCh38
rs544049922 Chr.19: 17276694-17276694 on Build GRCh38
rs1020875907 Chr.19: 17276549-17276549 on Build GRCh38
rs779490101 Chr.19: 17276806-17276810 on Build GRCh38
rs549514438 Chr.19: 17276437-17276437 on Build GRCh38
rs746164566 Chr.19: 17276712-17276712 on Build GRCh38
rs754879017 Chr.19: 17276483-17276483 on Build GRCh38
rs555163333 Chr.19: 17276545-17276545 on Build GRCh38
rs775022844 Chr.19: 17276791-17276791 on Build GRCh38
rs752598328 Chr.19: 17276711-17276711 on Build GRCh38
rs762016577 Chr.19: 17276689-17276689 on Build GRCh38
rs987269280 Chr.19: 17276667-17276667 on Build GRCh38
rs777717351 Chr.19: 17276715-17276715 on Build GRCh38
rs528538382 Chr.19: 17276811-17276811 on Build GRCh38
rs557124977 Chr.19: 17276701-17276701 on Build GRCh38
rs201403115 Chr.19: 17276419-17276419 on Build GRCh38
rs369680509 Chr.19: 17276775-17276775 on Build GRCh38
rs1056225246 Chr.19: 17276803-17276803 on Build GRCh38
rs367666058 Chr.19: 17276552-17276552 on Build GRCh38
rs749886493 Chr.19: 17276450-17276451 on Build GRCh38
rs923550383 Chr.19: 17276446-17276446 on Build GRCh38
rs755646838 Chr.19: 17276603-17276605 on Build GRCh38
rs914011822 Chr.19: 17276870-17276870 on Build GRCh38
rs200016717 Chr.19: 17276581-17276581 on Build GRCh38
rs765723650 Chr.19: 17276528-17276528 on Build GRCh38
rs771465795 Chr.19: 17276492-17276492 on Build GRCh38
rs772247808 Chr.19: 17276801-17276801 on Build GRCh38
rs553145535 Chr.19: 17276609-17276609 on Build GRCh38
rs751745605 Chr.19: 17276540-17276540 on Build GRCh38
rs758425891 Chr.19: 17276444-17276444 on Build GRCh38
rs763354003 Chr.19: 17276629-17276629 on Build GRCh38
rs772752367 Chr.19: 17276500-17276500 on Build GRCh38
rs762460102 Chr.19: 17276792-17276792 on Build GRCh38
rs773413132 Chr.19: 17276802-17276802 on Build GRCh38
rs865932567 Chr.19: 17276733-17276733 on Build GRCh38
rs765699252 Chr.19: 17276431-17276431 on Build GRCh38
rs375274426 Chr.19: 17276580-17276580 on Build GRCh38
rs770529132 Chr.19: 17276599-17276599 on Build GRCh38
rs373345419 Chr.19: 17276859-17276859 on Build GRCh38
rs746663940 Chr.19: 17276507-17276507 on Build GRCh38
rs746537560 Chr.19: 17276594-17276594 on Build GRCh38
rs1044961359 Chr.19: 17276497-17276497 on Build GRCh38
rs149371949 Chr.19: 17276612-17276612 on Build GRCh38
rs376541002 Chr.19: 17276814-17276814 on Build GRCh38
rs138909874 Chr.19: 17276461-17276461 on Build GRCh38
rs757494361 Chr.19: 17276541-17276541 on Build GRCh38
rs907148356 Chr.19: 17276544-17276544 on Build GRCh38
rs758916057 Chr.19: 17276456-17276456 on Build GRCh38
rs760230245 Chr.19: 17276422-17276422 on Build GRCh38
rs756733950 Chr.19: 17276768-17276768 on Build GRCh38
rs776284368 Chr.19: 17276607-17276607 on Build GRCh38
rs769248559 Chr.19: 17276777-17276777 on Build GRCh38
rs767799658 Chr.19: 17276695-17276695 on Build GRCh38
rs372034767 Chr.19: 17276464-17276464 on Build GRCh38
rs931575960 Chr.19: 17276773-17276773 on Build GRCh38
rs757371052 Chr.19: 17276611-17276611 on Build GRCh38
rs891626677 Chr.19: 17276530-17276530 on Build GRCh38
rs749110768 Chr.19: 17276468-17276468 on Build GRCh38
rs201681717 Chr.19: 17276560-17276560 on Build GRCh38
rs774507907 Chr.19: 17276681-17276681 on Build GRCh38
rs751018712 Chr.19: 17276767-17276767 on Build GRCh38
rs368476221 Chr.19: 17276460-17276460 on Build GRCh38
rs938887367 Chr.19: 17276800-17276800 on Build GRCh38
rs765664881 Chr.19: 17276440-17276440 on Build GRCh38
rs1037240474 Chr.19: 17276455-17276455 on Build GRCh38
rs920138696 Chr.19: 17276752-17276752 on Build GRCh38
rs763922306 Chr.19: 17276852-17276852 on Build GRCh38
rs61634114 Chr.19: 17276445-17276445 on Build GRCh38
rs754271614 Chr.19: 17276435-17276435 on Build GRCh38
rs754753033 Chr.19: 17276559-17276559 on Build GRCh38
rs753794856 Chr.19: 17276853-17276853 on Build GRCh38
rs375135043 Chr.19: 17276674-17276674 on Build GRCh38
rs896136975 Chr.19: 17276529-17276529 on Build GRCh38
rs766641020 Chr.19: 17276828-17276828 on Build GRCh38
rs764264413 Chr.19: 17276537-17276537 on Build GRCh38
rs752370874 Chr.19: 17276838-17276838 on Build GRCh38
rs577160894 Chr.19: 17276703-17276703 on Build GRCh38
rs550805861 Chr.19: 17276536-17276536 on Build GRCh38
rs558669016 Chr.19: 17276480-17276480 on Build GRCh38
rs896271183 Chr.19: 17276872-17276872 on Build GRCh38
rs753629716 Chr.19: 17276546-17276546 on Build GRCh38

View Full Product Details