Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17555604-17556108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLGALT1
Entrez Gene ID
79709
Gene Name
collagen beta(1-O)galactosyltransferase 1
Gene Aliases
GLT25D1
UniGene
Hs.418795
Gene Chromosome Location
Chr.19: 17555594-17583162 on Build GRCh38
Transcripts
NM_024656.3 XM_011527786.2

Gene Symbol
FAM129C
Entrez Gene ID
199786
Gene Name
family with sequence similarity 129 member C
Gene Aliases
BCNP1
UniGene
Hs.434133
Gene Chromosome Location
Chr.19: 17523301-17556755 on Build GRCh38
Transcripts
NM_024656.3 XM_011527786.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5659996 c.118G>A p.E40K Chr.19: 17555831-17555831 on Build GRCh38
4856139 c.155C>T p.P52L Chr.19: 17555868-17555868 on Build GRCh38
5761237 c.233G>A p.R78Q Chr.19: 17555946-17555946 on Build GRCh38
4131420 c.114C>T p.F38F Chr.19: 17555827-17555827 on Build GRCh38
5724325 c.115C>T p.P39S Chr.19: 17555828-17555828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938614876 Chr.19: 17555676-17555676 on Build GRCh38
rs762608439 Chr.19: 17555923-17555923 on Build GRCh38
rs1015946992 Chr.19: 17555742-17555742 on Build GRCh38
rs998480369 Chr.19: 17555881-17555881 on Build GRCh38
rs943415884 Chr.19: 17555644-17555644 on Build GRCh38
rs990071220 Chr.19: 17555964-17555964 on Build GRCh38
rs1026367282 Chr.19: 17556074-17556074 on Build GRCh38
rs888046568 Chr.19: 17555643-17555643 on Build GRCh38
rs915368953 Chr.19: 17555650-17555650 on Build GRCh38
rs964873130 Chr.19: 17555802-17555802 on Build GRCh38
rs895300504 Chr.19: 17555739-17555739 on Build GRCh38
rs1024052482 Chr.19: 17555862-17555862 on Build GRCh38
rs906929621 Chr.19: 17555873-17555873 on Build GRCh38
rs989671669 Chr.19: 17555811-17555811 on Build GRCh38
rs375187950 Chr.19: 17555649-17555649 on Build GRCh38
rs1013121754 Chr.19: 17555838-17555838 on Build GRCh38
rs754091915 Chr.19: 17555848-17555848 on Build GRCh38
rs903075606 Chr.19: 17555666-17555666 on Build GRCh38
rs894404579 Chr.19: 17555944-17555944 on Build GRCh38
rs1037507910 Chr.19: 17555984-17555984 on Build GRCh38
rs1042441669 Chr.19: 17555664-17555664 on Build GRCh38
rs894299582 Chr.19: 17555721-17555721 on Build GRCh38
rs532186337 Chr.19: 17555968-17555968 on Build GRCh38
rs1047719722 Chr.19: 17555637-17555637 on Build GRCh38
rs976285128 Chr.19: 17556040-17556040 on Build GRCh38
rs1018308980 Chr.19: 17555779-17555779 on Build GRCh38
rs978458440 Chr.19: 17555832-17555832 on Build GRCh38
rs955092262 Chr.19: 17555753-17555753 on Build GRCh38
rs746478240 Chr.19: 17555952-17555952 on Build GRCh38
rs1026000317 Chr.19: 17555747-17555747 on Build GRCh38
rs574361846 Chr.19: 17555673-17555673 on Build GRCh38
rs1055727816 Chr.19: 17555710-17555710 on Build GRCh38
rs542132867 Chr.19: 17555695-17555698 on Build GRCh38
rs964568112 Chr.19: 17556036-17556036 on Build GRCh38
rs549622223 Chr.19: 17556060-17556060 on Build GRCh38
rs767308333 Chr.19: 17555905-17555905 on Build GRCh38
rs898898209 Chr.19: 17555993-17555993 on Build GRCh38
rs898945714 Chr.19: 17555744-17555744 on Build GRCh38
rs929299166 Chr.19: 17555632-17555632 on Build GRCh38
rs988023799 Chr.19: 17555637-17555638 on Build GRCh38
rs1017893273 Chr.19: 17556009-17556009 on Build GRCh38
rs563277874 Chr.19: 17555899-17555899 on Build GRCh38
rs987750259 Chr.19: 17555763-17555763 on Build GRCh38
rs576623097 Chr.19: 17555651-17555651 on Build GRCh38
rs923351968 Chr.19: 17556050-17556050 on Build GRCh38
rs993213419 Chr.19: 17555996-17555996 on Build GRCh38
rs1012740258 Chr.19: 17555726-17555726 on Build GRCh38
rs7259723 Chr.19: 17555827-17555827 on Build GRCh38
rs945341411 Chr.19: 17555656-17555656 on Build GRCh38
rs766525393 Chr.19: 17555831-17555831 on Build GRCh38
rs760759942 Chr.19: 17555830-17555830 on Build GRCh38
rs1057044165 Chr.19: 17555908-17555908 on Build GRCh38
rs374320534 Chr.19: 17556054-17556054 on Build GRCh38
rs541815906 Chr.19: 17555627-17555627 on Build GRCh38

View Full Product Details