Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18588593-18589087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf60
Entrez Gene ID
55049
Gene Name
chromosome 19 open reading frame 60
Gene Aliases
-
UniGene
Hs.655317
Gene Chromosome Location
Chr.19: 18588685-18592337 on Build GRCh38
Transcripts
NM_001100418.1 NM_001100419.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5701452 c.92_93insG p.P32fs*20 Chr.19: 18588893-18588894 on Build GRCh38
5774083 c.180G>T p.E60D Chr.19: 18589075-18589075 on Build GRCh38
6443872 c.136C>A p.H46N Chr.19: 18589031-18589031 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs911810962 Chr.19: 18589001-18589001 on Build GRCh38
rs1034534298 Chr.19: 18588677-18588677 on Build GRCh38
rs1010688673 Chr.19: 18588874-18588874 on Build GRCh38
rs930561372 Chr.19: 18588851-18588851 on Build GRCh38
rs986244216 Chr.19: 18588817-18588817 on Build GRCh38
rs900191971 Chr.19: 18589036-18589036 on Build GRCh38
rs775795196 Chr.19: 18588969-18588969 on Build GRCh38
rs1014533996 Chr.19: 18588713-18588713 on Build GRCh38
rs747557742 Chr.19: 18588882-18588882 on Build GRCh38
rs903470856 Chr.19: 18588910-18588910 on Build GRCh38
rs978462230 Chr.19: 18588746-18588746 on Build GRCh38
rs992836646 Chr.19: 18588642-18588642 on Build GRCh38
rs746277198 Chr.19: 18588937-18588937 on Build GRCh38
rs371558866 Chr.19: 18588908-18588908 on Build GRCh38
rs1038706758 Chr.19: 18589014-18589014 on Build GRCh38
rs771099543 Chr.19: 18588731-18588731 on Build GRCh38
rs888412975 Chr.19: 18588870-18588870 on Build GRCh38
rs944512091 Chr.19: 18588829-18588829 on Build GRCh38
rs182766581 Chr.19: 18588627-18588627 on Build GRCh38
rs967610882 Chr.19: 18588735-18588735 on Build GRCh38
rs202026240 Chr.19: 18589005-18589005 on Build GRCh38
rs753507415 Chr.19: 18589060-18589060 on Build GRCh38
rs1038758360 Chr.19: 18588840-18588840 on Build GRCh38
rs953136117 Chr.19: 18588925-18588925 on Build GRCh38
rs925601456 Chr.19: 18588770-18588770 on Build GRCh38
rs1048365385 Chr.19: 18588650-18588650 on Build GRCh38
rs576981326 Chr.19: 18588921-18588921 on Build GRCh38
rs756160711 Chr.19: 18588776-18588776 on Build GRCh38
rs769842132 Chr.19: 18588944-18588944 on Build GRCh38
rs953162815 Chr.19: 18588775-18588775 on Build GRCh38
rs1036212546 Chr.19: 18588634-18588634 on Build GRCh38
rs919197978 Chr.19: 18588843-18588843 on Build GRCh38
rs929164590 Chr.19: 18588977-18588977 on Build GRCh38
rs1055786375 Chr.19: 18589010-18589010 on Build GRCh38
rs1048067583 Chr.19: 18588984-18588984 on Build GRCh38
rs911745076 Chr.19: 18588820-18588820 on Build GRCh38
rs763325527 Chr.19: 18588970-18588970 on Build GRCh38
rs971913582 Chr.19: 18588962-18588962 on Build GRCh38
rs757894866 Chr.19: 18588887-18588887 on Build GRCh38
rs937445540 Chr.19: 18588872-18588872 on Build GRCh38
rs369945153 Chr.19: 18588669-18588669 on Build GRCh38
rs1019136023 Chr.19: 18588949-18588949 on Build GRCh38
rs1047655017 Chr.19: 18588684-18588684 on Build GRCh38
rs537150436 Chr.19: 18588895-18588895 on Build GRCh38
rs763095689 Chr.19: 18588818-18588827 on Build GRCh38
rs552921750 Chr.19: 18588647-18588647 on Build GRCh38
rs977725138 Chr.19: 18588624-18588624 on Build GRCh38
rs10407212 Chr.19: 18588635-18588635 on Build GRCh38
rs559493496 Chr.19: 18588724-18588724 on Build GRCh38
rs1056260534 Chr.19: 18588877-18588877 on Build GRCh38
rs937311862 Chr.19: 18589007-18589007 on Build GRCh38
rs895927711 Chr.19: 18589051-18589051 on Build GRCh38
rs911679217 Chr.19: 18588626-18588626 on Build GRCh38
rs1048426697 Chr.19: 18588864-18588864 on Build GRCh38
rs374325797 Chr.19: 18588788-18588788 on Build GRCh38
rs985075733 Chr.19: 18588993-18588993 on Build GRCh38
rs752533255 Chr.19: 18588819-18588819 on Build GRCh38
rs890084360 Chr.19: 18588709-18588709 on Build GRCh38
rs754548897 Chr.19: 18588831-18588831 on Build GRCh38
rs754970178 Chr.19: 18588692-18588692 on Build GRCh38
rs769863181 Chr.19: 18588685-18588685 on Build GRCh38
rs568105805 Chr.19: 18588827-18588827 on Build GRCh38
rs565653109 Chr.19: 18588648-18588648 on Build GRCh38
rs778635257 Chr.19: 18588832-18588832 on Build GRCh38
rs888580684 Chr.19: 18588658-18588658 on Build GRCh38
rs113923497 Chr.19: 18588765-18588765 on Build GRCh38
rs10415257 Chr.19: 18588679-18588679 on Build GRCh38
rs930571098 Chr.19: 18588988-18588988 on Build GRCh38
rs965747210 Chr.19: 18588952-18588952 on Build GRCh38
rs753691744 Chr.19: 18588830-18588830 on Build GRCh38
rs777327574 Chr.19: 18588889-18588889 on Build GRCh38
rs771479774 Chr.19: 18588913-18588913 on Build GRCh38
rs774620541 Chr.19: 18588990-18588990 on Build GRCh38
rs746665131 Chr.19: 18588669-18588669 on Build GRCh38
rs764237847 Chr.19: 18588972-18588972 on Build GRCh38
rs973533422 Chr.19: 18588958-18588958 on Build GRCh38
rs750381280 Chr.19: 18588774-18588774 on Build GRCh38
rs553630345 Chr.19: 18588644-18588644 on Build GRCh38
rs766369158 Chr.19: 18589011-18589011 on Build GRCh38
rs773485202 Chr.19: 18588701-18588701 on Build GRCh38

View Full Product Details