Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19495918-19496437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATAD2A
Entrez Gene ID
54815
Gene Name
GATA zinc finger domain containing 2A
Gene Aliases
p66alpha
UniGene
Hs.118964
Gene Chromosome Location
Chr.19: 19385833-19508932 on Build GRCh38
Transcripts
NM_001300946.1 NM_017660.3 XM_011528105.1 XM_017026901.1 XM_017026902.1 XM_017026903.1 XM_017026904.1 XM_017026905.1 XM_017026906.1 XM_017026907.1 XM_017026908.1 XM_017026909.1 XM_017026910.1 XM_017026911.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4890006 c.383C>A p.A128D Chr.19: 19496107-19496107 on Build GRCh38
7105066 c.444C>T p.L148L Chr.19: 19496168-19496168 on Build GRCh38
5933814 c.862C>T p.Q288* Chr.19: 19496157-19496157 on Build GRCh38
4280114 c.790G>T p.G264W Chr.19: 19496085-19496085 on Build GRCh38
7156246 c.378C>T p.L126L Chr.19: 19496102-19496102 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112553051 Chr.19: 19496386-19496386 on Build GRCh38
rs73002973 Chr.19: 19496039-19496039 on Build GRCh38
rs889262409 Chr.19: 19495992-19495992 on Build GRCh38
rs906267179 Chr.19: 19496250-19496250 on Build GRCh38
rs1033561074 Chr.19: 19496403-19496403 on Build GRCh38
rs917637524 Chr.19: 19496165-19496165 on Build GRCh38
rs756000158 Chr.19: 19496168-19496168 on Build GRCh38
rs1000449309 Chr.19: 19495974-19495974 on Build GRCh38
rs536010506 Chr.19: 19495939-19495939 on Build GRCh38
rs964101106 Chr.19: 19496301-19496301 on Build GRCh38
rs967411617 Chr.19: 19496356-19496356 on Build GRCh38
rs777887495 Chr.19: 19496281-19496281 on Build GRCh38
rs768213461 Chr.19: 19496035-19496035 on Build GRCh38
rs756055279 Chr.19: 19496243-19496243 on Build GRCh38
rs748452735 Chr.19: 19496245-19496245 on Build GRCh38
rs977561912 Chr.19: 19496379-19496379 on Build GRCh38
rs758748693 Chr.19: 19496251-19496252 on Build GRCh38
rs201289342 Chr.19: 19496093-19496093 on Build GRCh38
rs750726616 Chr.19: 19495967-19495967 on Build GRCh38
rs750387323 Chr.19: 19496237-19496237 on Build GRCh38
rs761132140 Chr.19: 19496183-19496183 on Build GRCh38
rs771588933 Chr.19: 19496178-19496178 on Build GRCh38
rs138530563 Chr.19: 19496285-19496285 on Build GRCh38
rs916281264 Chr.19: 19496230-19496230 on Build GRCh38
rs549188396 Chr.19: 19496177-19496177 on Build GRCh38
rs1025593958 Chr.19: 19495991-19495991 on Build GRCh38
rs58845328 Chr.19: 19495952-19495952 on Build GRCh38
rs947728447 Chr.19: 19496394-19496394 on Build GRCh38
rs1029714966 Chr.19: 19496342-19496342 on Build GRCh38
rs577666541 Chr.19: 19496062-19496062 on Build GRCh38
rs984710023 Chr.19: 19496108-19496108 on Build GRCh38
rs1006649839 Chr.19: 19496401-19496401 on Build GRCh38
rs374965734 Chr.19: 19496172-19496172 on Build GRCh38
rs771291766 Chr.19: 19496184-19496184 on Build GRCh38
rs368175791 Chr.19: 19496113-19496113 on Build GRCh38
rs977530840 Chr.19: 19496126-19496126 on Build GRCh38
rs780124023 Chr.19: 19496257-19496257 on Build GRCh38
rs533125805 Chr.19: 19496294-19496294 on Build GRCh38
rs370944557 Chr.19: 19496135-19496135 on Build GRCh38
rs977875266 Chr.19: 19496206-19496206 on Build GRCh38
rs781180125 Chr.19: 19496001-19496001 on Build GRCh38
rs926108252 Chr.19: 19496382-19496382 on Build GRCh38
rs528154163 Chr.19: 19496187-19496187 on Build GRCh38
rs544317136 Chr.19: 19496016-19496016 on Build GRCh38
rs759612580 Chr.19: 19496116-19496116 on Build GRCh38
rs371903708 Chr.19: 19496231-19496231 on Build GRCh38
rs1055144788 Chr.19: 19495984-19495984 on Build GRCh38
rs367981840 Chr.19: 19496209-19496209 on Build GRCh38
rs768250542 Chr.19: 19496174-19496174 on Build GRCh38
rs1018947285 Chr.19: 19495998-19495998 on Build GRCh38
rs952168626 Chr.19: 19496105-19496105 on Build GRCh38
rs903550369 Chr.19: 19495972-19495972 on Build GRCh38
rs1042082934 Chr.19: 19495965-19495965 on Build GRCh38
rs771011514 Chr.19: 19496019-19496019 on Build GRCh38
rs754770631 Chr.19: 19496264-19496264 on Build GRCh38
rs779506191 Chr.19: 19496238-19496239 on Build GRCh38
rs751666844 Chr.19: 19496389-19496389 on Build GRCh38
rs767277394 Chr.19: 19496119-19496119 on Build GRCh38
rs374829526 Chr.19: 19496261-19496261 on Build GRCh38
rs1040514025 Chr.19: 19496320-19496320 on Build GRCh38
rs78079701 Chr.19: 19496286-19496286 on Build GRCh38
rs575347401 Chr.19: 19495990-19495990 on Build GRCh38
rs761371491 Chr.19: 19496040-19496040 on Build GRCh38
rs560361174 Chr.19: 19496117-19496117 on Build GRCh38
rs773677474 Chr.19: 19496092-19496092 on Build GRCh38
rs769686468 Chr.19: 19496003-19496003 on Build GRCh38
rs759934826 Chr.19: 19496193-19496193 on Build GRCh38
rs879012155 Chr.19: 19496033-19496033 on Build GRCh38
rs752937530 Chr.19: 19496227-19496227 on Build GRCh38
rs546990310 Chr.19: 19496317-19496317 on Build GRCh38
rs779960713 Chr.19: 19496256-19496257 on Build GRCh38
rs758505727 Chr.19: 19496120-19496120 on Build GRCh38
rs754092111 Chr.19: 19496115-19496115 on Build GRCh38
rs879084612 Chr.19: 19496031-19496031 on Build GRCh38
rs964385206 Chr.19: 19496067-19496067 on Build GRCh38
rs2288851 Chr.19: 19496182-19496182 on Build GRCh38
rs944982824 Chr.19: 19495959-19495959 on Build GRCh38
rs778621123 Chr.19: 19496175-19496175 on Build GRCh38
rs998129083 Chr.19: 19496313-19496313 on Build GRCh38
rs751099391 Chr.19: 19496000-19496000 on Build GRCh38
rs45580736 Chr.19: 19496156-19496156 on Build GRCh38
rs772521558 Chr.19: 19496072-19496072 on Build GRCh38
rs993527642 Chr.19: 19496381-19496381 on Build GRCh38
rs902060935 Chr.19: 19496372-19496372 on Build GRCh38
rs887753822 Chr.19: 19496391-19496391 on Build GRCh38
rs1007700986 Chr.19: 19495993-19495993 on Build GRCh38
rs371171214 Chr.19: 19496015-19496015 on Build GRCh38
rs748958687 Chr.19: 19496173-19496173 on Build GRCh38

View Full Product Details