Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19542775-19543269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CILP2
Entrez Gene ID
148113
Gene Name
cartilage intermediate layer protein 2
Gene Aliases
CLIP-2
UniGene
Hs.279574
Gene Chromosome Location
Chr.19: 19538265-19546659 on Build GRCh38
Transcripts
NM_153221.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6237373 c.978-9G>T p.? Chr.19: 19543239-19543239 on Build GRCh38
4894177 c.944C>T p.A315V Chr.19: 19542939-19542939 on Build GRCh38
3101460 c.949G>A p.G317R Chr.19: 19542944-19542944 on Build GRCh38
3531080 c.918C>T p.G306G Chr.19: 19542913-19542913 on Build GRCh38
5495971 c.977+7G>A p.? Chr.19: 19542979-19542979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769379948 Chr.19: 19542961-19542961 on Build GRCh38
rs778484094 Chr.19: 19543202-19543202 on Build GRCh38
rs770636449 Chr.19: 19542865-19542865 on Build GRCh38
rs573835129 Chr.19: 19543069-19543069 on Build GRCh38
rs751517386 Chr.19: 19542993-19542993 on Build GRCh38
rs539934829 Chr.19: 19542897-19542897 on Build GRCh38
rs758208409 Chr.19: 19542835-19542835 on Build GRCh38
rs780959337 Chr.19: 19542954-19542954 on Build GRCh38
rs753789478 Chr.19: 19542931-19542931 on Build GRCh38
rs767975609 Chr.19: 19542913-19542913 on Build GRCh38
rs759895052 Chr.19: 19542947-19542948 on Build GRCh38
rs779525026 Chr.19: 19543022-19543022 on Build GRCh38
rs367838537 Chr.19: 19542979-19542979 on Build GRCh38
rs58086295 Chr.19: 19543230-19543230 on Build GRCh38
rs139958428 Chr.19: 19543073-19543073 on Build GRCh38
rs751913381 Chr.19: 19543086-19543086 on Build GRCh38
rs374549449 Chr.19: 19543211-19543211 on Build GRCh38
rs986262226 Chr.19: 19543023-19543023 on Build GRCh38
rs777624556 Chr.19: 19542851-19542851 on Build GRCh38
rs779545782 Chr.19: 19542967-19542967 on Build GRCh38
rs1048553888 Chr.19: 19542907-19542907 on Build GRCh38
rs748735232 Chr.19: 19542987-19542987 on Build GRCh38
rs971753767 Chr.19: 19543068-19543068 on Build GRCh38
rs990338443 Chr.19: 19542803-19542803 on Build GRCh38
rs374606476 Chr.19: 19543097-19543097 on Build GRCh38
rs150832743 Chr.19: 19543045-19543045 on Build GRCh38
rs780871193 Chr.19: 19542870-19542870 on Build GRCh38
rs376812208 Chr.19: 19543232-19543232 on Build GRCh38
rs748665426 Chr.19: 19543208-19543208 on Build GRCh38
rs767312660 Chr.19: 19543000-19543000 on Build GRCh38
rs760978594 Chr.19: 19542915-19542915 on Build GRCh38
rs752752595 Chr.19: 19542826-19542826 on Build GRCh38
rs755888003 Chr.19: 19542924-19542924 on Build GRCh38
rs754489375 Chr.19: 19543200-19543200 on Build GRCh38
rs939810981 Chr.19: 19543166-19543166 on Build GRCh38
rs532143241 Chr.19: 19543079-19543079 on Build GRCh38
rs866713077 Chr.19: 19542896-19542896 on Build GRCh38
rs370889937 Chr.19: 19542988-19542988 on Build GRCh38
rs1050126017 Chr.19: 19543217-19543217 on Build GRCh38
rs895890853 Chr.19: 19543158-19543158 on Build GRCh38
rs1008204427 Chr.19: 19543034-19543034 on Build GRCh38
rs554443033 Chr.19: 19543237-19543237 on Build GRCh38
rs193028137 Chr.19: 19542806-19542806 on Build GRCh38
rs771397389 Chr.19: 19543228-19543228 on Build GRCh38
rs1056393212 Chr.19: 19543146-19543146 on Build GRCh38
rs926291904 Chr.19: 19542898-19542898 on Build GRCh38
rs774966489 Chr.19: 19542886-19542886 on Build GRCh38
rs773846239 Chr.19: 19543106-19543106 on Build GRCh38
rs376814408 Chr.19: 19542939-19542939 on Build GRCh38
rs748708643 Chr.19: 19542970-19542970 on Build GRCh38
rs747325214 Chr.19: 19543225-19543225 on Build GRCh38
rs201408948 Chr.19: 19542983-19542983 on Build GRCh38
rs760978689 Chr.19: 19542984-19542984 on Build GRCh38
rs766627413 Chr.19: 19542927-19542927 on Build GRCh38
rs1021805883 Chr.19: 19543072-19543072 on Build GRCh38
rs908208931 Chr.19: 19543138-19543138 on Build GRCh38
rs143695168 Chr.19: 19542943-19542943 on Build GRCh38
rs758387632 Chr.19: 19542937-19542937 on Build GRCh38
rs937471580 Chr.19: 19543145-19543145 on Build GRCh38
rs1048689805 Chr.19: 19543081-19543081 on Build GRCh38
rs938915966 Chr.19: 19543082-19543082 on Build GRCh38
rs866216323 Chr.19: 19542842-19542842 on Build GRCh38
rs552547008 Chr.19: 19543150-19543150 on Build GRCh38
rs201872220 Chr.19: 19542871-19542871 on Build GRCh38
rs534822338 Chr.19: 19543231-19543231 on Build GRCh38
rs774811655 Chr.19: 19542903-19542903 on Build GRCh38
rs745578974 Chr.19: 19542960-19542960 on Build GRCh38
rs750716481 Chr.19: 19542914-19542914 on Build GRCh38
rs140099724 Chr.19: 19542908-19542908 on Build GRCh38
rs769293535 Chr.19: 19542982-19542982 on Build GRCh38
rs956136122 Chr.19: 19543051-19543051 on Build GRCh38
rs769359842 Chr.19: 19542885-19542885 on Build GRCh38
rs966216792 Chr.19: 19543053-19543053 on Build GRCh38
rs146789008 Chr.19: 19542944-19542944 on Build GRCh38
rs991868937 Chr.19: 19543111-19543111 on Build GRCh38
rs750151744 Chr.19: 19543007-19543007 on Build GRCh38
rs113278598 Chr.19: 19543016-19543016 on Build GRCh38
rs772653120 Chr.19: 19543209-19543209 on Build GRCh38
rs755795649 Chr.19: 19543009-19543009 on Build GRCh38
rs910026540 Chr.19: 19543103-19543103 on Build GRCh38
rs1038043412 Chr.19: 19543173-19543173 on Build GRCh38
rs763530960 Chr.19: 19542902-19542902 on Build GRCh38
rs767404262 Chr.19: 19543247-19543247 on Build GRCh38
rs765326910 Chr.19: 19542989-19542989 on Build GRCh38
rs770542351 Chr.19: 19543078-19543078 on Build GRCh38
rs769231133 Chr.19: 19542973-19542973 on Build GRCh38
rs748950597 Chr.19: 19543197-19543197 on Build GRCh38
rs1014796041 Chr.19: 19543196-19543196 on Build GRCh38
rs746738302 Chr.19: 19542852-19542852 on Build GRCh38

View Full Product Details