Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19543156-19543651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CILP2
Entrez Gene ID
148113
Gene Name
cartilage intermediate layer protein 2
Gene Aliases
CLIP-2
UniGene
Hs.279574
Gene Chromosome Location
Chr.19: 19538265-19546659 on Build GRCh38
Transcripts
NM_153221.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5888397 c.1129G>T p.V377L Chr.19: 19543399-19543399 on Build GRCh38
4909711 c.1008A>G p.R336R Chr.19: 19543278-19543278 on Build GRCh38
993356 c.1006C>T p.R336* Chr.19: 19543276-19543276 on Build GRCh38
7108426 c.1054G>C p.D352H Chr.19: 19543324-19543324 on Build GRCh38
1129943 c.1110G>A p.S370S Chr.19: 19543380-19543380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042209173 Chr.19: 19543425-19543425 on Build GRCh38
rs762973440 Chr.19: 19543309-19543309 on Build GRCh38
rs778484094 Chr.19: 19543202-19543202 on Build GRCh38
rs143359521 Chr.19: 19543310-19543310 on Build GRCh38
rs372115427 Chr.19: 19543389-19543389 on Build GRCh38
rs139472405 Chr.19: 19543270-19543270 on Build GRCh38
rs754649866 Chr.19: 19543276-19543276 on Build GRCh38
rs751045943 Chr.19: 19543411-19543411 on Build GRCh38
rs778470110 Chr.19: 19543287-19543287 on Build GRCh38
rs781688994 Chr.19: 19543414-19543414 on Build GRCh38
rs58086295 Chr.19: 19543230-19543230 on Build GRCh38
rs150065892 Chr.19: 19543279-19543279 on Build GRCh38
rs7252453 Chr.19: 19543308-19543308 on Build GRCh38
rs747484764 Chr.19: 19543290-19543290 on Build GRCh38
rs992161977 Chr.19: 19543629-19543629 on Build GRCh38
rs757736188 Chr.19: 19543412-19543412 on Build GRCh38
rs137913643 Chr.19: 19543390-19543390 on Build GRCh38
rs532157215 Chr.19: 19543511-19543511 on Build GRCh38
rs374549449 Chr.19: 19543211-19543211 on Build GRCh38
rs552158329 Chr.19: 19543540-19543540 on Build GRCh38
rs201485988 Chr.19: 19543319-19543319 on Build GRCh38
rs763709825 Chr.19: 19543408-19543408 on Build GRCh38
rs752168071 Chr.19: 19543365-19543365 on Build GRCh38
rs759119693 Chr.19: 19543325-19543325 on Build GRCh38
rs376812208 Chr.19: 19543232-19543232 on Build GRCh38
rs748665426 Chr.19: 19543208-19543208 on Build GRCh38
rs754489375 Chr.19: 19543200-19543200 on Build GRCh38
rs1050126017 Chr.19: 19543217-19543217 on Build GRCh38
rs372602791 Chr.19: 19543430-19543430 on Build GRCh38
rs200587421 Chr.19: 19543354-19543354 on Build GRCh38
rs554443033 Chr.19: 19543237-19543237 on Build GRCh38
rs772017942 Chr.19: 19543437-19543437 on Build GRCh38
rs771397389 Chr.19: 19543228-19543228 on Build GRCh38
rs769932373 Chr.19: 19543375-19543375 on Build GRCh38
rs1018746651 Chr.19: 19543457-19543457 on Build GRCh38
rs147927361 Chr.19: 19543318-19543318 on Build GRCh38
rs369786318 Chr.19: 19543256-19543256 on Build GRCh38
rs758993213 Chr.19: 19543282-19543282 on Build GRCh38
rs141277324 Chr.19: 19543376-19543376 on Build GRCh38
rs1019229479 Chr.19: 19543305-19543305 on Build GRCh38
rs188436574 Chr.19: 19543277-19543277 on Build GRCh38
rs755953687 Chr.19: 19543257-19543257 on Build GRCh38
rs746090290 Chr.19: 19543295-19543295 on Build GRCh38
rs747325214 Chr.19: 19543225-19543225 on Build GRCh38
rs951803506 Chr.19: 19543552-19543552 on Build GRCh38
rs750873273 Chr.19: 19543419-19543419 on Build GRCh38
rs889024517 Chr.19: 19543444-19543444 on Build GRCh38
rs777415063 Chr.19: 19543364-19543364 on Build GRCh38
rs143503295 Chr.19: 19543428-19543428 on Build GRCh38
rs753876811 Chr.19: 19543345-19543345 on Build GRCh38
rs541629092 Chr.19: 19543382-19543382 on Build GRCh38
rs958949170 Chr.19: 19543628-19543628 on Build GRCh38
rs752462258 Chr.19: 19543399-19543399 on Build GRCh38
rs1011560257 Chr.19: 19543486-19543486 on Build GRCh38
rs891815257 Chr.19: 19543485-19543485 on Build GRCh38
rs534822338 Chr.19: 19543231-19543231 on Build GRCh38
rs376459373 Chr.19: 19543401-19543401 on Build GRCh38
rs141923821 Chr.19: 19543391-19543391 on Build GRCh38
rs749483136 Chr.19: 19543377-19543377 on Build GRCh38
rs757724397 Chr.19: 19543368-19543368 on Build GRCh38
rs745782206 Chr.19: 19543456-19543456 on Build GRCh38
rs770838972 Chr.19: 19543385-19543385 on Build GRCh38
rs758728651 Chr.19: 19543623-19543623 on Build GRCh38
rs752372369 Chr.19: 19543334-19543334 on Build GRCh38
rs568227779 Chr.19: 19543258-19543258 on Build GRCh38
rs781606092 Chr.19: 19543291-19543291 on Build GRCh38
rs960354302 Chr.19: 19543370-19543370 on Build GRCh38
rs753505078 Chr.19: 19543274-19543274 on Build GRCh38
rs1007928707 Chr.19: 19543451-19543451 on Build GRCh38
rs1035900482 Chr.19: 19543611-19543611 on Build GRCh38
rs772653120 Chr.19: 19543209-19543209 on Build GRCh38
rs749573038 Chr.19: 19543429-19543429 on Build GRCh38
rs376830577 Chr.19: 19543449-19543449 on Build GRCh38
rs768793354 Chr.19: 19543379-19543379 on Build GRCh38
rs201708106 Chr.19: 19543427-19543427 on Build GRCh38
rs368843344 Chr.19: 19543372-19543372 on Build GRCh38
rs767404262 Chr.19: 19543247-19543247 on Build GRCh38
rs764005696 Chr.19: 19543330-19543330 on Build GRCh38
rs145392584 Chr.19: 19543299-19543299 on Build GRCh38
rs563364364 Chr.19: 19543494-19543494 on Build GRCh38
rs993237248 Chr.19: 19543531-19543531 on Build GRCh38
rs748950597 Chr.19: 19543197-19543197 on Build GRCh38
rs756635511 Chr.19: 19543313-19543313 on Build GRCh38
rs1014796041 Chr.19: 19543196-19543196 on Build GRCh38
rs1026048680 Chr.19: 19543545-19543545 on Build GRCh38
rs750559369 Chr.19: 19543613-19543613 on Build GRCh38
rs45556231 Chr.19: 19543380-19543380 on Build GRCh38
rs984446117 Chr.19: 19543589-19543589 on Build GRCh38

View Full Product Details