Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39531469-39531965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EID2B
Entrez Gene ID
126272
Gene Name
EP300 interacting inhibitor of differentiation 2B
Gene Aliases
EID-2B, EID-3
UniGene
Hs.135181
Gene Chromosome Location
Chr.19: 39531533-39532867 on Build GRCh38
Transcripts
NM_152361.2

SNP Detail

SNP ID Chromosome Location
rs1015440244 Chr.19: 39531705-39531705 on Build GRCh38
rs986405630 Chr.19: 39531497-39531497 on Build GRCh38
rs866989943 Chr.19: 39531722-39531722 on Build GRCh38
rs574017925 Chr.19: 39531890-39531890 on Build GRCh38
rs190057538 Chr.19: 39531613-39531613 on Build GRCh38
rs1040516876 Chr.19: 39531931-39531931 on Build GRCh38
rs569360698 Chr.19: 39531609-39531609 on Build GRCh38
rs905158895 Chr.19: 39531671-39531671 on Build GRCh38
rs990508797 Chr.19: 39531806-39531806 on Build GRCh38
rs947648136 Chr.19: 39531867-39531867 on Build GRCh38
rs902642261 Chr.19: 39531512-39531512 on Build GRCh38
rs1043281863 Chr.19: 39531560-39531560 on Build GRCh38
rs186511548 Chr.19: 39531734-39531734 on Build GRCh38
rs924792429 Chr.19: 39531904-39531904 on Build GRCh38
rs574999549 Chr.19: 39531816-39531816 on Build GRCh38
rs773395280 Chr.19: 39531566-39531566 on Build GRCh38
rs542251970 Chr.19: 39531836-39531836 on Build GRCh38
rs937240914 Chr.19: 39531779-39531779 on Build GRCh38
rs563675211 Chr.19: 39531836-39531837 on Build GRCh38
rs764099695 Chr.19: 39531802-39531802 on Build GRCh38
rs896166755 Chr.19: 39531644-39531644 on Build GRCh38
rs1030712916 Chr.19: 39531795-39531795 on Build GRCh38
rs954735671 Chr.19: 39531844-39531844 on Build GRCh38
rs1019126782 Chr.19: 39531557-39531557 on Build GRCh38
rs986575212 Chr.19: 39531845-39531845 on Build GRCh38
rs983514333 Chr.19: 39531692-39531692 on Build GRCh38
rs971188295 Chr.19: 39531706-39531706 on Build GRCh38
rs996423559 Chr.19: 39531584-39531584 on Build GRCh38
rs758535068 Chr.19: 39531737-39531737 on Build GRCh38
rs935039518 Chr.19: 39531915-39531915 on Build GRCh38
rs981256664 Chr.19: 39531716-39531716 on Build GRCh38
rs181914491 Chr.19: 39531719-39531719 on Build GRCh38
rs540831268 Chr.19: 39531831-39531831 on Build GRCh38
rs1041444894 Chr.19: 39531874-39531874 on Build GRCh38
rs1009039009 Chr.19: 39531524-39531524 on Build GRCh38
rs1023556295 Chr.19: 39531667-39531667 on Build GRCh38
rs562178495 Chr.19: 39531935-39531935 on Build GRCh38
rs947532180 Chr.19: 39531517-39531517 on Build GRCh38
rs11458164 Chr.19: 39531835-39531836 on Build GRCh38
rs573480894 Chr.19: 39531849-39531849 on Build GRCh38
rs573469294 Chr.19: 39531651-39531651 on Build GRCh38
rs1041867220 Chr.19: 39531511-39531511 on Build GRCh38
rs1016073217 Chr.19: 39531686-39531686 on Build GRCh38
rs377158903 Chr.19: 39531845-39531845 on Build GRCh38
rs557411944 Chr.19: 39531493-39531493 on Build GRCh38
rs1005995042 Chr.19: 39531681-39531681 on Build GRCh38
rs952148779 Chr.19: 39531691-39531691 on Build GRCh38
rs1052510749 Chr.19: 39531611-39531611 on Build GRCh38
rs1013887555 Chr.19: 39531666-39531666 on Build GRCh38
rs369024638 Chr.19: 39531917-39531917 on Build GRCh38
rs542812976 Chr.19: 39531834-39531834 on Build GRCh38

View Full Product Details