Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39532633-39533096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EID2B
Entrez Gene ID
126272
Gene Name
EP300 interacting inhibitor of differentiation 2B
Gene Aliases
EID-2B, EID-3
UniGene
Hs.135181
Gene Chromosome Location
Chr.19: 39531533-39532867 on Build GRCh38
Transcripts
NM_152361.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6843048 c.120T>A p.V40V Chr.19: 39532683-39532683 on Build GRCh38
6843047 c.121C>G p.R41G Chr.19: 39532682-39532682 on Build GRCh38
4488258 c.32C>T p.S11F Chr.19: 39532771-39532771 on Build GRCh38
1681021 c.63G>T p.Q21H Chr.19: 39532740-39532740 on Build GRCh38
7238607 c.42C>A p.P14P Chr.19: 39532761-39532761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753715110 Chr.19: 39532728-39532728 on Build GRCh38
rs760352755 Chr.19: 39532739-39532739 on Build GRCh38
rs752442157 Chr.19: 39532730-39532730 on Build GRCh38
rs373907829 Chr.19: 39532801-39532801 on Build GRCh38
rs928813122 Chr.19: 39532986-39532986 on Build GRCh38
rs1123301 Chr.19: 39532668-39532668 on Build GRCh38
rs371551550 Chr.19: 39532839-39532839 on Build GRCh38
rs906148985 Chr.19: 39533002-39533002 on Build GRCh38
rs911689453 Chr.19: 39532902-39532902 on Build GRCh38
rs542792554 Chr.19: 39532988-39532988 on Build GRCh38
rs756595643 Chr.19: 39532836-39532836 on Build GRCh38
rs902992525 Chr.19: 39532889-39532889 on Build GRCh38
rs373058868 Chr.19: 39532704-39532704 on Build GRCh38
rs770290842 Chr.19: 39532712-39532712 on Build GRCh38
rs370642720 Chr.19: 39532812-39532812 on Build GRCh38
rs189876842 Chr.19: 39532895-39532895 on Build GRCh38
rs776198968 Chr.19: 39532843-39532843 on Build GRCh38
rs538140091 Chr.19: 39532885-39532885 on Build GRCh38
rs1017081013 Chr.19: 39533007-39533007 on Build GRCh38
rs538225711 Chr.19: 39532934-39532934 on Build GRCh38
rs758432913 Chr.19: 39532827-39532827 on Build GRCh38
rs775385014 Chr.19: 39532782-39532782 on Build GRCh38
rs761574399 Chr.19: 39532821-39532821 on Build GRCh38
rs10412570 Chr.19: 39532816-39532816 on Build GRCh38
rs374094786 Chr.19: 39532823-39532823 on Build GRCh38
rs199839095 Chr.19: 39532828-39532828 on Build GRCh38
rs934766861 Chr.19: 39533043-39533043 on Build GRCh38
rs868278339 Chr.19: 39532663-39532663 on Build GRCh38
rs773086092 Chr.19: 39532854-39532854 on Build GRCh38
rs1019508473 Chr.19: 39532774-39532774 on Build GRCh38
rs748534901 Chr.19: 39532796-39532796 on Build GRCh38
rs757225521 Chr.19: 39532760-39532760 on Build GRCh38
rs970631718 Chr.19: 39532844-39532844 on Build GRCh38
rs777547254 Chr.19: 39533027-39533027 on Build GRCh38
rs759637281 Chr.19: 39532875-39532876 on Build GRCh38
rs778983825 Chr.19: 39532669-39532669 on Build GRCh38
rs773790994 Chr.19: 39533004-39533004 on Build GRCh38
rs915857813 Chr.19: 39532999-39532999 on Build GRCh38
rs761641619 Chr.19: 39532850-39532850 on Build GRCh38
rs781632167 Chr.19: 39532792-39532792 on Build GRCh38
rs561056890 Chr.19: 39533015-39533015 on Build GRCh38
rs369438240 Chr.19: 39532692-39532692 on Build GRCh38
rs776964290 Chr.19: 39532701-39532701 on Build GRCh38
rs758526911 Chr.19: 39532776-39532776 on Build GRCh38
rs894516870 Chr.19: 39532874-39532874 on Build GRCh38
rs967244812 Chr.19: 39532675-39532675 on Build GRCh38
rs377059957 Chr.19: 39532721-39532721 on Build GRCh38
rs972212805 Chr.19: 39532985-39532985 on Build GRCh38
rs565194229 Chr.19: 39532740-39532740 on Build GRCh38
rs56024107 Chr.19: 39533024-39533024 on Build GRCh38
rs947428805 Chr.19: 39533020-39533020 on Build GRCh38
rs771743151 Chr.19: 39532805-39532805 on Build GRCh38
rs1040852665 Chr.19: 39533045-39533045 on Build GRCh38
rs938943981 Chr.19: 39532842-39532842 on Build GRCh38
rs778388711 Chr.19: 39532837-39532837 on Build GRCh38
rs963125673 Chr.19: 39532881-39532881 on Build GRCh38
rs766470003 Chr.19: 39532831-39532831 on Build GRCh38
rs553743549 Chr.19: 39532942-39532942 on Build GRCh38
rs1053078136 Chr.19: 39532967-39532967 on Build GRCh38
rs759181726 Chr.19: 39532729-39532729 on Build GRCh38
rs746570345 Chr.19: 39532841-39532841 on Build GRCh38
rs749841211 Chr.19: 39532838-39532838 on Build GRCh38
rs936095277 Chr.19: 39532951-39532951 on Build GRCh38
rs745348352 Chr.19: 39532802-39532802 on Build GRCh38
rs778784877 Chr.19: 39532893-39532893 on Build GRCh38
rs549142648 Chr.19: 39532835-39532835 on Build GRCh38
rs755235421 Chr.19: 39532833-39532833 on Build GRCh38
rs952421736 Chr.19: 39532938-39532938 on Build GRCh38
rs919167631 Chr.19: 39532713-39532713 on Build GRCh38
rs746025797 Chr.19: 39532689-39532689 on Build GRCh38
rs1043148298 Chr.19: 39532888-39532888 on Build GRCh38
rs754858684 Chr.19: 39532859-39532859 on Build GRCh38
rs962494684 Chr.19: 39532978-39532978 on Build GRCh38
rs572151317 Chr.19: 39532971-39532971 on Build GRCh38
rs200891311 Chr.19: 39532810-39532810 on Build GRCh38
rs373146106 Chr.19: 39532791-39532791 on Build GRCh38
rs1025946214 Chr.19: 39533064-39533064 on Build GRCh38
rs551774889 Chr.19: 39532817-39532817 on Build GRCh38
rs529720415 Chr.19: 39532892-39532892 on Build GRCh38
rs1015364530 Chr.19: 39532949-39532949 on Build GRCh38
rs533129244 Chr.19: 39532786-39532786 on Build GRCh38
rs763735279 Chr.19: 39532811-39532811 on Build GRCh38
rs775239667 Chr.19: 39532806-39532806 on Build GRCh38
rs950436794 Chr.19: 39533070-39533070 on Build GRCh38
rs747356364 Chr.19: 39532699-39532699 on Build GRCh38
rs751675548 Chr.19: 39532832-39532832 on Build GRCh38
rs755549569 Chr.19: 39532832-39532833 on Build GRCh38
rs991747377 Chr.19: 39532996-39532996 on Build GRCh38
rs183859311 Chr.19: 39532898-39532898 on Build GRCh38
rs113028706 Chr.19: 39532882-39532882 on Build GRCh38
rs770601552 Chr.19: 39532808-39532809 on Build GRCh38
rs769104709 Chr.19: 39532700-39532700 on Build GRCh38
rs940874963 Chr.19: 39532834-39532834 on Build GRCh38
rs996756879 Chr.19: 39532925-39532925 on Build GRCh38
rs775077074 Chr.19: 39532840-39532840 on Build GRCh38
rs963076265 Chr.19: 39533014-39533014 on Build GRCh38
rs567566721 Chr.19: 39532884-39532884 on Build GRCh38
rs765047854 Chr.19: 39532851-39532851 on Build GRCh38
rs367937546 Chr.19: 39532824-39532824 on Build GRCh38
rs975033580 Chr.19: 39532808-39532808 on Build GRCh38
rs780219670 Chr.19: 39532781-39532781 on Build GRCh38
rs753850231 Chr.19: 39532660-39532660 on Build GRCh38
rs778891784 Chr.19: 39532761-39532761 on Build GRCh38
rs371536672 Chr.19: 39532759-39532759 on Build GRCh38
rs750492513 Chr.19: 39532771-39532771 on Build GRCh38
rs780130396 Chr.19: 39532699-39532700 on Build GRCh38
rs755250869 Chr.19: 39532788-39532788 on Build GRCh38
rs780315608 Chr.19: 39532697-39532697 on Build GRCh38

View Full Product Details