Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43593071-43593567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRGQ
Entrez Gene ID
126298
Gene Name
immunity related GTPase Q
Gene Aliases
FKSG27, IRGQ1
UniGene
Hs.6217
Gene Chromosome Location
Chr.19: 43584367-43596234 on Build GRCh38
Transcripts
NM_001007561.2 XM_005258515.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5856046 c.636G>A p.W212* Chr.19: 43593262-43593262 on Build GRCh38
997648 c.606G>A p.E202E Chr.19: 43593292-43593292 on Build GRCh38
5794370 c.580C>T p.L194L Chr.19: 43593318-43593318 on Build GRCh38
1612357 c.745G>A p.D249N Chr.19: 43593153-43593153 on Build GRCh38
4078971 c.684C>T p.A228A Chr.19: 43593214-43593214 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763358670 Chr.19: 43593095-43593095 on Build GRCh38
rs752769629 Chr.19: 43593133-43593133 on Build GRCh38
rs939042981 Chr.19: 43593321-43593321 on Build GRCh38
rs747569879 Chr.19: 43593188-43593188 on Build GRCh38
rs952636686 Chr.19: 43593246-43593246 on Build GRCh38
rs377156282 Chr.19: 43593271-43593271 on Build GRCh38
rs144445040 Chr.19: 43593206-43593206 on Build GRCh38
rs374531772 Chr.19: 43593195-43593195 on Build GRCh38
rs759781908 Chr.19: 43593340-43593340 on Build GRCh38
rs1026818004 Chr.19: 43593223-43593223 on Build GRCh38
rs780719661 Chr.19: 43593184-43593184 on Build GRCh38
rs1041996597 Chr.19: 43593509-43593509 on Build GRCh38
rs748935968 Chr.19: 43593311-43593311 on Build GRCh38
rs772512253 Chr.19: 43593343-43593343 on Build GRCh38
rs550471546 Chr.19: 43593375-43593375 on Build GRCh38
rs754669610 Chr.19: 43593405-43593405 on Build GRCh38
rs753964544 Chr.19: 43593373-43593373 on Build GRCh38
rs770757614 Chr.19: 43593333-43593333 on Build GRCh38
rs914778474 Chr.19: 43593469-43593469 on Build GRCh38
rs369790576 Chr.19: 43593140-43593140 on Build GRCh38
rs767952058 Chr.19: 43593219-43593219 on Build GRCh38
rs780940124 Chr.19: 43593265-43593265 on Build GRCh38
rs944065345 Chr.19: 43593544-43593544 on Build GRCh38
rs867438697 Chr.19: 43593306-43593306 on Build GRCh38
rs777492227 Chr.19: 43593289-43593289 on Build GRCh38
rs145005666 Chr.19: 43593334-43593334 on Build GRCh38
rs750663738 Chr.19: 43593389-43593389 on Build GRCh38
rs547007493 Chr.19: 43593536-43593536 on Build GRCh38
rs755608843 Chr.19: 43593130-43593130 on Build GRCh38
rs752045444 Chr.19: 43593127-43593127 on Build GRCh38
rs113476402 Chr.19: 43593189-43593189 on Build GRCh38
rs779887523 Chr.19: 43593176-43593176 on Build GRCh38
rs755114237 Chr.19: 43593496-43593497 on Build GRCh38
rs770328804 Chr.19: 43593454-43593454 on Build GRCh38
rs764250666 Chr.19: 43593242-43593242 on Build GRCh38
rs201590682 Chr.19: 43593186-43593186 on Build GRCh38
rs748555242 Chr.19: 43593340-43593341 on Build GRCh38
rs765401306 Chr.19: 43593380-43593380 on Build GRCh38
rs767348291 Chr.19: 43593411-43593411 on Build GRCh38
rs991665452 Chr.19: 43593451-43593451 on Build GRCh38
rs777820330 Chr.19: 43593144-43593144 on Build GRCh38
rs771417943 Chr.19: 43593196-43593196 on Build GRCh38
rs769117567 Chr.19: 43593286-43593286 on Build GRCh38
rs201885151 Chr.19: 43593150-43593150 on Build GRCh38
rs149921319 Chr.19: 43593191-43593191 on Build GRCh38
rs561591619 Chr.19: 43593214-43593214 on Build GRCh38
rs753997747 Chr.19: 43593244-43593244 on Build GRCh38
rs766863233 Chr.19: 43593113-43593113 on Build GRCh38
rs764062784 Chr.19: 43593367-43593367 on Build GRCh38
rs772500323 Chr.19: 43593342-43593343 on Build GRCh38
rs759941723 Chr.19: 43593355-43593355 on Build GRCh38
rs370638315 Chr.19: 43593353-43593353 on Build GRCh38
rs138768669 Chr.19: 43593100-43593100 on Build GRCh38
rs756330662 Chr.19: 43593139-43593139 on Build GRCh38
rs1004117278 Chr.19: 43593247-43593247 on Build GRCh38
rs758135931 Chr.19: 43593165-43593165 on Build GRCh38
rs149099445 Chr.19: 43593535-43593535 on Build GRCh38
rs760624823 Chr.19: 43593241-43593241 on Build GRCh38
rs764835170 Chr.19: 43593093-43593093 on Build GRCh38
rs377356782 Chr.19: 43593256-43593256 on Build GRCh38
rs757529047 Chr.19: 43593254-43593254 on Build GRCh38
rs564627935 Chr.19: 43593377-43593377 on Build GRCh38
rs886234452 Chr.19: 43593543-43593543 on Build GRCh38
rs755572596 Chr.19: 43593399-43593399 on Build GRCh38
rs994538870 Chr.19: 43593486-43593486 on Build GRCh38
rs958823269 Chr.19: 43593445-43593445 on Build GRCh38
rs761500651 Chr.19: 43593092-43593092 on Build GRCh38
rs924593951 Chr.19: 43593512-43593512 on Build GRCh38
rs777151195 Chr.19: 43593331-43593331 on Build GRCh38
rs374969004 Chr.19: 43593261-43593261 on Build GRCh38
rs775805946 Chr.19: 43593352-43593352 on Build GRCh38
rs746931911 Chr.19: 43593183-43593183 on Build GRCh38
rs947615953 Chr.19: 43593492-43593492 on Build GRCh38

View Full Product Details