Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45777578-45778086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMPK
Entrez Gene ID
1760
Gene Name
dystrophia myotonica protein kinase
Gene Aliases
DM, DM1, DM1PK, DMK, MDPK, MT-PK
UniGene
Hs.631596
Gene Chromosome Location
Chr.19: 45769709-45782557 on Build GRCh38
Transcripts
NM_001081560.2 NM_001081562.2 NM_001081563.2 NM_001288764.1 NM_001288765.1 NM_001288766.1 NM_004409.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4079455 c.742G>A p.G248S Chr.19: 45777807-45777807 on Build GRCh38
5343356 c.713C>T p.S238L Chr.19: 45777866-45777866 on Build GRCh38
7139433 c.801C>A p.F267L Chr.19: 45777748-45777748 on Build GRCh38
1394669 c.832G>A p.A278T Chr.19: 45777747-45777747 on Build GRCh38
7095686 c.834C>G p.F278L Chr.19: 45777715-45777715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77315725 Chr.19: 45777962-45777962 on Build GRCh38
rs962168090 Chr.19: 45777674-45777674 on Build GRCh38
rs767282165 Chr.19: 45777865-45777865 on Build GRCh38
rs780885997 Chr.19: 45777683-45777683 on Build GRCh38
rs374895405 Chr.19: 45777654-45777654 on Build GRCh38
rs750253660 Chr.19: 45777655-45777655 on Build GRCh38
rs368945420 Chr.19: 45777711-45777711 on Build GRCh38
rs751225267 Chr.19: 45777662-45777662 on Build GRCh38
rs779683741 Chr.19: 45778005-45778005 on Build GRCh38
rs757728098 Chr.19: 45777895-45777895 on Build GRCh38
rs193021388 Chr.19: 45777981-45777981 on Build GRCh38
rs749666645 Chr.19: 45777699-45777699 on Build GRCh38
rs1020396097 Chr.19: 45777992-45777992 on Build GRCh38
rs149122951 Chr.19: 45777870-45777870 on Build GRCh38
rs772315750 Chr.19: 45777803-45777803 on Build GRCh38
rs1029162589 Chr.19: 45777833-45777833 on Build GRCh38
rs145261129 Chr.19: 45777712-45777712 on Build GRCh38
rs776266757 Chr.19: 45777611-45777611 on Build GRCh38
rs368913582 Chr.19: 45777794-45777794 on Build GRCh38
rs755865044 Chr.19: 45777999-45777999 on Build GRCh38
rs199698181 Chr.19: 45777778-45777778 on Build GRCh38
rs780860038 Chr.19: 45777877-45777877 on Build GRCh38
rs979400674 Chr.19: 45778030-45778030 on Build GRCh38
rs770389641 Chr.19: 45777617-45777617 on Build GRCh38
rs746534953 Chr.19: 45777605-45777605 on Build GRCh38
rs566079768 Chr.19: 45777946-45777946 on Build GRCh38
rs537756502 Chr.19: 45777986-45777986 on Build GRCh38
rs763177539 Chr.19: 45777625-45777625 on Build GRCh38
rs758707798 Chr.19: 45777908-45777908 on Build GRCh38
rs771438087 Chr.19: 45777790-45777790 on Build GRCh38
rs745732334 Chr.19: 45777901-45777914 on Build GRCh38
rs751334898 Chr.19: 45777758-45777758 on Build GRCh38
rs1030454205 Chr.19: 45777964-45777965 on Build GRCh38
rs201689719 Chr.19: 45777742-45777742 on Build GRCh38
rs1047593592 Chr.19: 45777898-45777898 on Build GRCh38
rs377307057 Chr.19: 45777907-45777907 on Build GRCh38
rs1006131174 Chr.19: 45777923-45777923 on Build GRCh38
rs781155031 Chr.19: 45777620-45777620 on Build GRCh38
rs758065350 Chr.19: 45777658-45777658 on Build GRCh38
rs372905696 Chr.19: 45777894-45777894 on Build GRCh38
rs555203925 Chr.19: 45777971-45777971 on Build GRCh38
rs991489635 Chr.19: 45777968-45777968 on Build GRCh38
rs751304727 Chr.19: 45777804-45777804 on Build GRCh38
rs375735536 Chr.19: 45777813-45777813 on Build GRCh38
rs754630789 Chr.19: 45777760-45777760 on Build GRCh38
rs775626849 Chr.19: 45777807-45777807 on Build GRCh38
rs954457574 Chr.19: 45777966-45777966 on Build GRCh38
rs779440487 Chr.19: 45777903-45777903 on Build GRCh38
rs749828765 Chr.19: 45777785-45777785 on Build GRCh38
rs766151412 Chr.19: 45777748-45777748 on Build GRCh38
rs528187884 Chr.19: 45777612-45777612 on Build GRCh38
rs916448731 Chr.19: 45777978-45777978 on Build GRCh38
rs953664488 Chr.19: 45777860-45777860 on Build GRCh38
rs747295381 Chr.19: 45777924-45777924 on Build GRCh38
rs755616825 Chr.19: 45777779-45777779 on Build GRCh38
rs1046236213 Chr.19: 45777746-45777746 on Build GRCh38
rs762835443 Chr.19: 45777822-45777822 on Build GRCh38
rs765166457 Chr.19: 45777730-45777730 on Build GRCh38
rs1052176549 Chr.19: 45778040-45778040 on Build GRCh38
rs772444621 Chr.19: 45778056-45778056 on Build GRCh38
rs370770361 Chr.19: 45777616-45777616 on Build GRCh38
rs774037322 Chr.19: 45777838-45777838 on Build GRCh38
rs570705950 Chr.19: 45777698-45777698 on Build GRCh38
rs747639886 Chr.19: 45777689-45777689 on Build GRCh38
rs140158944 Chr.19: 45777633-45777633 on Build GRCh38
rs767991260 Chr.19: 45777607-45777607 on Build GRCh38
rs771214913 Chr.19: 45777626-45777626 on Build GRCh38
rs927848480 Chr.19: 45778033-45778033 on Build GRCh38
rs1019132006 Chr.19: 45777879-45777879 on Build GRCh38
rs776764650 Chr.19: 45777820-45777820 on Build GRCh38
rs1015957267 Chr.19: 45777930-45777930 on Build GRCh38
rs779534958 Chr.19: 45777602-45777602 on Build GRCh38
rs779195244 Chr.19: 45777792-45777792 on Build GRCh38
rs369136836 Chr.19: 45777862-45777862 on Build GRCh38
rs557722032 Chr.19: 45778011-45778011 on Build GRCh38
rs763738699 Chr.19: 45777884-45777884 on Build GRCh38
rs376004763 Chr.19: 45777747-45777747 on Build GRCh38
rs147641100 Chr.19: 45777784-45777784 on Build GRCh38
rs372132101 Chr.19: 45777697-45777697 on Build GRCh38
rs554967270 Chr.19: 45777637-45777637 on Build GRCh38
rs764146518 Chr.19: 45777710-45777710 on Build GRCh38
rs141629958 Chr.19: 45777659-45777659 on Build GRCh38
rs577445591 Chr.19: 45778055-45778055 on Build GRCh38
rs900135585 Chr.19: 45777952-45777952 on Build GRCh38
rs768779192 Chr.19: 45777810-45777810 on Build GRCh38
rs771259599 Chr.19: 45777700-45777700 on Build GRCh38
rs988972137 Chr.19: 45777989-45777989 on Build GRCh38
rs374746286 Chr.19: 45777829-45777829 on Build GRCh38
rs141319926 Chr.19: 45777691-45777691 on Build GRCh38
rs201008625 Chr.19: 45777632-45777632 on Build GRCh38
rs1046633097 Chr.19: 45778016-45778016 on Build GRCh38
rs375336589 Chr.19: 45777709-45777709 on Build GRCh38
rs774774759 Chr.19: 45777701-45777701 on Build GRCh38
rs752743750 Chr.19: 45777640-45777640 on Build GRCh38
rs780386907 Chr.19: 45777920-45777920 on Build GRCh38
rs754651761 Chr.19: 45777675-45777675 on Build GRCh38
rs372404548 Chr.19: 45777808-45777808 on Build GRCh38
rs935074605 Chr.19: 45778038-45778038 on Build GRCh38
rs760708972 Chr.19: 45777722-45777722 on Build GRCh38
rs748856470 Chr.19: 45778042-45778042 on Build GRCh38
rs773760078 Chr.19: 45777622-45777622 on Build GRCh38
rs758092645 Chr.19: 45777601-45777601 on Build GRCh38

View Full Product Details