Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46838446-46838845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2S1
Entrez Gene ID
1175
Gene Name
adaptor related protein complex 2 sigma 1 subunit
Gene Aliases
AP17, CLAPS2, FBH3, FBHOk, HHC3
UniGene
Hs.119591
Gene Chromosome Location
Chr.19: 46838158-46850995 on Build GRCh38
Transcripts
NM_001301076.1 NM_001301078.1 NM_001301081.1 NM_004069.4 NM_021575.3 XM_011526423.2 XM_011526424.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
998494 c.342G>T p.V114V Chr.19: 46838534-46838534 on Build GRCh38
998495 c.339C>T p.V113V Chr.19: 46838537-46838537 on Build GRCh38
5856238 c.292G>A p.V98I Chr.19: 46838775-46838775 on Build GRCh38
1183129 c.268-1G>T p.? Chr.19: 46838800-46838800 on Build GRCh38
1394820 c.307C>T p.L103L Chr.19: 46838760-46838760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563483204 Chr.19: 46838639-46838639 on Build GRCh38
rs758527590 Chr.19: 46838693-46838693 on Build GRCh38
rs749954702 Chr.19: 46838511-46838511 on Build GRCh38
rs751195434 Chr.19: 46838586-46838586 on Build GRCh38
rs767303576 Chr.19: 46838588-46838588 on Build GRCh38
rs780903084 Chr.19: 46838537-46838537 on Build GRCh38
rs770400521 Chr.19: 46838773-46838773 on Build GRCh38
rs762532614 Chr.19: 46838513-46838513 on Build GRCh38
rs888089729 Chr.19: 46838682-46838682 on Build GRCh38
rs751283401 Chr.19: 46838519-46838519 on Build GRCh38
rs528395214 Chr.19: 46838797-46838797 on Build GRCh38
rs748463225 Chr.19: 46838717-46838717 on Build GRCh38
rs911290921 Chr.19: 46838598-46838598 on Build GRCh38
rs45527038 Chr.19: 46838570-46838570 on Build GRCh38
rs367711427 Chr.19: 46838732-46838732 on Build GRCh38
rs762350816 Chr.19: 46838581-46838582 on Build GRCh38
rs759178654 Chr.19: 46838788-46838788 on Build GRCh38
rs752456982 Chr.19: 46838597-46838597 on Build GRCh38
rs755173359 Chr.19: 46838707-46838707 on Build GRCh38
rs781559017 Chr.19: 46838710-46838710 on Build GRCh38
rs772355914 Chr.19: 46838557-46838557 on Build GRCh38
rs773768685 Chr.19: 46838782-46838782 on Build GRCh38
rs753056812 Chr.19: 46838819-46838819 on Build GRCh38
rs754791186 Chr.19: 46838531-46838531 on Build GRCh38
rs976603855 Chr.19: 46838606-46838606 on Build GRCh38
rs372783366 Chr.19: 46838584-46838584 on Build GRCh38
rs764760698 Chr.19: 46838498-46838498 on Build GRCh38
rs1047436145 Chr.19: 46838663-46838663 on Build GRCh38
rs777173659 Chr.19: 46838480-46838480 on Build GRCh38
rs942807609 Chr.19: 46838603-46838603 on Build GRCh38
rs780320260 Chr.19: 46838695-46838695 on Build GRCh38
rs780409762 Chr.19: 46838569-46838569 on Build GRCh38
rs532162280 Chr.19: 46838685-46838685 on Build GRCh38
rs143384246 Chr.19: 46838791-46838791 on Build GRCh38
rs779087525 Chr.19: 46838542-46838542 on Build GRCh38
rs200237364 Chr.19: 46838692-46838692 on Build GRCh38
rs968980596 Chr.19: 46838808-46838808 on Build GRCh38
rs565613118 Chr.19: 46838737-46838737 on Build GRCh38
rs202109457 Chr.19: 46838818-46838818 on Build GRCh38
rs147698652 Chr.19: 46838627-46838627 on Build GRCh38
rs762267195 Chr.19: 46838489-46838489 on Build GRCh38
rs541382293 Chr.19: 46838578-46838578 on Build GRCh38
rs772567984 Chr.19: 46838582-46838582 on Build GRCh38
rs756023462 Chr.19: 46838600-46838600 on Build GRCh38
rs1010457436 Chr.19: 46838734-46838734 on Build GRCh38
rs374245663 Chr.19: 46838577-46838577 on Build GRCh38
rs551984029 Chr.19: 46838686-46838686 on Build GRCh38
rs911045013 Chr.19: 46838617-46838617 on Build GRCh38
rs923308642 Chr.19: 46838664-46838664 on Build GRCh38
rs759154588 Chr.19: 46838587-46838587 on Build GRCh38
rs773698041 Chr.19: 46838730-46838730 on Build GRCh38
rs745961262 Chr.19: 46838551-46838551 on Build GRCh38
rs770208359 Chr.19: 46838720-46838720 on Build GRCh38
rs929876827 Chr.19: 46838662-46838662 on Build GRCh38
rs756038304 Chr.19: 46838541-46838541 on Build GRCh38
rs941012252 Chr.19: 46838684-46838684 on Build GRCh38
rs111373526 Chr.19: 46838582-46838582 on Build GRCh38
rs530162767 Chr.19: 46838604-46838604 on Build GRCh38
rs752653383 Chr.19: 46838540-46838540 on Build GRCh38

View Full Product Details