Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47319694-47320202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5AR1
Entrez Gene ID
728
Gene Name
complement component 5a receptor 1
Gene Aliases
C5A, C5AR, C5R1, CD88
UniGene
Hs.2161
Gene Chromosome Location
Chr.19: 47309847-47322070 on Build GRCh38
Transcripts
NM_001736.3 XM_005259190.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6561993 c.18T>C p.Y6Y Chr.19: 47319795-47319795 on Build GRCh38
3536290 c.120C>T p.A40A Chr.19: 47319897-47319897 on Build GRCh38
3536292 c.172G>A p.V58M Chr.19: 47319949-47319949 on Build GRCh38
73948 c.86C>T p.T29I Chr.19: 47319863-47319863 on Build GRCh38
1246868 c.141C>A p.V47V Chr.19: 47319918-47319918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189447051 Chr.19: 47319837-47319837 on Build GRCh38
rs540888456 Chr.19: 47319948-47319948 on Build GRCh38
rs780608850 Chr.19: 47319841-47319841 on Build GRCh38
rs779694046 Chr.19: 47319737-47319737 on Build GRCh38
rs199853893 Chr.19: 47319918-47319918 on Build GRCh38
rs201002949 Chr.19: 47320140-47320140 on Build GRCh38
rs200037309 Chr.19: 47319768-47319768 on Build GRCh38
rs752136515 Chr.19: 47319915-47319915 on Build GRCh38
rs946938645 Chr.19: 47319941-47319941 on Build GRCh38
rs924388393 Chr.19: 47319904-47319904 on Build GRCh38
rs140370073 Chr.19: 47319932-47319932 on Build GRCh38
rs895107917 Chr.19: 47320124-47320124 on Build GRCh38
rs758149300 Chr.19: 47320097-47320097 on Build GRCh38
rs1054010631 Chr.19: 47319910-47319910 on Build GRCh38
rs773589926 Chr.19: 47319965-47319965 on Build GRCh38
rs367862713 Chr.19: 47320044-47320044 on Build GRCh38
rs750171124 Chr.19: 47320093-47320093 on Build GRCh38
rs891257245 Chr.19: 47319840-47319840 on Build GRCh38
rs993586263 Chr.19: 47319996-47319996 on Build GRCh38
rs200581560 Chr.19: 47320065-47320065 on Build GRCh38
rs755648322 Chr.19: 47319993-47319993 on Build GRCh38
rs201037165 Chr.19: 47320040-47320040 on Build GRCh38
rs750975119 Chr.19: 47319832-47319832 on Build GRCh38
rs140039387 Chr.19: 47319923-47319923 on Build GRCh38
rs1011115887 Chr.19: 47319748-47319748 on Build GRCh38
rs750871770 Chr.19: 47319802-47319802 on Build GRCh38
rs1027623835 Chr.19: 47320020-47320020 on Build GRCh38
rs200003809 Chr.19: 47319962-47319962 on Build GRCh38
rs146067125 Chr.19: 47319901-47319901 on Build GRCh38
rs370494452 Chr.19: 47319775-47319775 on Build GRCh38
rs201523765 Chr.19: 47320013-47320013 on Build GRCh38
rs1046486771 Chr.19: 47320169-47320169 on Build GRCh38
rs1019469973 Chr.19: 47319807-47319807 on Build GRCh38
rs550245599 Chr.19: 47320071-47320071 on Build GRCh38
rs777420100 Chr.19: 47319931-47319931 on Build GRCh38
rs765672119 Chr.19: 47319801-47319801 on Build GRCh38
rs755024675 Chr.19: 47320102-47320102 on Build GRCh38
rs1032829565 Chr.19: 47319733-47319733 on Build GRCh38
rs771127357 Chr.19: 47320125-47320125 on Build GRCh38
rs776764274 Chr.19: 47319945-47319945 on Build GRCh38
rs773041435 Chr.19: 47319767-47319767 on Build GRCh38
rs769603168 Chr.19: 47319758-47319758 on Build GRCh38
rs375004252 Chr.19: 47319756-47319756 on Build GRCh38
rs200740207 Chr.19: 47319907-47319907 on Build GRCh38
rs1024931087 Chr.19: 47319744-47319744 on Build GRCh38
rs747847410 Chr.19: 47319751-47319751 on Build GRCh38
rs912744647 Chr.19: 47320037-47320037 on Build GRCh38
rs4467185 Chr.19: 47319781-47319781 on Build GRCh38
rs201084783 Chr.19: 47320078-47320078 on Build GRCh38
rs769494579 Chr.19: 47319856-47319856 on Build GRCh38
rs758286496 Chr.19: 47320034-47320034 on Build GRCh38
rs778616336 Chr.19: 47319942-47319942 on Build GRCh38
rs751509904 Chr.19: 47320101-47320101 on Build GRCh38
rs112812970 Chr.19: 47320015-47320015 on Build GRCh38
rs202146387 Chr.19: 47319943-47319943 on Build GRCh38
rs776358699 Chr.19: 47319750-47319750 on Build GRCh38
rs981720597 Chr.19: 47320135-47320135 on Build GRCh38
rs760084026 Chr.19: 47319980-47319980 on Build GRCh38
rs774370669 Chr.19: 47319877-47319877 on Build GRCh38
rs769216306 Chr.19: 47320177-47320177 on Build GRCh38
rs766830497 Chr.19: 47319971-47319971 on Build GRCh38
rs199559901 Chr.19: 47319828-47319828 on Build GRCh38
rs536711597 Chr.19: 47319990-47319990 on Build GRCh38
rs367724320 Chr.19: 47319760-47319760 on Build GRCh38
rs201817569 Chr.19: 47320141-47320141 on Build GRCh38
rs749928978 Chr.19: 47320132-47320132 on Build GRCh38
rs759458196 Chr.19: 47320122-47320122 on Build GRCh38
rs764241531 Chr.19: 47319969-47319969 on Build GRCh38
rs747815770 Chr.19: 47319850-47319850 on Build GRCh38
rs201003203 Chr.19: 47319857-47319857 on Build GRCh38
rs200942615 Chr.19: 47319963-47319963 on Build GRCh38
rs754063327 Chr.19: 47319970-47319970 on Build GRCh38
rs556451220 Chr.19: 47319726-47319726 on Build GRCh38
rs754123318 Chr.19: 47319782-47319782 on Build GRCh38
rs184513789 Chr.19: 47319731-47319731 on Build GRCh38
rs902490779 Chr.19: 47319728-47319728 on Build GRCh38
rs200429844 Chr.19: 47319774-47319774 on Build GRCh38
rs973204566 Chr.19: 47319799-47319799 on Build GRCh38
rs759019807 Chr.19: 47319736-47319736 on Build GRCh38
rs748991520 Chr.19: 47319738-47319738 on Build GRCh38
rs755697529 Chr.19: 47319927-47319927 on Build GRCh38
rs200400919 Chr.19: 47319872-47319872 on Build GRCh38
rs528096407 Chr.19: 47319944-47319944 on Build GRCh38
rs200668567 Chr.19: 47320174-47320174 on Build GRCh38
rs761979208 Chr.19: 47319800-47319800 on Build GRCh38
rs146759428 Chr.19: 47319740-47319740 on Build GRCh38
rs774711155 Chr.19: 47320059-47320059 on Build GRCh38
rs763699139 Chr.19: 47320012-47320012 on Build GRCh38
rs753400680 Chr.19: 47320081-47320081 on Build GRCh38
rs780742063 Chr.19: 47319922-47319922 on Build GRCh38
rs199837884 Chr.19: 47320058-47320058 on Build GRCh38
rs199684728 Chr.19: 47319889-47319889 on Build GRCh38
rs201964244 Chr.19: 47319916-47319916 on Build GRCh38
rs200630111 Chr.19: 47319989-47319989 on Build GRCh38
rs998506287 Chr.19: 47319721-47319721 on Build GRCh38
rs758968933 Chr.19: 47319816-47319816 on Build GRCh38
rs777997257 Chr.19: 47320103-47320103 on Build GRCh38
rs768023265 Chr.19: 47320063-47320063 on Build GRCh38
rs757665327 Chr.19: 47319730-47319730 on Build GRCh38
rs200546267 Chr.19: 47319891-47319891 on Build GRCh38
rs202194672 Chr.19: 47320014-47320014 on Build GRCh38
rs768747111 Chr.19: 47319882-47319882 on Build GRCh38
rs749509337 Chr.19: 47320045-47320045 on Build GRCh38
rs142732453 Chr.19: 47319892-47319892 on Build GRCh38
rs756895504 Chr.19: 47320089-47320089 on Build GRCh38
rs779999302 Chr.19: 47320098-47320098 on Build GRCh38
rs745712650 Chr.19: 47320027-47320027 on Build GRCh38
rs143680994 Chr.19: 47319938-47319938 on Build GRCh38
rs779868770 Chr.19: 47320036-47320036 on Build GRCh38
rs201655735 Chr.19: 47319769-47319769 on Build GRCh38
rs201947994 Chr.19: 47319979-47319979 on Build GRCh38
rs768126111 Chr.19: 47319741-47319741 on Build GRCh38
rs200066829 Chr.19: 47320171-47320171 on Build GRCh38
rs370739098 Chr.19: 47319873-47319873 on Build GRCh38
rs941662052 Chr.19: 47319785-47319785 on Build GRCh38
rs377031790 Chr.19: 47319734-47319734 on Build GRCh38
rs374799160 Chr.19: 47320178-47320178 on Build GRCh38
rs148454784 Chr.19: 47320173-47320173 on Build GRCh38
rs145650202 Chr.19: 47320092-47320092 on Build GRCh38
rs369395488 Chr.19: 47320041-47320041 on Build GRCh38
rs778753784 Chr.19: 47320026-47320026 on Build GRCh38
rs201183493 Chr.19: 47319878-47319878 on Build GRCh38
rs200095383 Chr.19: 47319896-47319896 on Build GRCh38
rs377325254 Chr.19: 47320072-47320072 on Build GRCh38
rs200289506 Chr.19: 47320021-47320021 on Build GRCh38
rs761154401 Chr.19: 47320176-47320176 on Build GRCh38

View Full Product Details