Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47745457-47745964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLTSCR2
Entrez Gene ID
29997
Gene Name
glioma tumor suppressor candidate region gene 2
Gene Aliases
PICT-1, PICT1
UniGene
Hs.421907
Gene Chromosome Location
Chr.19: 47745536-47757066 on Build GRCh38
Transcripts
NM_015710.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6041094 c.104_106delGGC p.R39delR Chr.19: 47745663-47745665 on Build GRCh38
7318669 c.19G>T p.G7C Chr.19: 47745578-47745578 on Build GRCh38
6220922 c.102G>T p.L34L Chr.19: 47745661-47745661 on Build GRCh38
1394950 c.23T>G p.V8G Chr.19: 47745582-47745582 on Build GRCh38
115754 c.47G>C p.S16T Chr.19: 47745606-47745606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939959610 Chr.19: 47745656-47745656 on Build GRCh38
rs754925914 Chr.19: 47745742-47745742 on Build GRCh38
rs541080070 Chr.19: 47745695-47745695 on Build GRCh38
rs758279843 Chr.19: 47745685-47745685 on Build GRCh38
rs891124853 Chr.19: 47745637-47745637 on Build GRCh38
rs377340728 Chr.19: 47745711-47745711 on Build GRCh38
rs368096847 Chr.19: 47745501-47745501 on Build GRCh38
rs571463636 Chr.19: 47745895-47745895 on Build GRCh38
rs752081752 Chr.19: 47745553-47745553 on Build GRCh38
rs763122158 Chr.19: 47745646-47745646 on Build GRCh38
rs557916840 Chr.19: 47745584-47745584 on Build GRCh38
rs778846533 Chr.19: 47745671-47745671 on Build GRCh38
rs370941645 Chr.19: 47745713-47745713 on Build GRCh38
rs1025115901 Chr.19: 47745575-47745575 on Build GRCh38
rs989886589 Chr.19: 47745529-47745529 on Build GRCh38
rs1025252836 Chr.19: 47745729-47745729 on Build GRCh38
rs531368956 Chr.19: 47745485-47745485 on Build GRCh38
rs766179264 Chr.19: 47745798-47745798 on Build GRCh38
rs762743448 Chr.19: 47745787-47745787 on Build GRCh38
rs949463712 Chr.19: 47745910-47745910 on Build GRCh38
rs753230193 Chr.19: 47745663-47745663 on Build GRCh38
rs986662440 Chr.19: 47745809-47745809 on Build GRCh38
rs924236840 Chr.19: 47745508-47745508 on Build GRCh38
rs746053167 Chr.19: 47745814-47745814 on Build GRCh38
rs757298179 Chr.19: 47745811-47745811 on Build GRCh38
rs375665192 Chr.19: 47745514-47745514 on Build GRCh38
rs750622962 Chr.19: 47745580-47745580 on Build GRCh38
rs935525798 Chr.19: 47745931-47745931 on Build GRCh38
rs1046545680 Chr.19: 47745920-47745920 on Build GRCh38
rs999681608 Chr.19: 47745746-47745746 on Build GRCh38
rs759002711 Chr.19: 47745516-47745516 on Build GRCh38
rs778901642 Chr.19: 47745612-47745612 on Build GRCh38
rs771024119 Chr.19: 47745749-47745749 on Build GRCh38
rs971089200 Chr.19: 47745573-47745573 on Build GRCh38
rs1013833316 Chr.19: 47745722-47745722 on Build GRCh38
rs746130777 Chr.19: 47745757-47745757 on Build GRCh38
rs569555970 Chr.19: 47745571-47745571 on Build GRCh38
rs780090942 Chr.19: 47745687-47745687 on Build GRCh38
rs921097936 Chr.19: 47745788-47745788 on Build GRCh38
rs982455672 Chr.19: 47745760-47745760 on Build GRCh38
rs529630048 Chr.19: 47745810-47745810 on Build GRCh38
rs761356503 Chr.19: 47745782-47745782 on Build GRCh38
rs916546707 Chr.19: 47745855-47745855 on Build GRCh38
rs779424808 Chr.19: 47745517-47745517 on Build GRCh38
rs780550153 Chr.19: 47745551-47745551 on Build GRCh38
rs200015322 Chr.19: 47745824-47745824 on Build GRCh38
rs376292602 Chr.19: 47745592-47745592 on Build GRCh38
rs770730864 Chr.19: 47745608-47745608 on Build GRCh38
rs749510337 Chr.19: 47745693-47745693 on Build GRCh38
rs1042439393 Chr.19: 47745659-47745659 on Build GRCh38
rs777299444 Chr.19: 47745562-47745562 on Build GRCh38
rs770656655 Chr.19: 47745567-47745567 on Build GRCh38
rs369247634 Chr.19: 47745635-47745635 on Build GRCh38
rs768980327 Chr.19: 47745778-47745778 on Build GRCh38
rs185060691 Chr.19: 47745550-47745550 on Build GRCh38
rs371377267 Chr.19: 47745556-47745556 on Build GRCh38
rs776951009 Chr.19: 47745662-47745663 on Build GRCh38
rs534027053 Chr.19: 47745596-47745596 on Build GRCh38
rs931190418 Chr.19: 47745853-47745853 on Build GRCh38
rs576263200 Chr.19: 47745657-47745657 on Build GRCh38
rs78530808 Chr.19: 47745650-47745650 on Build GRCh38
rs941159861 Chr.19: 47745578-47745578 on Build GRCh38
rs867575407 Chr.19: 47745593-47745593 on Build GRCh38
rs780929883 Chr.19: 47745806-47745806 on Build GRCh38
rs200592342 Chr.19: 47745783-47745783 on Build GRCh38
rs1054355951 Chr.19: 47745937-47745937 on Build GRCh38
rs1003841533 Chr.19: 47745928-47745928 on Build GRCh38
rs545243016 Chr.19: 47745495-47745495 on Build GRCh38
rs919891842 Chr.19: 47745848-47745848 on Build GRCh38
rs10404034 Chr.19: 47745577-47745577 on Build GRCh38
rs562807927 Chr.19: 47745479-47745479 on Build GRCh38
rs754441565 Chr.19: 47745554-47745554 on Build GRCh38
rs144346550 Chr.19: 47745668-47745668 on Build GRCh38
rs753358245 Chr.19: 47745716-47745716 on Build GRCh38
rs962724679 Chr.19: 47745936-47745936 on Build GRCh38
rs746948849 Chr.19: 47745689-47745689 on Build GRCh38
rs762122920 Chr.19: 47745576-47745576 on Build GRCh38
rs745789254 Chr.19: 47745615-47745615 on Build GRCh38
rs936778724 Chr.19: 47745845-47745845 on Build GRCh38
rs567766860 Chr.19: 47745923-47745923 on Build GRCh38
rs968169346 Chr.19: 47745544-47745544 on Build GRCh38
rs761894698 Chr.19: 47745627-47745627 on Build GRCh38
rs777529903 Chr.19: 47745545-47745545 on Build GRCh38
rs372832976 Chr.19: 47745541-47745541 on Build GRCh38
rs149422566 Chr.19: 47745618-47745618 on Build GRCh38
rs143725859 Chr.19: 47745625-47745625 on Build GRCh38
rs531876809 Chr.19: 47745829-47745829 on Build GRCh38
rs755612274 Chr.19: 47745665-47745665 on Build GRCh38
rs372294063 Chr.19: 47745520-47745520 on Build GRCh38
rs142807184 Chr.19: 47745589-47745589 on Build GRCh38
rs746449457 Chr.19: 47745518-47745518 on Build GRCh38
rs150656470 Chr.19: 47745510-47745510 on Build GRCh38
rs1042401 Chr.19: 47745607-47745607 on Build GRCh38
rs759848489 Chr.19: 47745697-47745697 on Build GRCh38
rs765506988 Chr.19: 47745548-47745548 on Build GRCh38
rs898469699 Chr.19: 47745565-47745565 on Build GRCh38
rs200112877 Chr.19: 47745682-47745682 on Build GRCh38
rs1055654013 Chr.19: 47745850-47745850 on Build GRCh38
rs754805111 Chr.19: 47745802-47745802 on Build GRCh38
rs369059053 Chr.19: 47745884-47745884 on Build GRCh38
rs766277052 Chr.19: 47745540-47745540 on Build GRCh38
rs895241965 Chr.19: 47745709-47745709 on Build GRCh38
rs751374197 Chr.19: 47745799-47745799 on Build GRCh38
rs746939560 Chr.19: 47745740-47745740 on Build GRCh38
rs746769722 Chr.19: 47745520-47745521 on Build GRCh38
rs776036516 Chr.19: 47745708-47745708 on Build GRCh38
rs534136235 Chr.19: 47745904-47745904 on Build GRCh38
rs547538337 Chr.19: 47745907-47745907 on Build GRCh38
rs200924988 Chr.19: 47745820-47745820 on Build GRCh38
rs772407067 Chr.19: 47745762-47745762 on Build GRCh38
rs1025199645 Chr.19: 47745897-47745897 on Build GRCh38
rs373099551 Chr.19: 47745720-47745720 on Build GRCh38
rs767903983 Chr.19: 47745704-47745704 on Build GRCh38
rs756919420 Chr.19: 47745669-47745669 on Build GRCh38
rs376506570 Chr.19: 47745739-47745739 on Build GRCh38
rs749408034 Chr.19: 47745566-47745566 on Build GRCh38
rs4802384 Chr.19: 47745870-47745870 on Build GRCh38
rs774520687 Chr.19: 47745756-47745756 on Build GRCh38
rs750121659 Chr.19: 47745730-47745730 on Build GRCh38
rs990840313 Chr.19: 47745854-47745854 on Build GRCh38
rs779736388 Chr.19: 47745493-47745493 on Build GRCh38
rs1012513533 Chr.19: 47745738-47745738 on Build GRCh38
rs368171673 Chr.19: 47745538-47745538 on Build GRCh38
rs761240743 Chr.19: 47745775-47745775 on Build GRCh38
rs774393641 Chr.19: 47745570-47745570 on Build GRCh38
rs61493325 Chr.19: 47745884-47745890 on Build GRCh38
rs1042391212 Chr.19: 47745828-47745828 on Build GRCh38
rs779376330 Chr.19: 47745512-47745512 on Build GRCh38
rs140003220 Chr.19: 47745531-47745531 on Build GRCh38
rs1000011170 Chr.19: 47745891-47745891 on Build GRCh38
rs567146666 Chr.19: 47745549-47745549 on Build GRCh38
rs752026650 Chr.19: 47745586-47745586 on Build GRCh38
rs754138301 Chr.19: 47745511-47745511 on Build GRCh38
rs775777313 Chr.19: 47745774-47745774 on Build GRCh38
rs201704848 Chr.19: 47745559-47745559 on Build GRCh38
rs763326567 Chr.19: 47745581-47745581 on Build GRCh38
rs756795856 Chr.19: 47745723-47745723 on Build GRCh38
rs1014198188 Chr.19: 47745881-47745881 on Build GRCh38
rs767958723 Chr.19: 47745661-47745661 on Build GRCh38
rs758226662 Chr.19: 47745731-47745731 on Build GRCh38
rs773442060 Chr.19: 47745645-47745645 on Build GRCh38
rs772348364 Chr.19: 47745830-47745830 on Build GRCh38
rs776909546 Chr.19: 47745574-47745574 on Build GRCh38
rs1019416918 Chr.19: 47745821-47745821 on Build GRCh38
rs769884573 Chr.19: 47745691-47745691 on Build GRCh38
rs6146540 Chr.19: 47745861-47745867 on Build GRCh38
rs749256313 Chr.19: 47745748-47745748 on Build GRCh38
rs201745947 Chr.19: 47745786-47745786 on Build GRCh38
rs372439824 Chr.19: 47745525-47745525 on Build GRCh38
rs757604840 Chr.19: 47745546-47745546 on Build GRCh38
rs768836011 Chr.19: 47745572-47745572 on Build GRCh38
rs771087850 Chr.19: 47745531-47745531 on Build GRCh38
rs773104170 Chr.19: 47745536-47745536 on Build GRCh38
rs780327411 Chr.19: 47745834-47745834 on Build GRCh38
rs894047871 Chr.19: 47745758-47745758 on Build GRCh38
rs760791411 Chr.19: 47745542-47745542 on Build GRCh38
rs3181870 Chr.19: 47745667-47745667 on Build GRCh38
rs375800820 Chr.19: 47745543-47745543 on Build GRCh38
rs902582461 Chr.19: 47745930-47745930 on Build GRCh38
rs192229430 Chr.19: 47745507-47745507 on Build GRCh38

View Full Product Details