Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48464501-48465009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ14
Entrez Gene ID
3770
Gene Name
potassium voltage-gated channel subfamily J member 14
Gene Aliases
IRK4, KIR2.4
UniGene
Hs.144011
Gene Chromosome Location
Chr.19: 48455707-48466110 on Build GRCh38
Transcripts
NM_013348.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4079859 c.1039C>T p.R347C Chr.19: 48464505-48464505 on Build GRCh38
7373107 c.1216G>A p.D406N Chr.19: 48464682-48464682 on Build GRCh38
4079860 c.1270C>T p.R424* Chr.19: 48464736-48464736 on Build GRCh38
7330983 c.1077G>T p.P359P Chr.19: 48464543-48464543 on Build GRCh38
5000272 c.1039C>A p.R347S Chr.19: 48464505-48464505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781352540 Chr.19: 48464736-48464736 on Build GRCh38
rs758664319 Chr.19: 48464775-48464775 on Build GRCh38
rs777727987 Chr.19: 48464535-48464535 on Build GRCh38
rs201423066 Chr.19: 48464571-48464571 on Build GRCh38
rs757138715 Chr.19: 48464542-48464542 on Build GRCh38
rs562447217 Chr.19: 48464526-48464526 on Build GRCh38
rs369152174 Chr.19: 48464809-48464809 on Build GRCh38
rs35593185 Chr.19: 48464532-48464532 on Build GRCh38
rs752666908 Chr.19: 48464807-48464807 on Build GRCh38
rs201381662 Chr.19: 48464821-48464821 on Build GRCh38
rs760390648 Chr.19: 48464679-48464679 on Build GRCh38
rs1050799044 Chr.19: 48464562-48464562 on Build GRCh38
rs752501523 Chr.19: 48464529-48464529 on Build GRCh38
rs763573793 Chr.19: 48464561-48464561 on Build GRCh38
rs990968538 Chr.19: 48464704-48464704 on Build GRCh38
rs916371267 Chr.19: 48464710-48464710 on Build GRCh38
rs774985981 Chr.19: 48464668-48464668 on Build GRCh38
rs144271715 Chr.19: 48464559-48464559 on Build GRCh38
rs374716744 Chr.19: 48464681-48464681 on Build GRCh38
rs774000705 Chr.19: 48464567-48464567 on Build GRCh38
rs915459734 Chr.19: 48464881-48464881 on Build GRCh38
rs139453142 Chr.19: 48464682-48464682 on Build GRCh38
rs890479462 Chr.19: 48464952-48464952 on Build GRCh38
rs750340467 Chr.19: 48464729-48464729 on Build GRCh38
rs754353829 Chr.19: 48464723-48464729 on Build GRCh38
rs142079550 Chr.19: 48464582-48464582 on Build GRCh38
rs151013666 Chr.19: 48464737-48464737 on Build GRCh38
rs375395301 Chr.19: 48464793-48464793 on Build GRCh38
rs779347599 Chr.19: 48464822-48464822 on Build GRCh38
rs908330806 Chr.19: 48464968-48464968 on Build GRCh38
rs543602716 Chr.19: 48464598-48464598 on Build GRCh38
rs149567009 Chr.19: 48464553-48464553 on Build GRCh38
rs757036433 Chr.19: 48464691-48464691 on Build GRCh38
rs373578741 Chr.19: 48464801-48464801 on Build GRCh38
rs138120407 Chr.19: 48464543-48464543 on Build GRCh38
rs147779902 Chr.19: 48464769-48464769 on Build GRCh38
rs778820492 Chr.19: 48464593-48464593 on Build GRCh38
rs753142015 Chr.19: 48464744-48464745 on Build GRCh38
rs116465275 Chr.19: 48464800-48464800 on Build GRCh38
rs749727205 Chr.19: 48464747-48464747 on Build GRCh38
rs949140366 Chr.19: 48464724-48464724 on Build GRCh38
rs935528330 Chr.19: 48464849-48464849 on Build GRCh38
rs535061721 Chr.19: 48464954-48464954 on Build GRCh38
rs757504462 Chr.19: 48464815-48464815 on Build GRCh38
rs754144498 Chr.19: 48464811-48464811 on Build GRCh38
rs1021141916 Chr.19: 48464661-48464661 on Build GRCh38
rs371118975 Chr.19: 48464753-48464753 on Build GRCh38
rs763901436 Chr.19: 48464579-48464579 on Build GRCh38
rs765213149 Chr.19: 48464708-48464708 on Build GRCh38
rs768359368 Chr.19: 48464770-48464770 on Build GRCh38
rs369293868 Chr.19: 48464643-48464643 on Build GRCh38
rs757193771 Chr.19: 48464585-48464585 on Build GRCh38
rs142080270 Chr.19: 48464612-48464612 on Build GRCh38
rs200428426 Chr.19: 48464778-48464778 on Build GRCh38
rs777018794 Chr.19: 48464572-48464572 on Build GRCh38
rs981843732 Chr.19: 48464827-48464827 on Build GRCh38
rs1013575371 Chr.19: 48464840-48464840 on Build GRCh38
rs150484317 Chr.19: 48464537-48464537 on Build GRCh38
rs781306557 Chr.19: 48464601-48464601 on Build GRCh38
rs773723083 Chr.19: 48464631-48464631 on Build GRCh38
rs770089449 Chr.19: 48464613-48464613 on Build GRCh38
rs140065287 Chr.19: 48464560-48464560 on Build GRCh38
rs778135213 Chr.19: 48464746-48464746 on Build GRCh38
rs562301083 Chr.19: 48464634-48464634 on Build GRCh38
rs746150051 Chr.19: 48464826-48464826 on Build GRCh38
rs75513035 Chr.19: 48464970-48464970 on Build GRCh38
rs755426284 Chr.19: 48464731-48464732 on Build GRCh38
rs765618547 Chr.19: 48464732-48464758 on Build GRCh38
rs560111886 Chr.19: 48464732-48464732 on Build GRCh38
rs750132842 Chr.19: 48464798-48464798 on Build GRCh38
rs968676239 Chr.19: 48464662-48464662 on Build GRCh38
rs780960686 Chr.19: 48464787-48464804 on Build GRCh38
rs377412381 Chr.19: 48464749-48464749 on Build GRCh38
rs1054445822 Chr.19: 48464862-48464862 on Build GRCh38
rs1050809864 Chr.19: 48464774-48464774 on Build GRCh38
rs376100200 Chr.19: 48464759-48464759 on Build GRCh38
rs557935092 Chr.19: 48464565-48464565 on Build GRCh38
rs552612830 Chr.19: 48464780-48464780 on Build GRCh38
rs144076553 Chr.19: 48464756-48464756 on Build GRCh38
rs867057466 Chr.19: 48464609-48464609 on Build GRCh38
rs774815907 Chr.19: 48464678-48464678 on Build GRCh38
rs1008999097 Chr.19: 48464789-48464789 on Build GRCh38
rs73050908 Chr.19: 48464758-48464758 on Build GRCh38
rs535271233 Chr.19: 48464895-48464895 on Build GRCh38
rs924091035 Chr.19: 48464842-48464842 on Build GRCh38
rs146272285 Chr.19: 48464675-48464675 on Build GRCh38
rs200152294 Chr.19: 48464773-48464773 on Build GRCh38
rs755684118 Chr.19: 48464578-48464578 on Build GRCh38
rs748506219 Chr.19: 48464557-48464557 on Build GRCh38
rs537566202 Chr.19: 48464591-48464591 on Build GRCh38
rs759009179 Chr.19: 48464673-48464673 on Build GRCh38
rs146487493 Chr.19: 48464806-48464806 on Build GRCh38
rs1050421834 Chr.19: 48464950-48464950 on Build GRCh38
rs186822771 Chr.19: 48464788-48464788 on Build GRCh38

View Full Product Details