Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48465564-48466070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ14
Entrez Gene ID
3770
Gene Name
potassium voltage-gated channel subfamily J member 14
Gene Aliases
IRK4, KIR2.4
UniGene
Hs.144011
Gene Chromosome Location
Chr.19: 48455707-48466110 on Build GRCh38
Transcripts
NM_013348.3

Gene Symbol
LOC105372430
Entrez Gene ID
105372430
Gene Name
uncharacterized LOC105372430
Gene Aliases
-
UniGene
Hs.736972
Gene Chromosome Location
Chr.19: 48465471-48469507 on Build GRCh38
Transcripts
NM_013348.3

SNP Detail

SNP ID Chromosome Location
rs1022497984 Chr.19: 48465896-48465896 on Build GRCh38
rs956456212 Chr.19: 48465594-48465594 on Build GRCh38
rs749978980 Chr.19: 48465670-48465670 on Build GRCh38
rs972326859 Chr.19: 48465943-48465943 on Build GRCh38
rs949627091 Chr.19: 48465614-48465614 on Build GRCh38
rs906427242 Chr.19: 48465765-48465765 on Build GRCh38
rs919239424 Chr.19: 48466008-48466008 on Build GRCh38
rs1050462924 Chr.19: 48465940-48465940 on Build GRCh38
rs545424437 Chr.19: 48465665-48465665 on Build GRCh38
rs953703473 Chr.19: 48465639-48465639 on Build GRCh38
rs375447015 Chr.19: 48465791-48465791 on Build GRCh38
rs530672799 Chr.19: 48465851-48465851 on Build GRCh38
rs919271971 Chr.19: 48465711-48465711 on Build GRCh38
rs773888935 Chr.19: 48465937-48465937 on Build GRCh38
rs113315621 Chr.19: 48465761-48465761 on Build GRCh38
rs999449261 Chr.19: 48466004-48466004 on Build GRCh38
rs78873884 Chr.19: 48465759-48465759 on Build GRCh38
rs77307854 Chr.19: 48465752-48465752 on Build GRCh38
rs1022443740 Chr.19: 48465603-48465603 on Build GRCh38
rs936425306 Chr.19: 48465588-48465588 on Build GRCh38
rs780054239 Chr.19: 48465883-48465883 on Build GRCh38
rs987074801 Chr.19: 48465749-48465749 on Build GRCh38
rs984881969 Chr.19: 48466045-48466045 on Build GRCh38
rs966626140 Chr.19: 48465990-48465990 on Build GRCh38
rs879097222 Chr.19: 48465630-48465630 on Build GRCh38
rs149803097 Chr.19: 48465653-48465653 on Build GRCh38
rs940957428 Chr.19: 48465834-48465834 on Build GRCh38
rs569989615 Chr.19: 48465880-48465880 on Build GRCh38
rs1031971082 Chr.19: 48466026-48466026 on Build GRCh38
rs539714329 Chr.19: 48465813-48465813 on Build GRCh38
rs1008045634 Chr.19: 48465959-48465959 on Build GRCh38
rs930720894 Chr.19: 48465732-48465732 on Build GRCh38
rs886724854 Chr.19: 48465622-48465622 on Build GRCh38
rs972024565 Chr.19: 48465683-48465683 on Build GRCh38
rs892151655 Chr.19: 48465824-48465824 on Build GRCh38
rs8100222 Chr.19: 48465592-48465592 on Build GRCh38
rs916371810 Chr.19: 48465790-48465790 on Build GRCh38
rs944943281 Chr.19: 48465676-48465676 on Build GRCh38
rs952170671 Chr.19: 48466025-48466025 on Build GRCh38
rs560775241 Chr.19: 48465621-48465622 on Build GRCh38
rs73050909 Chr.19: 48465818-48465818 on Build GRCh38
rs370137425 Chr.19: 48465909-48465909 on Build GRCh38
rs542322352 Chr.19: 48465908-48465908 on Build GRCh38
rs780169246 Chr.19: 48465693-48465693 on Build GRCh38
rs890566937 Chr.19: 48465941-48465941 on Build GRCh38
rs561465924 Chr.19: 48465755-48465755 on Build GRCh38
rs1010643861 Chr.19: 48465826-48465826 on Build GRCh38
rs531216116 Chr.19: 48465743-48465743 on Build GRCh38
rs990568999 Chr.19: 48465786-48465786 on Build GRCh38
rs560022474 Chr.19: 48465627-48465627 on Build GRCh38
rs989654649 Chr.19: 48465599-48465599 on Build GRCh38
rs748540327 Chr.19: 48465847-48465847 on Build GRCh38
rs899231221 Chr.19: 48465679-48465679 on Build GRCh38
rs1003431126 Chr.19: 48465779-48465779 on Build GRCh38
rs564099542 Chr.19: 48465742-48465742 on Build GRCh38
rs1019132176 Chr.19: 48465932-48465932 on Build GRCh38
rs188584392 Chr.19: 48465769-48465769 on Build GRCh38

View Full Product Details