Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49036617-49037113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGB1
Entrez Gene ID
114335
Gene Name
chorionic gonadotropin beta subunit 1
Gene Aliases
-
UniGene
Hs.659014
Gene Chromosome Location
Chr.19: 49035569-49036934 on Build GRCh38
Transcripts
NM_033377.1 NR_004383.1

Gene Symbol
SNAR-G1
Entrez Gene ID
100126780
Gene Name
small ILF3/NF90-associated RNA G1
Gene Aliases
-
UniGene
Hs.621635
Gene Chromosome Location
Chr.19: 49037020-49037147 on Build GRCh38
Transcripts
NM_033377.1 NR_004383.1

SNP Detail

SNP ID Chromosome Location
rs762305269 Chr.19: 49036668-49036668 on Build GRCh38
rs578004631 Chr.19: 49036954-49036954 on Build GRCh38
rs1006593052 Chr.19: 49036667-49036667 on Build GRCh38
rs34228596 Chr.19: 49036843-49036843 on Build GRCh38
rs34161060 Chr.19: 49036663-49036663 on Build GRCh38
rs1002813143 Chr.19: 49036922-49036922 on Build GRCh38
rs759076930 Chr.19: 49036682-49036682 on Build GRCh38
rs1035632080 Chr.19: 49036993-49036993 on Build GRCh38
rs186958845 Chr.19: 49036729-49036729 on Build GRCh38
rs112449679 Chr.19: 49037048-49037048 on Build GRCh38
rs540461564 Chr.19: 49037079-49037079 on Build GRCh38
rs377394615 Chr.19: 49036687-49036687 on Build GRCh38
rs373088178 Chr.19: 49036808-49036808 on Build GRCh38
rs532799393 Chr.19: 49037029-49037029 on Build GRCh38
rs373660013 Chr.19: 49036777-49036777 on Build GRCh38
rs979141736 Chr.19: 49037062-49037062 on Build GRCh38
rs1018028071 Chr.19: 49037040-49037041 on Build GRCh38
rs573599743 Chr.19: 49036780-49036780 on Build GRCh38
rs987996329 Chr.19: 49037052-49037052 on Build GRCh38
rs73945410 Chr.19: 49037090-49037090 on Build GRCh38
rs575677556 Chr.19: 49037068-49037068 on Build GRCh38
rs377556920 Chr.19: 49036816-49036816 on Build GRCh38
rs919268307 Chr.19: 49036936-49036936 on Build GRCh38
rs767091827 Chr.19: 49036691-49036692 on Build GRCh38
rs896667761 Chr.19: 49037028-49037028 on Build GRCh38
rs530534046 Chr.19: 49037042-49037042 on Build GRCh38
rs373044257 Chr.19: 49036844-49036844 on Build GRCh38
rs557353531 Chr.19: 49036987-49036987 on Build GRCh38
rs573432166 Chr.19: 49037075-49037075 on Build GRCh38
rs369942258 Chr.19: 49036784-49036784 on Build GRCh38
rs375233005 Chr.19: 49036773-49036773 on Build GRCh38
rs772289035 Chr.19: 49036717-49036717 on Build GRCh38
rs969506134 Chr.19: 49036852-49036852 on Build GRCh38
rs575971914 Chr.19: 49036873-49036873 on Build GRCh38
rs375604106 Chr.19: 49036681-49036681 on Build GRCh38
rs942622263 Chr.19: 49036947-49036947 on Build GRCh38
rs12460801 Chr.19: 49037009-49037009 on Build GRCh38
rs141531677 Chr.19: 49037086-49037086 on Build GRCh38
rs984787443 Chr.19: 49037011-49037011 on Build GRCh38
rs1031649274 Chr.19: 49036742-49036742 on Build GRCh38
rs746052565 Chr.19: 49036716-49036716 on Build GRCh38
rs904849332 Chr.19: 49037022-49037022 on Build GRCh38
rs769039759 Chr.19: 49036725-49036725 on Build GRCh38
rs555079477 Chr.19: 49037070-49037070 on Build GRCh38
rs887888354 Chr.19: 49037033-49037033 on Build GRCh38
rs762625218 Chr.19: 49036737-49036737 on Build GRCh38
rs571679811 Chr.19: 49036825-49036825 on Build GRCh38
rs377327911 Chr.19: 49036759-49036759 on Build GRCh38
rs200379679 Chr.19: 49037011-49037012 on Build GRCh38
rs751081848 Chr.19: 49036696-49036696 on Build GRCh38
rs766140640 Chr.19: 49036744-49036744 on Build GRCh38
rs369872271 Chr.19: 49036940-49036940 on Build GRCh38
rs551501295 Chr.19: 49036888-49036888 on Build GRCh38
rs1029551871 Chr.19: 49037047-49037047 on Build GRCh38
rs369471697 Chr.19: 49036831-49036831 on Build GRCh38
rs765647516 Chr.19: 49036731-49036731 on Build GRCh38
rs764951320 Chr.19: 49036679-49036679 on Build GRCh38
rs139185981 Chr.19: 49036905-49036905 on Build GRCh38
rs775492190 Chr.19: 49036723-49036723 on Build GRCh38
rs919258507 Chr.19: 49036869-49036869 on Build GRCh38
rs541595731 Chr.19: 49036750-49036750 on Build GRCh38
rs1057060606 Chr.19: 49036792-49036792 on Build GRCh38
rs532769722 Chr.19: 49036886-49036886 on Build GRCh38
rs538882424 Chr.19: 49037063-49037063 on Build GRCh38
rs769475713 Chr.19: 49036734-49036734 on Build GRCh38
rs960808642 Chr.19: 49036861-49036861 on Build GRCh38
rs927988907 Chr.19: 49036772-49036772 on Build GRCh38
rs1026763373 Chr.19: 49036650-49036650 on Build GRCh38
rs1057398455 Chr.19: 49036638-49036638 on Build GRCh38
rs189595614 Chr.19: 49037066-49037066 on Build GRCh38
rs781245225 Chr.19: 49036703-49036703 on Build GRCh38
rs1048265754 Chr.19: 49036876-49036876 on Build GRCh38
rs987901357 Chr.19: 49037014-49037014 on Build GRCh38
rs960872992 Chr.19: 49036996-49036996 on Build GRCh38
rs536700356 Chr.19: 49036893-49036893 on Build GRCh38
rs900948651 Chr.19: 49036882-49036882 on Build GRCh38
rs546911785 Chr.19: 49037059-49037059 on Build GRCh38
rs952061980 Chr.19: 49037010-49037010 on Build GRCh38
rs764676756 Chr.19: 49036662-49036662 on Build GRCh38
rs1048255288 Chr.19: 49036821-49036821 on Build GRCh38
rs993545872 Chr.19: 49037004-49037004 on Build GRCh38
rs548357040 Chr.19: 49036952-49036952 on Build GRCh38
rs942506736 Chr.19: 49036881-49036881 on Build GRCh38
rs35533886 Chr.19: 49036834-49036834 on Build GRCh38
rs551202632 Chr.19: 49036990-49036990 on Build GRCh38
rs972497937 Chr.19: 49036866-49036866 on Build GRCh38
rs377385500 Chr.19: 49036870-49036870 on Build GRCh38
rs929929757 Chr.19: 49036874-49036874 on Build GRCh38
rs528757170 Chr.19: 49036651-49036651 on Build GRCh38
rs550963946 Chr.19: 49037030-49037030 on Build GRCh38
rs897107347 Chr.19: 49036800-49036800 on Build GRCh38
rs377397813 Chr.19: 49037012-49037012 on Build GRCh38
rs975752180 Chr.19: 49037045-49037045 on Build GRCh38
rs1006262953 Chr.19: 49037077-49037077 on Build GRCh38
rs1030729100 Chr.19: 49037090-49037091 on Build GRCh38
rs969595549 Chr.19: 49036910-49036910 on Build GRCh38
rs769630365 Chr.19: 49036708-49036708 on Build GRCh38
rs569543184 Chr.19: 49036761-49036761 on Build GRCh38
rs1022530667 Chr.19: 49036903-49036903 on Build GRCh38
rs565436762 Chr.19: 49037025-49037025 on Build GRCh38
rs1006251902 Chr.19: 49037034-49037034 on Build GRCh38
rs766762733 Chr.19: 49036752-49036752 on Build GRCh38
rs775580246 Chr.19: 49036656-49036656 on Build GRCh38
rs182402915 Chr.19: 49036823-49036823 on Build GRCh38
rs375110238 Chr.19: 49036704-49036704 on Build GRCh38
rs555163674 Chr.19: 49036902-49036902 on Build GRCh38
rs1030646237 Chr.19: 49036841-49036841 on Build GRCh38
rs1057133643 Chr.19: 49037064-49037064 on Build GRCh38
rs900513857 Chr.19: 49037085-49037085 on Build GRCh38
rs191756795 Chr.19: 49036785-49036785 on Build GRCh38
rs562915171 Chr.19: 49037039-49037039 on Build GRCh38
rs981230197 Chr.19: 49036769-49036769 on Build GRCh38
rs928009459 Chr.19: 49036858-49036858 on Build GRCh38
rs1035176772 Chr.19: 49036925-49036925 on Build GRCh38
rs1056679048 Chr.19: 49037026-49037026 on Build GRCh38
rs561760700 Chr.19: 49036928-49036928 on Build GRCh38
rs34566775 Chr.19: 49036762-49036762 on Build GRCh38
rs34291483 Chr.19: 49036845-49036845 on Build GRCh38
rs868818970 Chr.19: 49036860-49036860 on Build GRCh38
rs534511803 Chr.19: 49037055-49037055 on Build GRCh38
rs545067814 Chr.19: 49036985-49036985 on Build GRCh38
rs1010546302 Chr.19: 49036983-49036983 on Build GRCh38
rs984023502 Chr.19: 49036945-49036945 on Build GRCh38
rs972166199 Chr.19: 49036934-49036934 on Build GRCh38
rs117204966 Chr.19: 49036946-49036946 on Build GRCh38
rs897003433 Chr.19: 49036639-49036639 on Build GRCh38
rs569421954 Chr.19: 49036890-49036890 on Build GRCh38
rs929877689 Chr.19: 49036817-49036817 on Build GRCh38
rs371968536 Chr.19: 49036766-49036766 on Build GRCh38
rs185491599 Chr.19: 49036883-49036883 on Build GRCh38
rs567478918 Chr.19: 49037049-49037049 on Build GRCh38
rs1006650939 Chr.19: 49036829-49036829 on Build GRCh38
rs761022684 Chr.19: 49036658-49036658 on Build GRCh38
rs777570430 Chr.19: 49036715-49036715 on Build GRCh38
rs751016313 Chr.19: 49036747-49036747 on Build GRCh38
rs913682038 Chr.19: 49037015-49037015 on Build GRCh38
rs10853805 Chr.19: 49036836-49036836 on Build GRCh38
rs78433317 Chr.19: 49037013-49037014 on Build GRCh38
rs1052143054 Chr.19: 49036962-49036962 on Build GRCh38
rs368649819 Chr.19: 49036641-49036641 on Build GRCh38
rs528663650 Chr.19: 49037013-49037013 on Build GRCh38
rs980898105 Chr.19: 49036853-49036853 on Build GRCh38
rs774444681 Chr.19: 49036776-49036776 on Build GRCh38
rs772093173 Chr.19: 49036654-49036654 on Build GRCh38
rs946430260 Chr.19: 49037016-49037016 on Build GRCh38
rs892154503 Chr.19: 49036963-49036963 on Build GRCh38
rs1043399401 Chr.19: 49036986-49036986 on Build GRCh38
rs113383936 Chr.19: 49036690-49036690 on Build GRCh38
rs754810554 Chr.19: 49036699-49036699 on Build GRCh38
rs182245410 Chr.19: 49037069-49037069 on Build GRCh38
rs571464318 Chr.19: 49037061-49037061 on Build GRCh38
rs755172926 Chr.19: 49037006-49037006 on Build GRCh38
rs956980716 Chr.19: 49036842-49036842 on Build GRCh38
rs933740243 Chr.19: 49036957-49036957 on Build GRCh38
rs544467426 Chr.19: 49037092-49037092 on Build GRCh38
rs900856459 Chr.19: 49036833-49036833 on Build GRCh38
rs373873835 Chr.19: 49036736-49036736 on Build GRCh38
rs761692328 Chr.19: 49036899-49036899 on Build GRCh38
rs202189266 Chr.19: 49036877-49036878 on Build GRCh38
rs540949768 Chr.19: 49037021-49037021 on Build GRCh38
rs777235070 Chr.19: 49036728-49036728 on Build GRCh38

View Full Product Details