Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 11447517-11448040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F6
Entrez Gene ID
1876
Gene Name
E2F transcription factor 6
Gene Aliases
E2F-6
UniGene
Hs.603093
Gene Chromosome Location
Chr.2: 11444375-11466177 on Build GRCh38
Transcripts
NM_001278275.1 NM_001278276.1 NM_001278277.1 NM_001278278.1 NM_198256.3 NM_212540.2 NR_103490.1 XM_017003547.1 XM_017003548.1 XM_017003549.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4507972 c.761C>T p.S254F Chr.2: 11447665-11447665 on Build GRCh38
459664 c.784G>C p.E262Q Chr.2: 11447642-11447642 on Build GRCh38
3836636 c.754G>T p.G252W Chr.2: 11447672-11447672 on Build GRCh38
3836637 c.694G>A p.D232N Chr.2: 11447732-11447732 on Build GRCh38
1250881 c.758C>G p.T253S Chr.2: 11447668-11447668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759667979 Chr.2: 11447777-11447777 on Build GRCh38
rs752149525 Chr.2: 11447820-11447820 on Build GRCh38
rs767520603 Chr.2: 11447615-11447615 on Build GRCh38
rs781047542 Chr.2: 11447658-11447658 on Build GRCh38
rs549277986 Chr.2: 11447908-11447908 on Build GRCh38
rs746681494 Chr.2: 11447576-11447576 on Build GRCh38
rs114855961 Chr.2: 11447855-11447855 on Build GRCh38
rs773174739 Chr.2: 11447687-11447687 on Build GRCh38
rs764631711 Chr.2: 11447947-11447947 on Build GRCh38
rs979455966 Chr.2: 11447979-11447979 on Build GRCh38
rs752319722 Chr.2: 11447743-11447743 on Build GRCh38
rs993756344 Chr.2: 11447709-11447709 on Build GRCh38
rs769513768 Chr.2: 11447666-11447666 on Build GRCh38
rs763177174 Chr.2: 11447605-11447605 on Build GRCh38
rs754581757 Chr.2: 11447741-11447741 on Build GRCh38
rs766335655 Chr.2: 11447794-11447794 on Build GRCh38
rs762790924 Chr.2: 11447772-11447773 on Build GRCh38
rs374415036 Chr.2: 11447793-11447793 on Build GRCh38
rs754671847 Chr.2: 11447657-11447657 on Build GRCh38
rs747061994 Chr.2: 11447770-11447770 on Build GRCh38
rs963443422 Chr.2: 11447998-11447998 on Build GRCh38
rs180877401 Chr.2: 11447584-11447584 on Build GRCh38
rs1002155129 Chr.2: 11447900-11447900 on Build GRCh38
rs562290379 Chr.2: 11447557-11447557 on Build GRCh38
rs960116334 Chr.2: 11447652-11447652 on Build GRCh38
rs772076652 Chr.2: 11447673-11447673 on Build GRCh38
rs774705345 Chr.2: 11447704-11447704 on Build GRCh38
rs778848930 Chr.2: 11447965-11447965 on Build GRCh38
rs987171892 Chr.2: 11447762-11447762 on Build GRCh38
rs143096401 Chr.2: 11447567-11447567 on Build GRCh38
rs574653171 Chr.2: 11447654-11447654 on Build GRCh38
rs755884950 Chr.2: 11447746-11447746 on Build GRCh38
rs376579949 Chr.2: 11447735-11447735 on Build GRCh38
rs763467255 Chr.2: 11447796-11447796 on Build GRCh38
rs1004937313 Chr.2: 11447659-11447659 on Build GRCh38
rs906135767 Chr.2: 11447881-11447881 on Build GRCh38
rs761954613 Chr.2: 11447788-11447788 on Build GRCh38
rs768754792 Chr.2: 11447772-11447772 on Build GRCh38
rs556079148 Chr.2: 11447639-11447639 on Build GRCh38
rs534390088 Chr.2: 11447595-11447595 on Build GRCh38
rs960791616 Chr.2: 11447936-11447936 on Build GRCh38
rs368492081 Chr.2: 11447795-11447795 on Build GRCh38
rs767708204 Chr.2: 11447821-11447821 on Build GRCh38
rs537491718 Chr.2: 11447631-11447631 on Build GRCh38
rs1020913237 Chr.2: 11447958-11447958 on Build GRCh38
rs147393250 Chr.2: 11447929-11447929 on Build GRCh38
rs780957670 Chr.2: 11447742-11447742 on Build GRCh38
rs779722761 Chr.2: 11447653-11447653 on Build GRCh38
rs770844155 Chr.2: 11447767-11447767 on Build GRCh38
rs769753812 Chr.2: 11447592-11447592 on Build GRCh38
rs1033366683 Chr.2: 11447983-11447983 on Build GRCh38
rs760894895 Chr.2: 11447628-11447628 on Build GRCh38
rs776864919 Chr.2: 11447782-11447782 on Build GRCh38
rs371114161 Chr.2: 11447768-11447768 on Build GRCh38
rs554030188 Chr.2: 11447696-11447696 on Build GRCh38
rs148210519 Chr.2: 11447603-11447603 on Build GRCh38
rs776570333 Chr.2: 11447583-11447583 on Build GRCh38
rs770887484 Chr.2: 11447672-11447672 on Build GRCh38
rs1013599065 Chr.2: 11447956-11447956 on Build GRCh38
rs776759468 Chr.2: 11447711-11447711 on Build GRCh38
rs775635341 Chr.2: 11447619-11447619 on Build GRCh38
rs1043591563 Chr.2: 11447861-11447861 on Build GRCh38
rs957298074 Chr.2: 11447997-11447997 on Build GRCh38
rs142815155 Chr.2: 11447668-11447668 on Build GRCh38
rs749176304 Chr.2: 11447757-11447757 on Build GRCh38
rs186311898 Chr.2: 11447827-11447827 on Build GRCh38
rs550137850 Chr.2: 11447542-11447542 on Build GRCh38
rs777437822 Chr.2: 11447753-11447753 on Build GRCh38
rs765543732 Chr.2: 11447732-11447732 on Build GRCh38
rs747951018 Chr.2: 11447661-11447661 on Build GRCh38
rs760977565 Chr.2: 11447917-11447917 on Build GRCh38
rs572296961 Chr.2: 11447697-11447697 on Build GRCh38
rs762198936 Chr.2: 11447724-11447724 on Build GRCh38
rs994692138 Chr.2: 11448000-11448000 on Build GRCh38
rs189688920 Chr.2: 11447581-11447581 on Build GRCh38
rs757875915 Chr.2: 11447651-11447651 on Build GRCh38
rs567983469 Chr.2: 11447930-11447930 on Build GRCh38
rs750924441 Chr.2: 11447645-11447645 on Build GRCh38
rs754190161 Chr.2: 11447636-11447636 on Build GRCh38
rs1033839189 Chr.2: 11447904-11447904 on Build GRCh38
rs935087871 Chr.2: 11447774-11447774 on Build GRCh38
rs1057140840 Chr.2: 11447926-11447926 on Build GRCh38
rs767057682 Chr.2: 11447733-11447733 on Build GRCh38
rs564608078 Chr.2: 11447854-11447854 on Build GRCh38

View Full Product Details