Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25233250-25233748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT3A
Entrez Gene ID
1788
Gene Name
DNA methyltransferase 3 alpha
Gene Aliases
DNMT3A2, M.HsaIIIA, TBRS
UniGene
Hs.515840
Gene Chromosome Location
Chr.2: 25232961-25342590 on Build GRCh38
Transcripts
NM_001320893.1 NM_022552.4 NM_153759.3 NM_175629.2 NR_135490.1 XM_005264175.4 XM_005264177.4 XM_011532662.2 XM_011532663.2 XM_011532666.2 XM_011532667.2 XM_017003526.1 XM_017003527.1

SNP Detail

SNP ID Chromosome Location
rs905459753 Chr.2: 25233400-25233400 on Build GRCh38
rs141339922 Chr.2: 25233539-25233539 on Build GRCh38
rs922303874 Chr.2: 25233290-25233290 on Build GRCh38
rs1031862233 Chr.2: 25233653-25233653 on Build GRCh38
rs35163679 Chr.2: 25233565-25233566 on Build GRCh38
rs536862337 Chr.2: 25233276-25233276 on Build GRCh38
rs1009508234 Chr.2: 25233455-25233456 on Build GRCh38
rs982444798 Chr.2: 25233720-25233720 on Build GRCh38
rs564377554 Chr.2: 25233374-25233374 on Build GRCh38
rs372931913 Chr.2: 25233362-25233362 on Build GRCh38
rs999246536 Chr.2: 25233598-25233598 on Build GRCh38
rs572153377 Chr.2: 25233275-25233275 on Build GRCh38
rs900201917 Chr.2: 25233403-25233403 on Build GRCh38
rs10538 Chr.2: 25233303-25233303 on Build GRCh38
rs980244463 Chr.2: 25233313-25233313 on Build GRCh38
rs903910244 Chr.2: 25233498-25233498 on Build GRCh38
rs188646626 Chr.2: 25233518-25233518 on Build GRCh38
rs867332343 Chr.2: 25233590-25233590 on Build GRCh38
rs759068352 Chr.2: 25233428-25233428 on Build GRCh38
rs970450351 Chr.2: 25233311-25233311 on Build GRCh38
rs1038507064 Chr.2: 25233386-25233386 on Build GRCh38
rs1031811636 Chr.2: 25233282-25233282 on Build GRCh38
rs956289289 Chr.2: 25233295-25233295 on Build GRCh38
rs938727063 Chr.2: 25233340-25233340 on Build GRCh38
rs768332317 Chr.2: 25233713-25233713 on Build GRCh38
rs577776952 Chr.2: 25233401-25233401 on Build GRCh38
rs1021478025 Chr.2: 25233463-25233463 on Build GRCh38
rs955048617 Chr.2: 25233648-25233648 on Build GRCh38
rs145421172 Chr.2: 25233301-25233301 on Build GRCh38
rs199673579 Chr.2: 25233529-25233529 on Build GRCh38
rs1051391673 Chr.2: 25233522-25233522 on Build GRCh38
rs928718784 Chr.2: 25233318-25233318 on Build GRCh38
rs1021650345 Chr.2: 25233310-25233310 on Build GRCh38
rs975070973 Chr.2: 25233288-25233288 on Build GRCh38
rs1038011155 Chr.2: 25233402-25233402 on Build GRCh38
rs890219458 Chr.2: 25233587-25233587 on Build GRCh38
rs543601250 Chr.2: 25233513-25233513 on Build GRCh38
rs1029572903 Chr.2: 25233631-25233631 on Build GRCh38

View Full Product Details