Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 31186333-31186880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN14
Entrez Gene ID
440854
Gene Name
calpain 14
Gene Aliases
-
UniGene
Hs.468059
Gene Chromosome Location
Chr.2: 31173056-31233957 on Build GRCh38
Transcripts
NM_001145122.1 NM_001321270.1 XM_011532863.2 XM_011532864.2 XM_011532865.1 XM_011532866.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1728120 c.1629C>A p.N543K Chr.2: 31186444-31186444 on Build GRCh38
6586427 c.1610A>G p.Q537R Chr.2: 31186463-31186463 on Build GRCh38
1725171 c.1628A>G p.N543S Chr.2: 31186445-31186445 on Build GRCh38
7309174 c.1628A>G p.N543S Chr.2: 31186445-31186445 on Build GRCh38
5460808 c.1613T>C p.L538P Chr.2: 31186460-31186460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917709163 Chr.2: 31186646-31186646 on Build GRCh38
rs922122303 Chr.2: 31186597-31186597 on Build GRCh38
rs142999800 Chr.2: 31186773-31186773 on Build GRCh38
rs994591308 Chr.2: 31186806-31186806 on Build GRCh38
rs572233371 Chr.2: 31186624-31186624 on Build GRCh38
rs937622191 Chr.2: 31186680-31186680 on Build GRCh38
rs558541544 Chr.2: 31186620-31186620 on Build GRCh38
rs771064055 Chr.2: 31186446-31186446 on Build GRCh38
rs183623141 Chr.2: 31186822-31186822 on Build GRCh38
rs143752754 Chr.2: 31186400-31186400 on Build GRCh38
rs955200260 Chr.2: 31186458-31186458 on Build GRCh38
rs185618621 Chr.2: 31186405-31186405 on Build GRCh38
rs145817869 Chr.2: 31186523-31186523 on Build GRCh38
rs893136440 Chr.2: 31186730-31186730 on Build GRCh38
rs113479164 Chr.2: 31186477-31186477 on Build GRCh38
rs906086304 Chr.2: 31186672-31186672 on Build GRCh38
rs1010258544 Chr.2: 31186439-31186439 on Build GRCh38
rs540851894 Chr.2: 31186650-31186650 on Build GRCh38
rs1045903136 Chr.2: 31186670-31186670 on Build GRCh38
rs769666959 Chr.2: 31186410-31186410 on Build GRCh38
rs200015647 Chr.2: 31186471-31186471 on Build GRCh38
rs755851236 Chr.2: 31186391-31186391 on Build GRCh38
rs950776793 Chr.2: 31186820-31186820 on Build GRCh38
rs983825667 Chr.2: 31186536-31186536 on Build GRCh38
rs192602949 Chr.2: 31186733-31186733 on Build GRCh38
rs770417574 Chr.2: 31186447-31186447 on Build GRCh38
rs762292339 Chr.2: 31186498-31186498 on Build GRCh38
rs371139161 Chr.2: 31186462-31186462 on Build GRCh38
rs929653114 Chr.2: 31186628-31186628 on Build GRCh38
rs891212032 Chr.2: 31186766-31186766 on Build GRCh38
rs1003390835 Chr.2: 31186444-31186444 on Build GRCh38
rs1033906628 Chr.2: 31186452-31186452 on Build GRCh38
rs1020291264 Chr.2: 31186765-31186765 on Build GRCh38
rs150924980 Chr.2: 31186792-31186792 on Build GRCh38
rs535893370 Chr.2: 31186491-31186491 on Build GRCh38
rs576627812 Chr.2: 31186696-31186696 on Build GRCh38
rs577611123 Chr.2: 31186816-31186816 on Build GRCh38
rs945069358 Chr.2: 31186644-31186644 on Build GRCh38
rs982527798 Chr.2: 31186633-31186633 on Build GRCh38
rs762563911 Chr.2: 31186841-31186841 on Build GRCh38
rs149004599 Chr.2: 31186659-31186659 on Build GRCh38
rs771820770 Chr.2: 31186485-31186485 on Build GRCh38
rs975006540 Chr.2: 31186586-31186586 on Build GRCh38
rs569266196 Chr.2: 31186553-31186553 on Build GRCh38
rs944932657 Chr.2: 31186559-31186559 on Build GRCh38
rs190442235 Chr.2: 31186711-31186711 on Build GRCh38
rs765639630 Chr.2: 31186502-31186502 on Build GRCh38
rs373281105 Chr.2: 31186758-31186758 on Build GRCh38
rs1027693579 Chr.2: 31186521-31186521 on Build GRCh38
rs543800953 Chr.2: 31186746-31186746 on Build GRCh38
rs538938235 Chr.2: 31186604-31186604 on Build GRCh38

View Full Product Details