Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38070921-38071433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP1B1
Entrez Gene ID
1545
Gene Name
cytochrome P450 family 1 subfamily B member 1
Gene Aliases
CP1B, CYPIB1, GLC3A, P4501B1
UniGene
Hs.154654
Gene Chromosome Location
Chr.2: 38067603-38076181 on Build GRCh38
Transcripts
NM_000104.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1306783 c.1099G>T p.D367Y Chr.2: 38071255-38071255 on Build GRCh38
170331 c.1420G>T p.E474* Chr.2: 38070934-38070934 on Build GRCh38
1249565 c.1102C>T p.R368C Chr.2: 38071252-38071252 on Build GRCh38
5936986 c.1198C>T p.P400S Chr.2: 38071156-38071156 on Build GRCh38
5924916 c.1220C>T p.T407I Chr.2: 38071134-38071134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775261951 Chr.2: 38071106-38071106 on Build GRCh38
rs745785396 Chr.2: 38071090-38071090 on Build GRCh38
rs775328093 Chr.2: 38071251-38071252 on Build GRCh38
rs770930904 Chr.2: 38071086-38071086 on Build GRCh38
rs759284349 Chr.2: 38071031-38071031 on Build GRCh38
rs764499798 Chr.2: 38071249-38071249 on Build GRCh38
rs761216127 Chr.2: 38071313-38071313 on Build GRCh38
rs104893629 Chr.2: 38071087-38071087 on Build GRCh38
rs768138533 Chr.2: 38071119-38071119 on Build GRCh38
rs771894894 Chr.2: 38071099-38071099 on Build GRCh38
rs745873505 Chr.2: 38071181-38071181 on Build GRCh38
rs756276196 Chr.2: 38071069-38071069 on Build GRCh38
rs760897392 Chr.2: 38071248-38071248 on Build GRCh38
rs104894979 Chr.2: 38070945-38070945 on Build GRCh38
rs745378110 Chr.2: 38071247-38071247 on Build GRCh38
rs373896870 Chr.2: 38071016-38071016 on Build GRCh38
rs573739126 Chr.2: 38071039-38071039 on Build GRCh38
rs781034244 Chr.2: 38071282-38071282 on Build GRCh38
rs776027129 Chr.2: 38071241-38071241 on Build GRCh38
rs141245683 Chr.2: 38071276-38071276 on Build GRCh38
rs940036742 Chr.2: 38071187-38071187 on Build GRCh38
rs998489422 Chr.2: 38071091-38071091 on Build GRCh38
rs759196527 Chr.2: 38070981-38070981 on Build GRCh38
rs139251365 Chr.2: 38070989-38070989 on Build GRCh38
rs749258520 Chr.2: 38071084-38071084 on Build GRCh38
rs370488779 Chr.2: 38071141-38071141 on Build GRCh38
rs772853286 Chr.2: 38071293-38071293 on Build GRCh38
rs916479886 Chr.2: 38071255-38071255 on Build GRCh38
rs4398252 Chr.2: 38071240-38071240 on Build GRCh38
rs104893622 Chr.2: 38071234-38071234 on Build GRCh38
rs56010818 Chr.2: 38071185-38071185 on Build GRCh38
rs72549378 Chr.2: 38071158-38071158 on Build GRCh38
rs369661494 Chr.2: 38071235-38071235 on Build GRCh38
rs148461905 Chr.2: 38071265-38071265 on Build GRCh38
rs768380638 Chr.2: 38071332-38071333 on Build GRCh38
rs768970224 Chr.2: 38071318-38071318 on Build GRCh38
rs750535605 Chr.2: 38071347-38071347 on Build GRCh38
rs780250973 Chr.2: 38070946-38070946 on Build GRCh38
rs149049138 Chr.2: 38071009-38071009 on Build GRCh38
rs781513633 Chr.2: 38071010-38071010 on Build GRCh38
rs71541925 Chr.2: 38071121-38071121 on Build GRCh38
rs770030868 Chr.2: 38071019-38071019 on Build GRCh38
rs776212230 Chr.2: 38071127-38071127 on Build GRCh38
rs766412697 Chr.2: 38071360-38071360 on Build GRCh38
rs370691828 Chr.2: 38071027-38071027 on Build GRCh38
rs1035845063 Chr.2: 38071150-38071150 on Build GRCh38
rs1056836 Chr.2: 38071060-38071060 on Build GRCh38
rs747324108 Chr.2: 38070948-38070948 on Build GRCh38
rs766732450 Chr.2: 38071048-38071048 on Build GRCh38
rs1031608836 Chr.2: 38071100-38071100 on Build GRCh38
rs1003079068 Chr.2: 38071108-38071108 on Build GRCh38
rs761190485 Chr.2: 38071134-38071134 on Build GRCh38
rs753971512 Chr.2: 38071171-38071171 on Build GRCh38
rs754048211 Chr.2: 38071250-38071250 on Build GRCh38
rs770799584 Chr.2: 38071029-38071029 on Build GRCh38
rs868208502 Chr.2: 38070957-38070957 on Build GRCh38
rs113974874 Chr.2: 38071050-38071050 on Build GRCh38
rs756452522 Chr.2: 38071028-38071028 on Build GRCh38
rs906925939 Chr.2: 38071273-38071273 on Build GRCh38
rs771737035 Chr.2: 38071035-38071035 on Build GRCh38
rs4986888 Chr.2: 38071026-38071026 on Build GRCh38
rs893198212 Chr.2: 38071052-38071052 on Build GRCh38
rs994749853 Chr.2: 38071378-38071378 on Build GRCh38
rs780770167 Chr.2: 38071385-38071389 on Build GRCh38
rs143977153 Chr.2: 38070956-38070956 on Build GRCh38
rs758353407 Chr.2: 38071088-38071088 on Build GRCh38
rs377049098 Chr.2: 38071024-38071024 on Build GRCh38
rs920199421 Chr.2: 38071201-38071201 on Build GRCh38
rs367993908 Chr.2: 38071400-38071400 on Build GRCh38
rs769873517 Chr.2: 38070969-38070969 on Build GRCh38
rs773249406 Chr.2: 38070971-38070971 on Build GRCh38
rs533651620 Chr.2: 38071398-38071398 on Build GRCh38
rs769250429 Chr.2: 38071217-38071217 on Build GRCh38
rs55771538 Chr.2: 38071261-38071261 on Build GRCh38
rs79204362 Chr.2: 38071251-38071251 on Build GRCh38
rs762973729 Chr.2: 38071350-38071350 on Build GRCh38
rs557905773 Chr.2: 38071204-38071204 on Build GRCh38
rs72480442 Chr.2: 38071252-38071252 on Build GRCh38
rs764779087 Chr.2: 38071135-38071135 on Build GRCh38
rs766872841 Chr.2: 38070973-38070973 on Build GRCh38
rs1024011253 Chr.2: 38071076-38071076 on Build GRCh38
rs747801707 Chr.2: 38071214-38071214 on Build GRCh38
rs113962019 Chr.2: 38071320-38071320 on Build GRCh38
rs932710268 Chr.2: 38070988-38070988 on Build GRCh38
rs869145989 Chr.2: 38071145-38071154 on Build GRCh38
rs777515179 Chr.2: 38070963-38070964 on Build GRCh38
rs72549381 Chr.2: 38071291-38071291 on Build GRCh38
rs4986887 Chr.2: 38071033-38071033 on Build GRCh38
rs55989760 Chr.2: 38071195-38071195 on Build GRCh38
rs985156509 Chr.2: 38071213-38071214 on Build GRCh38
rs751808441 Chr.2: 38070994-38070994 on Build GRCh38
rs986621621 Chr.2: 38071396-38071396 on Build GRCh38
rs373628592 Chr.2: 38071321-38071321 on Build GRCh38
rs1056837 Chr.2: 38071007-38071007 on Build GRCh38
rs72481808 Chr.2: 38071030-38071030 on Build GRCh38
rs587778873 Chr.2: 38071144-38071145 on Build GRCh38
rs758827228 Chr.2: 38071006-38071006 on Build GRCh38
rs540049009 Chr.2: 38070977-38070977 on Build GRCh38
rs367941075 Chr.2: 38071083-38071083 on Build GRCh38
rs750448258 Chr.2: 38071263-38071263 on Build GRCh38
rs766259019 Chr.2: 38070974-38070974 on Build GRCh38
rs200182794 Chr.2: 38071309-38071309 on Build GRCh38
rs777962233 Chr.2: 38071045-38071045 on Build GRCh38
rs757331407 Chr.2: 38071172-38071172 on Build GRCh38
rs775908245 Chr.2: 38071307-38071307 on Build GRCh38
rs148233007 Chr.2: 38071021-38071021 on Build GRCh38
rs4986889 Chr.2: 38070962-38070962 on Build GRCh38
rs72549373 Chr.2: 38070951-38070952 on Build GRCh38
rs72549376 Chr.2: 38071023-38071023 on Build GRCh38
rs201181935 Chr.2: 38071064-38071064 on Build GRCh38
rs28936701 Chr.2: 38070949-38070949 on Build GRCh38
rs778934619 Chr.2: 38071178-38071178 on Build GRCh38
rs56175199 Chr.2: 38071044-38071044 on Build GRCh38
rs761954795 Chr.2: 38071329-38071329 on Build GRCh38
rs775035231 Chr.2: 38071140-38071140 on Build GRCh38
rs886055997 Chr.2: 38071152-38071152 on Build GRCh38
rs768470862 Chr.2: 38071339-38071339 on Build GRCh38
rs768047511 Chr.2: 38071156-38071156 on Build GRCh38
rs947487251 Chr.2: 38071278-38071278 on Build GRCh38
rs1055330213 Chr.2: 38071244-38071244 on Build GRCh38
rs72549379 Chr.2: 38071264-38071264 on Build GRCh38
rs760132470 Chr.2: 38070992-38070992 on Build GRCh38
rs747810555 Chr.2: 38071290-38071290 on Build GRCh38
rs762767808 Chr.2: 38070972-38070972 on Build GRCh38
rs760381965 Chr.2: 38071114-38071114 on Build GRCh38
rs756363215 Chr.2: 38071164-38071164 on Build GRCh38
rs148542782 Chr.2: 38071186-38071186 on Build GRCh38
rs72481806 Chr.2: 38071159-38071159 on Build GRCh38
rs190572437 Chr.2: 38071298-38071298 on Build GRCh38
rs760289994 Chr.2: 38071043-38071043 on Build GRCh38
rs755439631 Chr.2: 38071056-38071056 on Build GRCh38
rs56305281 Chr.2: 38071218-38071218 on Build GRCh38
rs764181460 Chr.2: 38071170-38071170 on Build GRCh38
rs1800440 Chr.2: 38070996-38070996 on Build GRCh38
rs751992316 Chr.2: 38071055-38071055 on Build GRCh38
rs772830856 Chr.2: 38071222-38071222 on Build GRCh38
rs957253424 Chr.2: 38071129-38071129 on Build GRCh38
rs72549380 Chr.2: 38071278-38071290 on Build GRCh38
rs771691649 Chr.2: 38071248-38071249 on Build GRCh38
rs748355427 Chr.2: 38071018-38071018 on Build GRCh38
rs749073455 Chr.2: 38071009-38071009 on Build GRCh38

View Full Product Details