Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 41938143-41938653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf91
Entrez Gene ID
400950
Gene Name
chromosome 2 open reading frame 91
Gene Aliases
-
UniGene
Hs.738713
Gene Chromosome Location
Chr.2: 41935368-41961382 on Build GRCh38
Transcripts
NM_001242815.1 XM_017004097.1

SNP Detail

SNP ID Chromosome Location
rs190805221 Chr.2: 41938424-41938424 on Build GRCh38
rs943330809 Chr.2: 41938450-41938450 on Build GRCh38
rs1039109454 Chr.2: 41938331-41938331 on Build GRCh38
rs909646942 Chr.2: 41938216-41938216 on Build GRCh38
rs982344404 Chr.2: 41938206-41938206 on Build GRCh38
rs182352882 Chr.2: 41938440-41938440 on Build GRCh38
rs7602635 Chr.2: 41938322-41938322 on Build GRCh38
rs777805660 Chr.2: 41938602-41938602 on Build GRCh38
rs10192292 Chr.2: 41938614-41938614 on Build GRCh38
rs1039056943 Chr.2: 41938467-41938467 on Build GRCh38
rs992792952 Chr.2: 41938170-41938170 on Build GRCh38
rs963477957 Chr.2: 41938221-41938221 on Build GRCh38
rs1052931565 Chr.2: 41938354-41938354 on Build GRCh38
rs796835871 Chr.2: 41938231-41938231 on Build GRCh38
rs919463281 Chr.2: 41938277-41938277 on Build GRCh38
rs1053215710 Chr.2: 41938524-41938524 on Build GRCh38
rs959615894 Chr.2: 41938453-41938453 on Build GRCh38
rs891653714 Chr.2: 41938360-41938360 on Build GRCh38
rs936057365 Chr.2: 41938497-41938497 on Build GRCh38
rs755307332 Chr.2: 41938576-41938576 on Build GRCh38
rs576046146 Chr.2: 41938337-41938337 on Build GRCh38
rs926893062 Chr.2: 41938209-41938209 on Build GRCh38
rs7576690 Chr.2: 41938617-41938617 on Build GRCh38
rs1042511774 Chr.2: 41938274-41938274 on Build GRCh38
rs115338687 Chr.2: 41938213-41938213 on Build GRCh38
rs369270992 Chr.2: 41938224-41938224 on Build GRCh38
rs919408571 Chr.2: 41938437-41938437 on Build GRCh38
rs756225325 Chr.2: 41938599-41938599 on Build GRCh38
rs148352143 Chr.2: 41938609-41938609 on Build GRCh38
rs766073304 Chr.2: 41938167-41938167 on Build GRCh38
rs970875071 Chr.2: 41938432-41938432 on Build GRCh38
rs748326053 Chr.2: 41938592-41938592 on Build GRCh38
rs555082810 Chr.2: 41938369-41938369 on Build GRCh38
rs1007139816 Chr.2: 41938364-41938364 on Build GRCh38
rs929541480 Chr.2: 41938297-41938297 on Build GRCh38
rs945460533 Chr.2: 41938270-41938270 on Build GRCh38
rs745886049 Chr.2: 41938400-41938400 on Build GRCh38
rs574722340 Chr.2: 41938380-41938380 on Build GRCh38
rs1014568756 Chr.2: 41938367-41938367 on Build GRCh38
rs868823367 Chr.2: 41938605-41938605 on Build GRCh38
rs748851754 Chr.2: 41938350-41938350 on Build GRCh38
rs1016348812 Chr.2: 41938365-41938365 on Build GRCh38
rs925983425 Chr.2: 41938489-41938489 on Build GRCh38
rs962431568 Chr.2: 41938419-41938419 on Build GRCh38
rs768234037 Chr.2: 41938401-41938401 on Build GRCh38
rs1052053700 Chr.2: 41938314-41938314 on Build GRCh38
rs143497793 Chr.2: 41938193-41938193 on Build GRCh38
rs911865501 Chr.2: 41938319-41938319 on Build GRCh38
rs1016598438 Chr.2: 41938417-41938417 on Build GRCh38
rs988107279 Chr.2: 41938443-41938443 on Build GRCh38
rs912161185 Chr.2: 41938449-41938449 on Build GRCh38
rs1014101261 Chr.2: 41938597-41938597 on Build GRCh38
rs945437244 Chr.2: 41938509-41938509 on Build GRCh38
rs2374389 Chr.2: 41938184-41938184 on Build GRCh38
rs750824542 Chr.2: 41938569-41938569 on Build GRCh38
rs989463004 Chr.2: 41938485-41938485 on Build GRCh38
rs575532862 Chr.2: 41938263-41938263 on Build GRCh38
rs781412618 Chr.2: 41938582-41938582 on Build GRCh38

View Full Product Details