Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 58162732-58163235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCL
Entrez Gene ID
55120
Gene Name
Fanconi anemia complementation group L
Gene Aliases
FAAP43, PHF9, POG
UniGene
Hs.631890
Gene Chromosome Location
Chr.2: 58159243-58241380 on Build GRCh38
Transcripts
NM_001114636.1 NM_018062.3 XM_005264395.3 XM_005264397.4 XM_011532939.2 XM_011532940.2 XM_011532941.1 XM_011532942.2 XM_011532944.2 XM_011532945.2 XM_017004414.1 XM_017004416.1 XM_006712091.3 XM_006712092.3 XM_006712093.3 XM_017004859.1 XM_017004860.1 XM_017004861.1 XM_017004862.1 XM_017004863.1 XM_017004864.1 XM_017004865.1 XM_017004866.1

Gene Symbol
VRK2
Entrez Gene ID
7444
Gene Name
vaccinia related kinase 2
Gene Aliases
-
UniGene
Hs.715298
Gene Chromosome Location
Chr.2: 57907651-58163991 on Build GRCh38
Transcripts
NM_001114636.1 NM_018062.3 XM_005264395.3 XM_005264397.4 XM_011532939.2 XM_011532940.2 XM_011532941.1 XM_011532942.2 XM_011532944.2 XM_011532945.2 XM_017004414.1 XM_017004416.1 XM_006712091.3 XM_006712092.3 XM_006712093.3 XM_017004859.1 XM_017004860.1 XM_017004861.1 XM_017004862.1 XM_017004863.1 XM_017004864.1 XM_017004865.1 XM_017004866.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4853876 c.865G>C p.E289Q Chr.2: 58162904-58162904 on Build GRCh38
4141178 c.812T>C p.I271T Chr.2: 58163038-58163038 on Build GRCh38
6467348 c.902C>T p.S301F Chr.2: 58162867-58162867 on Build GRCh38
1021953 c.895G>A p.E299K Chr.2: 58162874-58162874 on Build GRCh38
1408754 c.836G>A p.S279N Chr.2: 58162933-58162933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557652860 Chr.2: 58163096-58163096 on Build GRCh38
rs766351502 Chr.2: 58163065-58163065 on Build GRCh38
rs960298272 Chr.2: 58163148-58163148 on Build GRCh38
rs374236117 Chr.2: 58163084-58163084 on Build GRCh38
rs751642410 Chr.2: 58163109-58163109 on Build GRCh38
rs553599985 Chr.2: 58162961-58162961 on Build GRCh38
rs779043657 Chr.2: 58162977-58162977 on Build GRCh38
rs750050199 Chr.2: 58163040-58163040 on Build GRCh38
rs766417160 Chr.2: 58163096-58163096 on Build GRCh38
rs750013831 Chr.2: 58163041-58163041 on Build GRCh38
rs758712673 Chr.2: 58163093-58163093 on Build GRCh38
rs763417767 Chr.2: 58163084-58163085 on Build GRCh38
rs748425411 Chr.2: 58162978-58162978 on Build GRCh38
rs777259158 Chr.2: 58162995-58162995 on Build GRCh38
rs935920468 Chr.2: 58162927-58162927 on Build GRCh38
rs141444545 Chr.2: 58162925-58162925 on Build GRCh38
rs543029154 Chr.2: 58163118-58163118 on Build GRCh38
rs755952682 Chr.2: 58162855-58162855 on Build GRCh38
rs369259658 Chr.2: 58162845-58162845 on Build GRCh38
rs779834767 Chr.2: 58162856-58162856 on Build GRCh38
rs1040113767 Chr.2: 58163005-58163005 on Build GRCh38
rs778413514 Chr.2: 58162985-58162985 on Build GRCh38
rs569946109 Chr.2: 58162782-58162782 on Build GRCh38
rs753690162 Chr.2: 58162969-58162969 on Build GRCh38
rs772128300 Chr.2: 58162980-58162980 on Build GRCh38
rs61757387 Chr.2: 58162909-58162909 on Build GRCh38
rs373364584 Chr.2: 58162875-58162875 on Build GRCh38
rs760163232 Chr.2: 58163063-58163063 on Build GRCh38
rs1008510137 Chr.2: 58162996-58162996 on Build GRCh38
rs749841169 Chr.2: 58162957-58162957 on Build GRCh38
rs754860957 Chr.2: 58163084-58163084 on Build GRCh38
rs376528147 Chr.2: 58162886-58162886 on Build GRCh38
rs888731378 Chr.2: 58163054-58163054 on Build GRCh38
rs775664760 Chr.2: 58163028-58163028 on Build GRCh38
rs1024412127 Chr.2: 58163146-58163146 on Build GRCh38
rs555757548 Chr.2: 58162838-58162838 on Build GRCh38
rs980090918 Chr.2: 58162757-58162757 on Build GRCh38
rs756695293 Chr.2: 58162888-58162888 on Build GRCh38
rs761047182 Chr.2: 58162817-58162817 on Build GRCh38
rs777205908 Chr.2: 58162911-58162911 on Build GRCh38
rs1034349027 Chr.2: 58162795-58162795 on Build GRCh38
rs35244714 Chr.2: 58162981-58162981 on Build GRCh38
rs535158133 Chr.2: 58162923-58162923 on Build GRCh38
rs371059417 Chr.2: 58162976-58162976 on Build GRCh38
rs773117904 Chr.2: 58162896-58162896 on Build GRCh38
rs779587713 Chr.2: 58162865-58162865 on Build GRCh38
rs777175455 Chr.2: 58162849-58162849 on Build GRCh38
rs575916313 Chr.2: 58163104-58163104 on Build GRCh38
rs761259325 Chr.2: 58162900-58162900 on Build GRCh38
rs897556023 Chr.2: 58163021-58163021 on Build GRCh38
rs199875654 Chr.2: 58162894-58162894 on Build GRCh38
rs539245901 Chr.2: 58163150-58163150 on Build GRCh38
rs770095807 Chr.2: 58163083-58163084 on Build GRCh38
rs771742741 Chr.2: 58162864-58162865 on Build GRCh38
rs201749725 Chr.2: 58163200-58163203 on Build GRCh38
rs745845830 Chr.2: 58162850-58162850 on Build GRCh38
rs763837257 Chr.2: 58162843-58162843 on Build GRCh38
rs1010330712 Chr.2: 58163011-58163011 on Build GRCh38
rs776927609 Chr.2: 58163059-58163060 on Build GRCh38
rs373237140 Chr.2: 58163116-58163116 on Build GRCh38
rs61753272 Chr.2: 58163033-58163033 on Build GRCh38
rs567636275 Chr.2: 58162839-58162839 on Build GRCh38
rs753745302 Chr.2: 58163082-58163082 on Build GRCh38
rs959083891 Chr.2: 58162799-58162799 on Build GRCh38
rs770309872 Chr.2: 58163004-58163004 on Build GRCh38
rs767072559 Chr.2: 58162935-58162935 on Build GRCh38
rs375526911 Chr.2: 58162885-58162885 on Build GRCh38
rs756047554 Chr.2: 58162960-58162960 on Build GRCh38
rs761327399 Chr.2: 58162934-58162934 on Build GRCh38
rs778782244 Chr.2: 58163088-58163088 on Build GRCh38
rs768342310 Chr.2: 58163001-58163001 on Build GRCh38
rs369702659 Chr.2: 58163110-58163110 on Build GRCh38
rs898216685 Chr.2: 58162887-58162887 on Build GRCh38
rs771626484 Chr.2: 58162986-58162986 on Build GRCh38
rs756773258 Chr.2: 58162847-58162847 on Build GRCh38
rs200895973 Chr.2: 58162997-58162997 on Build GRCh38
rs891982888 Chr.2: 58163002-58163002 on Build GRCh38
rs967436474 Chr.2: 58163184-58163184 on Build GRCh38
rs764085319 Chr.2: 58162808-58162808 on Build GRCh38
rs1047208127 Chr.2: 58163036-58163036 on Build GRCh38
rs772224853 Chr.2: 58162891-58162891 on Build GRCh38
rs570217357 Chr.2: 58162961-58162962 on Build GRCh38
rs149412402 Chr.2: 58162834-58162834 on Build GRCh38
rs771100855 Chr.2: 58163108-58163108 on Build GRCh38
rs185440106 Chr.2: 58162756-58162756 on Build GRCh38
rs992476147 Chr.2: 58162836-58162836 on Build GRCh38
rs752366828 Chr.2: 58163090-58163090 on Build GRCh38

View Full Product Details