Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 60452517-60453018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL11A
Entrez Gene ID
53335
Gene Name
B-cell CLL/lymphoma 11A
Gene Aliases
BCL11A-L, BCL11A-S, BCL11A-XL, BCL11a-M, CTIP1, EVI9, HBFQTL5, ZNF856
UniGene
Hs.370549
Gene Chromosome Location
Chr.2: 60451167-60553498 on Build GRCh38
Transcripts
NM_018014.3 NM_138559.1 XM_011532909.1 XM_011532910.1 XM_011532912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6158757 c.662C>A p.P221H Chr.2: 60452635-60452635 on Build GRCh38
3053722 c.2292G>A p.S764S Chr.2: 60452605-60452605 on Build GRCh38
6534467 c.2275G>C p.D759H Chr.2: 60452622-60452622 on Build GRCh38
6503896 c.2234C>T p.S745F Chr.2: 60452663-60452663 on Build GRCh38
3972290 c.704G>A p.R235Q Chr.2: 60452593-60452593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568594980 Chr.2: 60452990-60452990 on Build GRCh38
rs376955234 Chr.2: 60452554-60452554 on Build GRCh38
rs987569721 Chr.2: 60452797-60452797 on Build GRCh38
rs372945270 Chr.2: 60452630-60452630 on Build GRCh38
rs754200283 Chr.2: 60452541-60452541 on Build GRCh38
rs939006533 Chr.2: 60452589-60452589 on Build GRCh38
rs775483177 Chr.2: 60452561-60452561 on Build GRCh38
rs768519844 Chr.2: 60452697-60452697 on Build GRCh38
rs942086284 Chr.2: 60452897-60452916 on Build GRCh38
rs751555664 Chr.2: 60452558-60452558 on Build GRCh38
rs765467462 Chr.2: 60452643-60452643 on Build GRCh38
rs756558789 Chr.2: 60452537-60452537 on Build GRCh38
rs756792680 Chr.2: 60452656-60452656 on Build GRCh38
rs768802721 Chr.2: 60452593-60452593 on Build GRCh38
rs981376605 Chr.2: 60452930-60452930 on Build GRCh38
rs929395760 Chr.2: 60452721-60452721 on Build GRCh38
rs374828093 Chr.2: 60452945-60452945 on Build GRCh38
rs753260917 Chr.2: 60452676-60452676 on Build GRCh38
rs757675521 Chr.2: 60452705-60452706 on Build GRCh38
rs1022643055 Chr.2: 60452679-60452679 on Build GRCh38
rs761923443 Chr.2: 60452534-60452534 on Build GRCh38
rs1057196069 Chr.2: 60452723-60452723 on Build GRCh38
rs774666438 Chr.2: 60452607-60452607 on Build GRCh38
rs201277008 Chr.2: 60452610-60452610 on Build GRCh38
rs752196143 Chr.2: 60452692-60452692 on Build GRCh38
rs564609137 Chr.2: 60452642-60452642 on Build GRCh38
rs1031803390 Chr.2: 60452547-60452547 on Build GRCh38
rs954561164 Chr.2: 60452932-60452932 on Build GRCh38
rs954797218 Chr.2: 60452743-60452743 on Build GRCh38
rs755552426 Chr.2: 60452691-60452692 on Build GRCh38
rs184641807 Chr.2: 60452543-60452543 on Build GRCh38
rs919060514 Chr.2: 60452699-60452699 on Build GRCh38
rs973641222 Chr.2: 60452894-60452894 on Build GRCh38
rs113307140 Chr.2: 60452670-60452671 on Build GRCh38
rs917264287 Chr.2: 60452805-60452805 on Build GRCh38
rs954383915 Chr.2: 60452602-60452602 on Build GRCh38
rs555661121 Chr.2: 60452697-60452697 on Build GRCh38
rs754847260 Chr.2: 60452632-60452632 on Build GRCh38
rs1007223648 Chr.2: 60452993-60452993 on Build GRCh38
rs532074730 Chr.2: 60452709-60452709 on Build GRCh38
rs920884212 Chr.2: 60452922-60452922 on Build GRCh38
rs143388171 Chr.2: 60452581-60452581 on Build GRCh38
rs1044705119 Chr.2: 60452826-60452826 on Build GRCh38
rs376248698 Chr.2: 60452690-60452690 on Build GRCh38
rs771438128 Chr.2: 60452568-60452568 on Build GRCh38
rs891944804 Chr.2: 60452882-60452883 on Build GRCh38
rs112822457 Chr.2: 60452548-60452548 on Build GRCh38
rs774916881 Chr.2: 60452576-60452576 on Build GRCh38
rs1029073024 Chr.2: 60452727-60452727 on Build GRCh38
rs751233129 Chr.2: 60452646-60452646 on Build GRCh38
rs772183313 Chr.2: 60452712-60452712 on Build GRCh38
rs760251532 Chr.2: 60452841-60452841 on Build GRCh38
rs1053292593 Chr.2: 60452856-60452856 on Build GRCh38
rs749937730 Chr.2: 60452673-60452673 on Build GRCh38
rs768182495 Chr.2: 60452718-60452718 on Build GRCh38
rs904776945 Chr.2: 60452839-60452839 on Build GRCh38
rs867405159 Chr.2: 60452599-60452599 on Build GRCh38
rs939081501 Chr.2: 60452983-60452983 on Build GRCh38
rs748839477 Chr.2: 60452696-60452697 on Build GRCh38
rs764702531 Chr.2: 60452827-60452827 on Build GRCh38
rs773213278 Chr.2: 60452611-60452611 on Build GRCh38
rs948237645 Chr.2: 60452926-60452926 on Build GRCh38
rs369170414 Chr.2: 60452536-60452536 on Build GRCh38
rs767456942 Chr.2: 60452535-60452535 on Build GRCh38
rs749199891 Chr.2: 60452695-60452695 on Build GRCh38
rs761242116 Chr.2: 60452619-60452619 on Build GRCh38
rs920738352 Chr.2: 60452549-60452549 on Build GRCh38
rs756810180 Chr.2: 60452546-60452546 on Build GRCh38
rs17331129 Chr.2: 60452807-60452807 on Build GRCh38
rs551839025 Chr.2: 60452697-60452698 on Build GRCh38
rs369693637 Chr.2: 60452540-60452540 on Build GRCh38
rs778514336 Chr.2: 60452698-60452698 on Build GRCh38
rs551784376 Chr.2: 60452636-60452636 on Build GRCh38
rs1009754418 Chr.2: 60452883-60452883 on Build GRCh38
rs780820021 Chr.2: 60452658-60452658 on Build GRCh38
rs865844321 Chr.2: 60452747-60452747 on Build GRCh38
rs758107245 Chr.2: 60452542-60452542 on Build GRCh38
rs766860941 Chr.2: 60452628-60452628 on Build GRCh38

View Full Product Details