Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69960002-69960510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPRV1
Entrez Gene ID
151516
Gene Name
aspartic peptidase, retroviral-like 1
Gene Aliases
MUNO, SASP, SASPase, Taps
UniGene
Hs.516253
Gene Chromosome Location
Chr.2: 69960089-69962265 on Build GRCh38
Transcripts
NM_152792.2

SNP Detail

SNP ID Chromosome Location
rs1047561538 Chr.2: 69960115-69960115 on Build GRCh38
rs928574584 Chr.2: 69960104-69960104 on Build GRCh38
rs3821276 Chr.2: 69960041-69960041 on Build GRCh38
rs989996398 Chr.2: 69960067-69960067 on Build GRCh38
rs1055347826 Chr.2: 69960235-69960235 on Build GRCh38
rs562458602 Chr.2: 69960127-69960127 on Build GRCh38
rs375594346 Chr.2: 69960150-69960150 on Build GRCh38
rs983372062 Chr.2: 69960438-69960438 on Build GRCh38
rs1038485411 Chr.2: 69960152-69960152 on Build GRCh38
rs914299068 Chr.2: 69960035-69960035 on Build GRCh38
rs1031927558 Chr.2: 69960241-69960241 on Build GRCh38
rs576065677 Chr.2: 69960141-69960141 on Build GRCh38
rs765645434 Chr.2: 69960464-69960464 on Build GRCh38
rs891392760 Chr.2: 69960198-69960198 on Build GRCh38
rs1025393872 Chr.2: 69960283-69960283 on Build GRCh38
rs938571552 Chr.2: 69960108-69960108 on Build GRCh38
rs956269032 Chr.2: 69960292-69960292 on Build GRCh38
rs757569782 Chr.2: 69960220-69960220 on Build GRCh38
rs979752872 Chr.2: 69960385-69960385 on Build GRCh38
rs139593764 Chr.2: 69960196-69960196 on Build GRCh38
rs139791269 Chr.2: 69960349-69960349 on Build GRCh38
rs920947730 Chr.2: 69960079-69960079 on Build GRCh38
rs771936232 Chr.2: 69960183-69960183 on Build GRCh38
rs72910670 Chr.2: 69960120-69960120 on Build GRCh38
rs986520374 Chr.2: 69960433-69960433 on Build GRCh38
rs3087933 Chr.2: 69960406-69960406 on Build GRCh38
rs145600005 Chr.2: 69960330-69960330 on Build GRCh38
rs902621107 Chr.2: 69960300-69960300 on Build GRCh38
rs931573671 Chr.2: 69960214-69960214 on Build GRCh38
rs990028991 Chr.2: 69960411-69960411 on Build GRCh38
rs990710293 Chr.2: 69960309-69960309 on Build GRCh38
rs34123968 Chr.2: 69960460-69960460 on Build GRCh38
rs541517125 Chr.2: 69960184-69960184 on Build GRCh38
rs201896761 Chr.2: 69960302-69960302 on Build GRCh38
rs774147053 Chr.2: 69960060-69960060 on Build GRCh38
rs186756796 Chr.2: 69960202-69960202 on Build GRCh38
rs149739285 Chr.2: 69960215-69960215 on Build GRCh38
rs912347263 Chr.2: 69960130-69960130 on Build GRCh38
rs569224129 Chr.2: 69960423-69960423 on Build GRCh38

View Full Product Details