Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69960792-69961293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPRV1
Entrez Gene ID
151516
Gene Name
aspartic peptidase, retroviral-like 1
Gene Aliases
MUNO, SASP, SASPase, Taps
UniGene
Hs.516253
Gene Chromosome Location
Chr.2: 69960089-69962265 on Build GRCh38
Transcripts
NM_152792.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3056266 c.851T>C p.I284T Chr.2: 69960838-69960838 on Build GRCh38
3056270 c.702C>A p.D234E Chr.2: 69960987-69960987 on Build GRCh38
4553642 c.597G>A p.K199K Chr.2: 69961092-69961092 on Build GRCh38
279008 c.620T>C p.V207A Chr.2: 69961069-69961069 on Build GRCh38
6297366 c.396C>A p.I132I Chr.2: 69961293-69961293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771815800 Chr.2: 69961113-69961113 on Build GRCh38
rs755318799 Chr.2: 69960851-69960851 on Build GRCh38
rs147056246 Chr.2: 69960910-69960910 on Build GRCh38
rs751978821 Chr.2: 69960826-69960826 on Build GRCh38
rs747852584 Chr.2: 69960995-69960995 on Build GRCh38
rs754850728 Chr.2: 69961095-69961095 on Build GRCh38
rs371446435 Chr.2: 69961130-69961130 on Build GRCh38
rs200651018 Chr.2: 69961099-69961099 on Build GRCh38
rs1001647646 Chr.2: 69960942-69960942 on Build GRCh38
rs750697488 Chr.2: 69960881-69960881 on Build GRCh38
rs780295597 Chr.2: 69960845-69960845 on Build GRCh38
rs766173456 Chr.2: 69961199-69961199 on Build GRCh38
rs1014974080 Chr.2: 69961013-69961013 on Build GRCh38
rs200484241 Chr.2: 69961236-69961236 on Build GRCh38
rs372410120 Chr.2: 69960927-69960927 on Build GRCh38
rs746685284 Chr.2: 69961223-69961223 on Build GRCh38
rs568592170 Chr.2: 69961083-69961083 on Build GRCh38
rs768183966 Chr.2: 69960969-69960969 on Build GRCh38
rs746968623 Chr.2: 69960922-69960922 on Build GRCh38
rs949673516 Chr.2: 69960829-69960829 on Build GRCh38
rs763217648 Chr.2: 69961237-69961237 on Build GRCh38
rs761331152 Chr.2: 69960876-69960876 on Build GRCh38
rs773693551 Chr.2: 69960998-69960998 on Build GRCh38
rs760082473 Chr.2: 69961044-69961044 on Build GRCh38
rs144160396 Chr.2: 69961162-69961162 on Build GRCh38
rs761972154 Chr.2: 69961194-69961194 on Build GRCh38
rs763475963 Chr.2: 69961267-69961267 on Build GRCh38
rs746832214 Chr.2: 69960855-69960855 on Build GRCh38
rs144572836 Chr.2: 69960949-69960949 on Build GRCh38
rs1026634764 Chr.2: 69960991-69960991 on Build GRCh38
rs775826144 Chr.2: 69961244-69961244 on Build GRCh38
rs111819900 Chr.2: 69960934-69960934 on Build GRCh38
rs368339100 Chr.2: 69961027-69961027 on Build GRCh38
rs751747496 Chr.2: 69961152-69961152 on Build GRCh38
rs767945598 Chr.2: 69960877-69960877 on Build GRCh38
rs1008148767 Chr.2: 69960944-69960945 on Build GRCh38
rs139581474 Chr.2: 69960858-69960858 on Build GRCh38
rs752301953 Chr.2: 69961220-69961220 on Build GRCh38
rs752649063 Chr.2: 69961066-69961066 on Build GRCh38
rs779269238 Chr.2: 69960852-69960852 on Build GRCh38
rs769863291 Chr.2: 69961007-69961009 on Build GRCh38
rs762432577 Chr.2: 69960863-69960863 on Build GRCh38
rs906077985 Chr.2: 69960923-69960923 on Build GRCh38
rs747608966 Chr.2: 69961110-69961110 on Build GRCh38
rs765655077 Chr.2: 69961149-69961149 on Build GRCh38
rs750968820 Chr.2: 69960834-69960834 on Build GRCh38
rs199781771 Chr.2: 69961233-69961233 on Build GRCh38
rs775692995 Chr.2: 69961156-69961156 on Build GRCh38
rs761270468 Chr.2: 69960887-69960887 on Build GRCh38
rs773684455 Chr.2: 69960972-69960972 on Build GRCh38
rs76975752 Chr.2: 69960846-69960846 on Build GRCh38
rs764416223 Chr.2: 69961166-69961166 on Build GRCh38
rs201257906 Chr.2: 69960861-69960861 on Build GRCh38
rs138437547 Chr.2: 69960957-69960957 on Build GRCh38
rs996197429 Chr.2: 69960925-69960925 on Build GRCh38
rs777302363 Chr.2: 69961222-69961222 on Build GRCh38
rs143824935 Chr.2: 69960996-69960996 on Build GRCh38
rs760137345 Chr.2: 69961142-69961142 on Build GRCh38
rs892063213 Chr.2: 69960860-69960860 on Build GRCh38
rs963329777 Chr.2: 69961024-69961024 on Build GRCh38
rs756531444 Chr.2: 69960843-69960843 on Build GRCh38
rs142181991 Chr.2: 69961203-69961203 on Build GRCh38
rs777250815 Chr.2: 69961041-69961041 on Build GRCh38
rs913183708 Chr.2: 69961225-69961225 on Build GRCh38
rs747421482 Chr.2: 69961120-69961120 on Build GRCh38
rs769536694 Chr.2: 69961143-69961143 on Build GRCh38
rs114182672 Chr.2: 69961071-69961071 on Build GRCh38
rs753904171 Chr.2: 69960849-69960849 on Build GRCh38
rs762215216 Chr.2: 69960831-69960831 on Build GRCh38
rs1050431886 Chr.2: 69961170-69961170 on Build GRCh38
rs146425292 Chr.2: 69960898-69960898 on Build GRCh38
rs199802764 Chr.2: 69961070-69961070 on Build GRCh38
rs147879189 Chr.2: 69960962-69960962 on Build GRCh38
rs753564634 Chr.2: 69961202-69961202 on Build GRCh38
rs777566583 Chr.2: 69960859-69960859 on Build GRCh38
rs752998743 Chr.2: 69960931-69960931 on Build GRCh38
rs778702426 Chr.2: 69961204-69961204 on Build GRCh38
rs765384025 Chr.2: 69961059-69961059 on Build GRCh38
rs367637047 Chr.2: 69960832-69960832 on Build GRCh38
rs757820035 Chr.2: 69961221-69961221 on Build GRCh38
rs751811211 Chr.2: 69961182-69961182 on Build GRCh38
rs144015283 Chr.2: 69960873-69960873 on Build GRCh38
rs781731211 Chr.2: 69961231-69961231 on Build GRCh38
rs770653376 Chr.2: 69961230-69961230 on Build GRCh38
rs139245310 Chr.2: 69961207-69961207 on Build GRCh38
rs778491957 Chr.2: 69961097-69961097 on Build GRCh38
rs200429927 Chr.2: 69961132-69961132 on Build GRCh38
rs146705876 Chr.2: 69960866-69960866 on Build GRCh38
rs779783710 Chr.2: 69961081-69961081 on Build GRCh38
rs990541288 Chr.2: 69961002-69961002 on Build GRCh38
rs531721434 Chr.2: 69961034-69961034 on Build GRCh38
rs755091728 Chr.2: 69960919-69960919 on Build GRCh38
rs758489117 Chr.2: 69960933-69960933 on Build GRCh38
rs745641281 Chr.2: 69960953-69960953 on Build GRCh38
rs148341435 Chr.2: 69960909-69960909 on Build GRCh38
rs201753195 Chr.2: 69961116-69961116 on Build GRCh38
rs994280980 Chr.2: 69960984-69960984 on Build GRCh38
rs944077133 Chr.2: 69961193-69961193 on Build GRCh38
rs866249998 Chr.2: 69961056-69961056 on Build GRCh38
rs766580374 Chr.2: 69960971-69960972 on Build GRCh38

View Full Product Details