Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73257731-73258238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO41
Entrez Gene ID
150726
Gene Name
F-box protein 41
Gene Aliases
FBX41
UniGene
Hs.23158
Gene Chromosome Location
Chr.2: 73254682-73284453 on Build GRCh38
Transcripts
NM_001080410.2 XM_005264158.3 XM_006711950.2 XM_011532562.1 XM_011532563.2 XM_017003421.1 XM_017003422.1 XM_017003423.1

SNP Detail

SNP ID Chromosome Location
rs142350495 Chr.2: 73257972-73257972 on Build GRCh38
rs1031640859 Chr.2: 73258059-73258059 on Build GRCh38
rs576683967 Chr.2: 73258150-73258150 on Build GRCh38
rs906245759 Chr.2: 73258031-73258031 on Build GRCh38
rs997355491 Chr.2: 73257840-73257840 on Build GRCh38
rs536426462 Chr.2: 73258134-73258134 on Build GRCh38
rs75729698 Chr.2: 73257839-73257839 on Build GRCh38
rs965187216 Chr.2: 73257898-73257898 on Build GRCh38
rs1051575982 Chr.2: 73258046-73258046 on Build GRCh38
rs929012900 Chr.2: 73257978-73257978 on Build GRCh38
rs761243156 Chr.2: 73258107-73258107 on Build GRCh38
rs1009273597 Chr.2: 73258137-73258137 on Build GRCh38
rs559003005 Chr.2: 73258202-73258202 on Build GRCh38
rs1035842315 Chr.2: 73258164-73258164 on Build GRCh38
rs981617126 Chr.2: 73257904-73257904 on Build GRCh38
rs371085709 Chr.2: 73258088-73258088 on Build GRCh38
rs957621509 Chr.2: 73257877-73257877 on Build GRCh38
rs936569863 Chr.2: 73257976-73257976 on Build GRCh38
rs1006100136 Chr.2: 73257808-73257808 on Build GRCh38
rs982031683 Chr.2: 73258161-73258161 on Build GRCh38
rs1047449598 Chr.2: 73257980-73257980 on Build GRCh38
rs995086353 Chr.2: 73258081-73258081 on Build GRCh38
rs555269044 Chr.2: 73258145-73258145 on Build GRCh38
rs1018994519 Chr.2: 73258149-73258149 on Build GRCh38
rs548325412 Chr.2: 73257838-73257838 on Build GRCh38
rs1026766469 Chr.2: 73258094-73258094 on Build GRCh38
rs955470415 Chr.2: 73257847-73257847 on Build GRCh38
rs144590210 Chr.2: 73258015-73258015 on Build GRCh38
rs553448027 Chr.2: 73258049-73258049 on Build GRCh38
rs147839957 Chr.2: 73257857-73257857 on Build GRCh38
rs748248023 Chr.2: 73258014-73258014 on Build GRCh38
rs941667535 Chr.2: 73258012-73258012 on Build GRCh38
rs1011531027 Chr.2: 73258108-73258108 on Build GRCh38
rs1033937004 Chr.2: 73257757-73257757 on Build GRCh38
rs975235713 Chr.2: 73258201-73258201 on Build GRCh38
rs998856594 Chr.2: 73258052-73258052 on Build GRCh38
rs567060468 Chr.2: 73257956-73257956 on Build GRCh38
rs1031609708 Chr.2: 73257861-73257861 on Build GRCh38
rs532670533 Chr.2: 73257766-73257766 on Build GRCh38
rs899383847 Chr.2: 73257821-73257821 on Build GRCh38
rs1023663928 Chr.2: 73258110-73258110 on Build GRCh38
rs887710262 Chr.2: 73257798-73257798 on Build GRCh38
rs749850826 Chr.2: 73257902-73257902 on Build GRCh38
rs990329173 Chr.2: 73257888-73257888 on Build GRCh38
rs886439101 Chr.2: 73258113-73258113 on Build GRCh38
rs771645211 Chr.2: 73258169-73258169 on Build GRCh38
rs566300073 Chr.2: 73258118-73258118 on Build GRCh38
rs1030182750 Chr.2: 73257843-73257843 on Build GRCh38
rs184920102 Chr.2: 73258048-73258048 on Build GRCh38
rs970323697 Chr.2: 73258141-73258141 on Build GRCh38
rs544061982 Chr.2: 73258191-73258191 on Build GRCh38
rs570867909 Chr.2: 73258033-73258033 on Build GRCh38
rs531366427 Chr.2: 73257996-73257996 on Build GRCh38
rs916158386 Chr.2: 73257892-73257892 on Build GRCh38
rs549300706 Chr.2: 73257876-73257876 on Build GRCh38

View Full Product Details