Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73258909-73259401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO41
Entrez Gene ID
150726
Gene Name
F-box protein 41
Gene Aliases
FBX41
UniGene
Hs.23158
Gene Chromosome Location
Chr.2: 73254682-73284453 on Build GRCh38
Transcripts
NM_001080410.2 XM_005264158.3 XM_006711950.2 XM_011532562.1 XM_011532563.2 XM_017003421.1 XM_017003422.1 XM_017003423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7195689 c.2758C>T p.R920W Chr.2: 73259035-73259035 on Build GRCh38
3372865 c.2580G>A p.R860R Chr.2: 73259030-73259030 on Build GRCh38
7250900 c.2768G>C p.G923A Chr.2: 73259025-73259025 on Build GRCh38
6092376 c.2673G>A p.G891G Chr.2: 73259256-73259256 on Build GRCh38
4962286 c.2642G>A p.R881Q Chr.2: 73259287-73259287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755445450 Chr.2: 73259008-73259008 on Build GRCh38
rs557012136 Chr.2: 73259165-73259165 on Build GRCh38
rs956587579 Chr.2: 73259178-73259178 on Build GRCh38
rs745407270 Chr.2: 73259311-73259311 on Build GRCh38
rs775914052 Chr.2: 73259323-73259323 on Build GRCh38
rs775195418 Chr.2: 73258966-73258966 on Build GRCh38
rs754869381 Chr.2: 73259277-73259277 on Build GRCh38
rs369748133 Chr.2: 73259253-73259253 on Build GRCh38
rs746489249 Chr.2: 73259199-73259199 on Build GRCh38
rs778737158 Chr.2: 73259283-73259283 on Build GRCh38
rs370166126 Chr.2: 73258967-73258967 on Build GRCh38
rs559290672 Chr.2: 73259104-73259104 on Build GRCh38
rs765695065 Chr.2: 73259143-73259143 on Build GRCh38
rs766971909 Chr.2: 73259131-73259131 on Build GRCh38
rs771707604 Chr.2: 73259197-73259197 on Build GRCh38
rs764352742 Chr.2: 73259250-73259250 on Build GRCh38
rs145166005 Chr.2: 73259226-73259226 on Build GRCh38
rs542275895 Chr.2: 73259164-73259164 on Build GRCh38
rs761196272 Chr.2: 73258978-73258978 on Build GRCh38
rs952492182 Chr.2: 73259098-73259098 on Build GRCh38
rs747891713 Chr.2: 73259066-73259066 on Build GRCh38
rs774545012 Chr.2: 73259330-73259330 on Build GRCh38
rs758399047 Chr.2: 73259027-73259027 on Build GRCh38
rs149178635 Chr.2: 73259201-73259201 on Build GRCh38
rs538622406 Chr.2: 73259049-73259049 on Build GRCh38
rs749057797 Chr.2: 73259053-73259053 on Build GRCh38
rs768245332 Chr.2: 73258968-73258968 on Build GRCh38
rs757374921 Chr.2: 73259156-73259156 on Build GRCh38
rs747059282 Chr.2: 73259035-73259035 on Build GRCh38
rs767121287 Chr.2: 73258979-73258979 on Build GRCh38
rs370901292 Chr.2: 73259054-73259054 on Build GRCh38
rs779934544 Chr.2: 73259161-73259161 on Build GRCh38
rs553462213 Chr.2: 73259145-73259145 on Build GRCh38
rs373913609 Chr.2: 73259251-73259251 on Build GRCh38
rs777267477 Chr.2: 73259214-73259214 on Build GRCh38
rs760285002 Chr.2: 73259093-73259093 on Build GRCh38
rs1005115354 Chr.2: 73259336-73259336 on Build GRCh38
rs577351688 Chr.2: 73259108-73259108 on Build GRCh38
rs767731502 Chr.2: 73259346-73259346 on Build GRCh38
rs780508991 Chr.2: 73259324-73259324 on Build GRCh38
rs765528062 Chr.2: 73259025-73259025 on Build GRCh38
rs760066147 Chr.2: 73259224-73259224 on Build GRCh38
rs115044409 Chr.2: 73259149-73259149 on Build GRCh38
rs1044512636 Chr.2: 73259234-73259234 on Build GRCh38
rs959109134 Chr.2: 73258975-73258975 on Build GRCh38
rs532007287 Chr.2: 73259307-73259307 on Build GRCh38
rs750516783 Chr.2: 73259275-73259275 on Build GRCh38
rs374556639 Chr.2: 73259078-73259078 on Build GRCh38
rs768901836 Chr.2: 73259017-73259018 on Build GRCh38
rs1005066392 Chr.2: 73258981-73258981 on Build GRCh38
rs780991069 Chr.2: 73258940-73258940 on Build GRCh38
rs761177597 Chr.2: 73258991-73258991 on Build GRCh38
rs767641457 Chr.2: 73259272-73259272 on Build GRCh38
rs777475319 Chr.2: 73259292-73259292 on Build GRCh38
rs112931206 Chr.2: 73259116-73259116 on Build GRCh38
rs368143126 Chr.2: 73259293-73259293 on Build GRCh38
rs895009717 Chr.2: 73259364-73259364 on Build GRCh38
rs753033564 Chr.2: 73259026-73259026 on Build GRCh38
rs536316404 Chr.2: 73258942-73258942 on Build GRCh38
rs759978017 Chr.2: 73259141-73259141 on Build GRCh38
rs1003280425 Chr.2: 73258953-73258953 on Build GRCh38
rs752514254 Chr.2: 73259287-73259287 on Build GRCh38
rs199625082 Chr.2: 73258990-73258990 on Build GRCh38
rs367854019 Chr.2: 73259326-73259326 on Build GRCh38
rs772791259 Chr.2: 73259198-73259198 on Build GRCh38
rs138268745 Chr.2: 73258974-73258974 on Build GRCh38
rs764250946 Chr.2: 73259152-73259152 on Build GRCh38
rs779986017 Chr.2: 73259052-73259052 on Build GRCh38
rs767024986 Chr.2: 73258999-73258999 on Build GRCh38
rs771627612 Chr.2: 73259071-73259071 on Build GRCh38
rs756526388 Chr.2: 73259210-73259210 on Build GRCh38
rs922140947 Chr.2: 73259122-73259122 on Build GRCh38
rs775880863 Chr.2: 73259229-73259229 on Build GRCh38
rs370814153 Chr.2: 73259233-73259233 on Build GRCh38
rs754754460 Chr.2: 73259176-73259176 on Build GRCh38
rs770290950 Chr.2: 73259320-73259320 on Build GRCh38
rs769732247 Chr.2: 73258961-73258961 on Build GRCh38
rs995905907 Chr.2: 73259000-73259000 on Build GRCh38
rs758165731 Chr.2: 73259291-73259291 on Build GRCh38
rs560927594 Chr.2: 73259182-73259182 on Build GRCh38
rs761965966 Chr.2: 73259343-73259343 on Build GRCh38
rs754207965 Chr.2: 73259006-73259006 on Build GRCh38
rs200807832 Chr.2: 73259030-73259030 on Build GRCh38
rs770716961 Chr.2: 73259299-73259299 on Build GRCh38
rs899361007 Chr.2: 73259353-73259353 on Build GRCh38

View Full Product Details