Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73260448-73260913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO41
Entrez Gene ID
150726
Gene Name
F-box protein 41
Gene Aliases
FBX41
UniGene
Hs.23158
Gene Chromosome Location
Chr.2: 73254682-73284453 on Build GRCh38
Transcripts
NM_001080410.2 XM_005264158.3 XM_006711950.2 XM_011532562.1 XM_011532563.2 XM_017003421.1 XM_017003422.1 XM_017003423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4436424 c.2402G>A p.R801H Chr.2: 73260811-73260811 on Build GRCh38
6433185 c.2275G>T p.G759C Chr.2: 73260755-73260755 on Build GRCh38
4095584 c.2305C>T p.R769W Chr.2: 73260533-73260533 on Build GRCh38
4813463 c.2557G>A p.E853K Chr.2: 73260464-73260464 on Build GRCh38
4565557 c.2411_2412CC>TA p.P804L Chr.2: 73260801-73260802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056824440 Chr.2: 73260720-73260720 on Build GRCh38
rs748480982 Chr.2: 73260495-73260495 on Build GRCh38
rs776626029 Chr.2: 73260868-73260868 on Build GRCh38
rs755400177 Chr.2: 73260486-73260486 on Build GRCh38
rs759679772 Chr.2: 73260534-73260534 on Build GRCh38
rs1022956831 Chr.2: 73260714-73260714 on Build GRCh38
rs753265961 Chr.2: 73260592-73260592 on Build GRCh38
rs372882768 Chr.2: 73260741-73260741 on Build GRCh38
rs369498847 Chr.2: 73260485-73260485 on Build GRCh38
rs988287669 Chr.2: 73260874-73260874 on Build GRCh38
rs62149575 Chr.2: 73260797-73260797 on Build GRCh38
rs912156717 Chr.2: 73260844-73260844 on Build GRCh38
rs767480644 Chr.2: 73260558-73260558 on Build GRCh38
rs913657923 Chr.2: 73260550-73260550 on Build GRCh38
rs771005973 Chr.2: 73260774-73260774 on Build GRCh38
rs995300045 Chr.2: 73260789-73260789 on Build GRCh38
rs757685558 Chr.2: 73260875-73260875 on Build GRCh38
rs771186356 Chr.2: 73260513-73260513 on Build GRCh38
rs758916516 Chr.2: 73260747-73260747 on Build GRCh38
rs772490678 Chr.2: 73260500-73260500 on Build GRCh38
rs765916084 Chr.2: 73260832-73260832 on Build GRCh38
rs960106859 Chr.2: 73260596-73260596 on Build GRCh38
rs781269654 Chr.2: 73260876-73260876 on Build GRCh38
rs761699672 Chr.2: 73260547-73260547 on Build GRCh38
rs752206156 Chr.2: 73260837-73260837 on Build GRCh38
rs766532299 Chr.2: 73260623-73260623 on Build GRCh38
rs772844439 Chr.2: 73260813-73260813 on Build GRCh38
rs763484568 Chr.2: 73260525-73260525 on Build GRCh38
rs375233061 Chr.2: 73260564-73260564 on Build GRCh38
rs766086813 Chr.2: 73260569-73260569 on Build GRCh38
rs569031158 Chr.2: 73260770-73260770 on Build GRCh38
rs758859272 Chr.2: 73260861-73260861 on Build GRCh38
rs148018926 Chr.2: 73260507-73260507 on Build GRCh38
rs745908703 Chr.2: 73260780-73260780 on Build GRCh38
rs770068903 Chr.2: 73260783-73260783 on Build GRCh38
rs779445655 Chr.2: 73260489-73260489 on Build GRCh38
rs760541730 Chr.2: 73260822-73260822 on Build GRCh38
rs755638807 Chr.2: 73260478-73260478 on Build GRCh38
rs776965262 Chr.2: 73260779-73260779 on Build GRCh38
rs761785764 Chr.2: 73260788-73260788 on Build GRCh38
rs369615563 Chr.2: 73260567-73260567 on Build GRCh38
rs776530331 Chr.2: 73260524-73260524 on Build GRCh38
rs568400647 Chr.2: 73260546-73260546 on Build GRCh38
rs1027724234 Chr.2: 73260794-73260794 on Build GRCh38
rs772141532 Chr.2: 73260803-73260803 on Build GRCh38
rs904886434 Chr.2: 73260601-73260601 on Build GRCh38
rs764668422 Chr.2: 73260835-73260835 on Build GRCh38
rs755363542 Chr.2: 73260833-73260833 on Build GRCh38
rs764105116 Chr.2: 73260539-73260539 on Build GRCh38
rs999040170 Chr.2: 73260557-73260557 on Build GRCh38
rs746153629 Chr.2: 73260881-73260881 on Build GRCh38
rs535822841 Chr.2: 73260667-73260667 on Build GRCh38
rs377270722 Chr.2: 73260483-73260483 on Build GRCh38
rs778363298 Chr.2: 73260862-73260862 on Build GRCh38
rs557202458 Chr.2: 73260718-73260718 on Build GRCh38
rs761966696 Chr.2: 73260879-73260879 on Build GRCh38
rs774419963 Chr.2: 73260785-73260785 on Build GRCh38
rs773407794 Chr.2: 73260506-73260506 on Build GRCh38
rs865871335 Chr.2: 73260533-73260533 on Build GRCh38
rs896395283 Chr.2: 73260729-73260729 on Build GRCh38
rs868335347 Chr.2: 73260793-73260793 on Build GRCh38
rs754521790 Chr.2: 73260690-73260690 on Build GRCh38
rs373617282 Chr.2: 73260873-73260873 on Build GRCh38
rs747428399 Chr.2: 73260771-73260771 on Build GRCh38
rs372169842 Chr.2: 73260882-73260882 on Build GRCh38
rs962137864 Chr.2: 73260782-73260782 on Build GRCh38

View Full Product Details