Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96144070-96144564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP2
Entrez Gene ID
1844
Gene Name
dual specificity phosphatase 2
Gene Aliases
PAC-1, PAC1
UniGene
Hs.1183
Gene Chromosome Location
Chr.2: 96143166-96145468 on Build GRCh38
Transcripts
NM_004418.3 XM_017003546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1284665 c.684G>T p.Q228H Chr.2: 96144200-96144200 on Build GRCh38
1291549 c.722G>A p.G241D Chr.2: 96144162-96144162 on Build GRCh38
5418358 c.555C>T p.C185C Chr.2: 96144329-96144329 on Build GRCh38
4968389 c.661A>T p.S221C Chr.2: 96144223-96144223 on Build GRCh38
6206680 c.651C>T p.F217F Chr.2: 96144233-96144233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750087780 Chr.2: 96144350-96144350 on Build GRCh38
rs888503932 Chr.2: 96144093-96144093 on Build GRCh38
rs764502826 Chr.2: 96144261-96144261 on Build GRCh38
rs773604439 Chr.2: 96144167-96144167 on Build GRCh38
rs745737498 Chr.2: 96144298-96144306 on Build GRCh38
rs201412020 Chr.2: 96144280-96144280 on Build GRCh38
rs771854789 Chr.2: 96144309-96144309 on Build GRCh38
rs541334714 Chr.2: 96144452-96144452 on Build GRCh38
rs370042908 Chr.2: 96144393-96144393 on Build GRCh38
rs768679007 Chr.2: 96144417-96144417 on Build GRCh38
rs907856081 Chr.2: 96144449-96144449 on Build GRCh38
rs775535169 Chr.2: 96144326-96144326 on Build GRCh38
rs780828125 Chr.2: 96144205-96144205 on Build GRCh38
rs779589878 Chr.2: 96144282-96144282 on Build GRCh38
rs983558187 Chr.2: 96144431-96144431 on Build GRCh38
rs1033196477 Chr.2: 96144209-96144209 on Build GRCh38
rs556235439 Chr.2: 96144249-96144249 on Build GRCh38
rs976622932 Chr.2: 96144461-96144461 on Build GRCh38
rs903412229 Chr.2: 96144444-96144444 on Build GRCh38
rs201028960 Chr.2: 96144248-96144248 on Build GRCh38
rs144192490 Chr.2: 96144231-96144231 on Build GRCh38
rs746076346 Chr.2: 96144321-96144321 on Build GRCh38
rs762195420 Chr.2: 96144344-96144344 on Build GRCh38
rs749292287 Chr.2: 96144293-96144293 on Build GRCh38
rs375737797 Chr.2: 96144354-96144354 on Build GRCh38
rs529392486 Chr.2: 96144527-96144527 on Build GRCh38
rs775901984 Chr.2: 96144253-96144253 on Build GRCh38
rs929910250 Chr.2: 96144388-96144388 on Build GRCh38
rs576247409 Chr.2: 96144332-96144332 on Build GRCh38
rs771694150 Chr.2: 96144327-96144328 on Build GRCh38
rs199945308 Chr.2: 96144191-96144191 on Build GRCh38
rs999088239 Chr.2: 96144468-96144468 on Build GRCh38
rs369390154 Chr.2: 96144281-96144281 on Build GRCh38
rs372344461 Chr.2: 96144238-96144238 on Build GRCh38
rs758707947 Chr.2: 96144125-96144125 on Build GRCh38
rs749202296 Chr.2: 96144229-96144229 on Build GRCh38
rs369325506 Chr.2: 96144333-96144333 on Build GRCh38
rs369831290 Chr.2: 96144299-96144299 on Build GRCh38
rs745878412 Chr.2: 96144144-96144144 on Build GRCh38
rs375163568 Chr.2: 96144097-96144097 on Build GRCh38
rs765869354 Chr.2: 96144353-96144353 on Build GRCh38
rs753164081 Chr.2: 96144157-96144157 on Build GRCh38
rs922492414 Chr.2: 96144380-96144380 on Build GRCh38
rs922543482 Chr.2: 96144483-96144483 on Build GRCh38
rs760953286 Chr.2: 96144170-96144170 on Build GRCh38
rs963742224 Chr.2: 96144307-96144307 on Build GRCh38
rs776753272 Chr.2: 96144319-96144319 on Build GRCh38
rs767892413 Chr.2: 96144348-96144348 on Build GRCh38
rs780547246 Chr.2: 96144142-96144142 on Build GRCh38
rs746625044 Chr.2: 96144139-96144139 on Build GRCh38
rs759596126 Chr.2: 96144187-96144187 on Build GRCh38
rs749389431 Chr.2: 96144415-96144415 on Build GRCh38
rs1033289591 Chr.2: 96144496-96144496 on Build GRCh38
rs770469798 Chr.2: 96144141-96144141 on Build GRCh38
rs751263132 Chr.2: 96144197-96144197 on Build GRCh38
rs1010565498 Chr.2: 96144303-96144303 on Build GRCh38
rs368110350 Chr.2: 96144136-96144136 on Build GRCh38
rs932501477 Chr.2: 96144533-96144533 on Build GRCh38
rs185121318 Chr.2: 96144537-96144537 on Build GRCh38
rs778571583 Chr.2: 96144161-96144167 on Build GRCh38
rs779404624 Chr.2: 96144107-96144107 on Build GRCh38
rs184661278 Chr.2: 96144233-96144233 on Build GRCh38
rs755773859 Chr.2: 96144352-96144352 on Build GRCh38
rs932503803 Chr.2: 96144263-96144263 on Build GRCh38
rs762677764 Chr.2: 96144156-96144156 on Build GRCh38
rs774790294 Chr.2: 96144337-96144337 on Build GRCh38
rs754959106 Chr.2: 96144298-96144298 on Build GRCh38
rs1014217323 Chr.2: 96144426-96144426 on Build GRCh38
rs548942138 Chr.2: 96144535-96144535 on Build GRCh38
rs769813572 Chr.2: 96144323-96144323 on Build GRCh38
rs900982752 Chr.2: 96144240-96144240 on Build GRCh38
rs533153952 Chr.2: 96144454-96144454 on Build GRCh38
rs775439306 Chr.2: 96144145-96144145 on Build GRCh38
rs957551028 Chr.2: 96144515-96144515 on Build GRCh38
rs952192391 Chr.2: 96144423-96144423 on Build GRCh38
rs143940850 Chr.2: 96144193-96144193 on Build GRCh38
rs145177272 Chr.2: 96144453-96144453 on Build GRCh38
rs372766743 Chr.2: 96144305-96144305 on Build GRCh38
rs202170773 Chr.2: 96144222-96144222 on Build GRCh38
rs201829031 Chr.2: 96144411-96144411 on Build GRCh38
rs753010881 Chr.2: 96144119-96144119 on Build GRCh38
rs549966683 Chr.2: 96144460-96144460 on Build GRCh38
rs369633913 Chr.2: 96144418-96144418 on Build GRCh38
rs1034872970 Chr.2: 96144124-96144124 on Build GRCh38
rs555380866 Chr.2: 96144180-96144180 on Build GRCh38
rs1005653932 Chr.2: 96144103-96144103 on Build GRCh38
rs747298849 Chr.2: 96144232-96144232 on Build GRCh38
rs745381899 Chr.2: 96144217-96144217 on Build GRCh38
rs757557678 Chr.2: 96144266-96144266 on Build GRCh38
rs763429869 Chr.2: 96144255-96144255 on Build GRCh38
rs756891334 Chr.2: 96144199-96144199 on Build GRCh38
rs766621091 Chr.2: 96144176-96144176 on Build GRCh38
rs758793748 Chr.2: 96144192-96144192 on Build GRCh38
rs140138990 Chr.2: 96144369-96144369 on Build GRCh38
rs1045773897 Chr.2: 96144427-96144427 on Build GRCh38
rs920699225 Chr.2: 96144101-96144101 on Build GRCh38
rs745721382 Chr.2: 96144120-96144120 on Build GRCh38
rs977557781 Chr.2: 96144206-96144206 on Build GRCh38
rs777430150 Chr.2: 96144129-96144129 on Build GRCh38
rs566028992 Chr.2: 96144190-96144190 on Build GRCh38
rs751919546 Chr.2: 96144265-96144265 on Build GRCh38
rs780185750 Chr.2: 96144414-96144414 on Build GRCh38
rs767097357 Chr.2: 96144271-96144271 on Build GRCh38
rs200368289 Chr.2: 96144400-96144400 on Build GRCh38
rs180817110 Chr.2: 96144503-96144503 on Build GRCh38
rs190041400 Chr.2: 96144389-96144389 on Build GRCh38

View Full Product Details